Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17261774-17262131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS3
Entrez Gene ID
8533
Gene Name
COP9 signalosome subunit 3
Gene Aliases
CSN3, SGN3
UniGene
Hs.6076
Gene Chromosome Location
Chr.17: 17246624-17281303 on Build GRCh38
Transcripts
NM_001199125.1 NM_001316354.1 NM_001316355.1 NM_001316356.1 NM_001316357.1 NM_001316358.1 NM_003653.3 XM_005256837.4 XM_005256840.3 XM_005256842.4 XM_017025246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325050 c.680A>T p.Y227F Chr.17: 17262048-17262048 on Build GRCh38
6634083 c.636T>G p.P212P Chr.17: 17262092-17262092 on Build GRCh38
976097 c.677A>C p.K226T Chr.17: 17262051-17262051 on Build GRCh38
976096 c.679T>C p.Y227H Chr.17: 17262049-17262049 on Build GRCh38
1381212 c.713T>C p.V238A Chr.17: 17262015-17262015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191538354 Chr.17: 17261903-17261903 on Build GRCh38
rs898424641 Chr.17: 17261912-17261912 on Build GRCh38
rs557665110 Chr.17: 17261897-17261897 on Build GRCh38
rs894998782 Chr.17: 17261877-17261877 on Build GRCh38
rs756555950 Chr.17: 17261928-17261928 on Build GRCh38
rs774496026 Chr.17: 17261953-17261953 on Build GRCh38
rs765139424 Chr.17: 17262028-17262028 on Build GRCh38
rs15316 Chr.17: 17262020-17262020 on Build GRCh38
rs1018137120 Chr.17: 17261851-17261851 on Build GRCh38
rs374574802 Chr.17: 17262083-17262083 on Build GRCh38
rs147736279 Chr.17: 17261921-17261921 on Build GRCh38
rs754353492 Chr.17: 17262084-17262084 on Build GRCh38
rs780429852 Chr.17: 17261997-17261998 on Build GRCh38
rs939137520 Chr.17: 17261870-17261870 on Build GRCh38
rs1008757997 Chr.17: 17261952-17261952 on Build GRCh38
rs757565947 Chr.17: 17262088-17262088 on Build GRCh38
rs376677984 Chr.17: 17261996-17261996 on Build GRCh38
rs776776336 Chr.17: 17262016-17262016 on Build GRCh38
rs957850826 Chr.17: 17261822-17261825 on Build GRCh38
rs1031928897 Chr.17: 17261810-17261810 on Build GRCh38
rs1042043703 Chr.17: 17261861-17261861 on Build GRCh38
rs771285669 Chr.17: 17261944-17261944 on Build GRCh38
rs372718839 Chr.17: 17261935-17261935 on Build GRCh38
rs909873859 Chr.17: 17261906-17261906 on Build GRCh38
rs923601187 Chr.17: 17261869-17261869 on Build GRCh38
rs750054179 Chr.17: 17262045-17262045 on Build GRCh38
rs775403442 Chr.17: 17261998-17261998 on Build GRCh38
rs751020203 Chr.17: 17262074-17262074 on Build GRCh38
rs965605899 Chr.17: 17261853-17261853 on Build GRCh38
rs746108127 Chr.17: 17261991-17261991 on Build GRCh38
rs778402150 Chr.17: 17261934-17261934 on Build GRCh38
rs1056196969 Chr.17: 17261873-17261873 on Build GRCh38
rs914708402 Chr.17: 17262091-17262091 on Build GRCh38
rs200577855 Chr.17: 17262070-17262070 on Build GRCh38
rs375456935 Chr.17: 17261917-17261917 on Build GRCh38
rs763929534 Chr.17: 17262002-17262002 on Build GRCh38
rs1736210 Chr.17: 17261822-17261822 on Build GRCh38
rs918515017 Chr.17: 17261864-17261864 on Build GRCh38
rs779284568 Chr.17: 17262103-17262103 on Build GRCh38
rs537774731 Chr.17: 17261882-17261882 on Build GRCh38
rs751874540 Chr.17: 17261999-17262000 on Build GRCh38
rs760861624 Chr.17: 17262000-17262000 on Build GRCh38
rs758091601 Chr.17: 17262056-17262056 on Build GRCh38
rs950279466 Chr.17: 17261957-17261957 on Build GRCh38

View Full Product Details