Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47975177-47975775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP3
Entrez Gene ID
80279
Gene Name
CDK5 regulatory subunit associated protein 3
Gene Aliases
C53, HSF-27, IC53, LZAP, MST016, OK/SW-cl.114, PP1553
UniGene
Hs.20157
Gene Chromosome Location
Chr.17: 47967904-47981786 on Build GRCh38
Transcripts
NM_001278197.1 NM_001278198.1 NM_001278216.1 NM_001278217.1 NM_176096.2 XM_011525297.1 XM_017025164.1 XM_017025165.1 XM_017025166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691654 c.571G>T p.A191S Chr.17: 47975571-47975571 on Build GRCh38
6625577 c.477G>A p.E159E Chr.17: 47975301-47975301 on Build GRCh38
5136940 c.581G>T p.G194V Chr.17: 47975581-47975581 on Build GRCh38
6625575 c.401C>T p.A134V Chr.17: 47975225-47975225 on Build GRCh38
5055549 c.587C>T p.A196V Chr.17: 47975587-47975587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754607032 Chr.17: 47975592-47975594 on Build GRCh38
rs1050376002 Chr.17: 47975263-47975263 on Build GRCh38
rs780584998 Chr.17: 47975578-47975578 on Build GRCh38
rs573174070 Chr.17: 47975539-47975539 on Build GRCh38
rs768462849 Chr.17: 47975684-47975684 on Build GRCh38
rs762702255 Chr.17: 47975677-47975677 on Build GRCh38
rs1040468747 Chr.17: 47975446-47975446 on Build GRCh38
rs746469111 Chr.17: 47975476-47975476 on Build GRCh38
rs1055630106 Chr.17: 47975525-47975525 on Build GRCh38
rs35494079 Chr.17: 47975601-47975602 on Build GRCh38
rs764290238 Chr.17: 47975244-47975244 on Build GRCh38
rs986117308 Chr.17: 47975419-47975419 on Build GRCh38
rs774592142 Chr.17: 47975499-47975499 on Build GRCh38
rs772196862 Chr.17: 47975597-47975597 on Build GRCh38
rs750283829 Chr.17: 47975258-47975258 on Build GRCh38
rs1045509285 Chr.17: 47975679-47975679 on Build GRCh38
rs374807113 Chr.17: 47975257-47975257 on Build GRCh38
rs912434412 Chr.17: 47975612-47975612 on Build GRCh38
rs140443377 Chr.17: 47975587-47975587 on Build GRCh38
rs919295888 Chr.17: 47975472-47975472 on Build GRCh38
rs778639980 Chr.17: 47975297-47975297 on Build GRCh38
rs551145994 Chr.17: 47975449-47975449 on Build GRCh38
rs201728980 Chr.17: 47975535-47975535 on Build GRCh38
rs748108750 Chr.17: 47975343-47975343 on Build GRCh38
rs12944318 Chr.17: 47975388-47975388 on Build GRCh38
rs1051705805 Chr.17: 47975213-47975213 on Build GRCh38
rs187620011 Chr.17: 47975396-47975396 on Build GRCh38
rs544321871 Chr.17: 47975601-47975601 on Build GRCh38
rs1002646692 Chr.17: 47975347-47975347 on Build GRCh38
rs763213122 Chr.17: 47975238-47975238 on Build GRCh38
rs898006583 Chr.17: 47975307-47975307 on Build GRCh38
rs765344081 Chr.17: 47975261-47975263 on Build GRCh38
rs779758702 Chr.17: 47975266-47975266 on Build GRCh38
rs752487753 Chr.17: 47975556-47975556 on Build GRCh38
rs761997333 Chr.17: 47975334-47975334 on Build GRCh38
rs751198396 Chr.17: 47975619-47975619 on Build GRCh38
rs758392060 Chr.17: 47975527-47975529 on Build GRCh38
rs369343510 Chr.17: 47975516-47975516 on Build GRCh38
rs766139944 Chr.17: 47975636-47975636 on Build GRCh38
rs970996439 Chr.17: 47975346-47975346 on Build GRCh38
rs1056062359 Chr.17: 47975644-47975644 on Build GRCh38
rs752366442 Chr.17: 47975381-47975381 on Build GRCh38
rs760546731 Chr.17: 47975348-47975348 on Build GRCh38
rs747108207 Chr.17: 47975600-47975600 on Build GRCh38
rs201230966 Chr.17: 47975296-47975296 on Build GRCh38
rs187828907 Chr.17: 47975660-47975660 on Build GRCh38
rs777168836 Chr.17: 47975465-47975465 on Build GRCh38
rs768983329 Chr.17: 47975496-47975496 on Build GRCh38
rs755434127 Chr.17: 47975586-47975586 on Build GRCh38
rs776812925 Chr.17: 47975607-47975607 on Build GRCh38
rs778868247 Chr.17: 47975318-47975318 on Build GRCh38
rs914522105 Chr.17: 47975403-47975403 on Build GRCh38
rs772252708 Chr.17: 47975592-47975592 on Build GRCh38
rs780684229 Chr.17: 47975635-47975636 on Build GRCh38
rs559597619 Chr.17: 47975743-47975743 on Build GRCh38
rs776171691 Chr.17: 47975734-47975734 on Build GRCh38
rs781738132 Chr.17: 47975313-47975313 on Build GRCh38
rs751707212 Chr.17: 47975253-47975253 on Build GRCh38
rs750359983 Chr.17: 47975379-47975379 on Build GRCh38
rs776945659 Chr.17: 47975203-47975203 on Build GRCh38
rs989628016 Chr.17: 47975426-47975426 on Build GRCh38
rs745875058 Chr.17: 47975662-47975662 on Build GRCh38
rs748344769 Chr.17: 47975507-47975507 on Build GRCh38
rs61741124 Chr.17: 47975515-47975515 on Build GRCh38
rs777898276 Chr.17: 47975647-47975647 on Build GRCh38
rs955909345 Chr.17: 47975374-47975374 on Build GRCh38
rs911908875 Chr.17: 47975494-47975494 on Build GRCh38
rs753201423 Chr.17: 47975719-47975719 on Build GRCh38
rs61741125 Chr.17: 47975517-47975517 on Build GRCh38
rs770014353 Chr.17: 47975209-47975209 on Build GRCh38
rs894254257 Chr.17: 47975526-47975526 on Build GRCh38
rs755483548 Chr.17: 47975631-47975631 on Build GRCh38
rs776688459 Chr.17: 47975531-47975531 on Build GRCh38
rs759568675 Chr.17: 47975609-47975609 on Build GRCh38
rs368081823 Chr.17: 47975288-47975288 on Build GRCh38
rs987463916 Chr.17: 47975459-47975459 on Build GRCh38
rs183342259 Chr.17: 47975588-47975588 on Build GRCh38
rs749707963 Chr.17: 47975495-47975495 on Build GRCh38
rs200358823 Chr.17: 47975336-47975336 on Build GRCh38
rs1021394163 Chr.17: 47975595-47975595 on Build GRCh38
rs369855838 Chr.17: 47975555-47975555 on Build GRCh38
rs1044661840 Chr.17: 47975704-47975704 on Build GRCh38
rs767040825 Chr.17: 47975629-47975629 on Build GRCh38
rs747654549 Chr.17: 47975200-47975200 on Build GRCh38
rs372048386 Chr.17: 47975289-47975289 on Build GRCh38
rs911939889 Chr.17: 47975602-47975602 on Build GRCh38
rs753063656 Chr.17: 47975643-47975643 on Build GRCh38
rs747674614 Chr.17: 47975302-47975302 on Build GRCh38
rs374055919 Chr.17: 47975648-47975648 on Build GRCh38
rs779473195 Chr.17: 47975634-47975634 on Build GRCh38
rs749650948 Chr.17: 47975324-47975324 on Build GRCh38
rs756775349 Chr.17: 47975562-47975562 on Build GRCh38
rs766275276 Chr.17: 47975353-47975353 on Build GRCh38
rs367948107 Chr.17: 47975638-47975638 on Build GRCh38
rs368510697 Chr.17: 47975322-47975322 on Build GRCh38
rs370792872 Chr.17: 47975560-47975560 on Build GRCh38
rs746402949 Chr.17: 47975315-47975315 on Build GRCh38
rs1013969439 Chr.17: 47975338-47975338 on Build GRCh38
rs370149330 Chr.17: 47975630-47975630 on Build GRCh38
rs1022626228 Chr.17: 47975342-47975342 on Build GRCh38
rs373053211 Chr.17: 47975561-47975561 on Build GRCh38
rs761778927 Chr.17: 47975252-47975252 on Build GRCh38
rs902900471 Chr.17: 47975724-47975724 on Build GRCh38
rs769916668 Chr.17: 47975670-47975670 on Build GRCh38
rs192054881 Chr.17: 47975444-47975444 on Build GRCh38
rs747873025 Chr.17: 47975672-47975672 on Build GRCh38
rs768873249 Chr.17: 47975331-47975331 on Build GRCh38
rs371908146 Chr.17: 47975337-47975337 on Build GRCh38
rs766484127 Chr.17: 47975527-47975527 on Build GRCh38
rs775802632 Chr.17: 47975234-47975234 on Build GRCh38
rs953949268 Chr.17: 47975404-47975404 on Build GRCh38
rs747388090 Chr.17: 47975363-47975363 on Build GRCh38
rs114871157 Chr.17: 47975359-47975359 on Build GRCh38
rs757634684 Chr.17: 47975364-47975364 on Build GRCh38
rs761348862 Chr.17: 47975621-47975621 on Build GRCh38
rs754308503 Chr.17: 47975583-47975583 on Build GRCh38
rs556847315 Chr.17: 47975613-47975613 on Build GRCh38
rs771472549 Chr.17: 47975202-47975202 on Build GRCh38
rs758769916 Chr.17: 47975645-47975645 on Build GRCh38
rs752683425 Chr.17: 47975537-47975540 on Build GRCh38
rs369286052 Chr.17: 47975254-47975254 on Build GRCh38
rs2597172 Chr.17: 47975427-47975427 on Build GRCh38
rs976486770 Chr.17: 47975624-47975624 on Build GRCh38
rs775482336 Chr.17: 47975672-47975672 on Build GRCh38
rs780532066 Chr.17: 47975480-47975480 on Build GRCh38
rs528069332 Chr.17: 47975205-47975205 on Build GRCh38
rs375023997 Chr.17: 47975611-47975611 on Build GRCh38
rs200176581 Chr.17: 47975618-47975618 on Build GRCh38
rs958306121 Chr.17: 47975565-47975565 on Build GRCh38
rs751383513 Chr.17: 47975588-47975588 on Build GRCh38
rs377557038 Chr.17: 47975571-47975571 on Build GRCh38
rs781434034 Chr.17: 47975661-47975661 on Build GRCh38
rs757797602 Chr.17: 47975308-47975308 on Build GRCh38
rs756580769 Chr.17: 47975477-47975477 on Build GRCh38

View Full Product Details