Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 59435202-59435729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCBE1
Entrez Gene ID
147372
Gene Name
collagen and calcium binding EGF domains 1
Gene Aliases
HKLLS1
UniGene
Hs.34333
Gene Chromosome Location
Chr.18: 59430939-59698181 on Build GRCh38
Transcripts
NM_133459.3 XM_017025556.1 XM_017025557.1

SNP Detail

SNP ID Chromosome Location
rs901846951 Chr.18: 59435239-59435239 on Build GRCh38
rs1029494083 Chr.18: 59435359-59435359 on Build GRCh38
rs923238907 Chr.18: 59435573-59435573 on Build GRCh38
rs937375572 Chr.18: 59435603-59435603 on Build GRCh38
rs558281381 Chr.18: 59435548-59435548 on Build GRCh38
rs887164633 Chr.18: 59435286-59435286 on Build GRCh38
rs529266103 Chr.18: 59435705-59435705 on Build GRCh38
rs548222123 Chr.18: 59435454-59435454 on Build GRCh38
rs532612240 Chr.18: 59435292-59435292 on Build GRCh38
rs886054071 Chr.18: 59435601-59435602 on Build GRCh38
rs971047847 Chr.18: 59435437-59435437 on Build GRCh38
rs551011131 Chr.18: 59435298-59435298 on Build GRCh38
rs909291474 Chr.18: 59435531-59435531 on Build GRCh38
rs193200245 Chr.18: 59435419-59435419 on Build GRCh38
rs35018840 Chr.18: 59435388-59435389 on Build GRCh38
rs777333567 Chr.18: 59435539-59435539 on Build GRCh38
rs895322664 Chr.18: 59435698-59435698 on Build GRCh38
rs902794023 Chr.18: 59435666-59435666 on Build GRCh38
rs1003676029 Chr.18: 59435258-59435258 on Build GRCh38
rs997228345 Chr.18: 59435598-59435598 on Build GRCh38
rs1038855053 Chr.18: 59435558-59435558 on Build GRCh38
rs561328038 Chr.18: 59435641-59435641 on Build GRCh38
rs941578080 Chr.18: 59435544-59435544 on Build GRCh38
rs977195637 Chr.18: 59435362-59435362 on Build GRCh38
rs1029631176 Chr.18: 59435636-59435636 on Build GRCh38
rs566748160 Chr.18: 59435478-59435478 on Build GRCh38
rs943393485 Chr.18: 59435564-59435564 on Build GRCh38
rs748705977 Chr.18: 59435325-59435325 on Build GRCh38
rs555760247 Chr.18: 59435685-59435685 on Build GRCh38
rs906793571 Chr.18: 59435253-59435253 on Build GRCh38
rs991743220 Chr.18: 59435606-59435606 on Build GRCh38
rs887255868 Chr.18: 59435536-59435536 on Build GRCh38
rs902656511 Chr.18: 59435310-59435310 on Build GRCh38
rs1047281804 Chr.18: 59435279-59435279 on Build GRCh38
rs886054072 Chr.18: 59435647-59435647 on Build GRCh38
rs373421502 Chr.18: 59435284-59435284 on Build GRCh38
rs950590882 Chr.18: 59435491-59435491 on Build GRCh38
rs80296667 Chr.18: 59435571-59435571 on Build GRCh38
rs559237621 Chr.18: 59435264-59435264 on Build GRCh38
rs955309063 Chr.18: 59435360-59435360 on Build GRCh38
rs928180465 Chr.18: 59435462-59435462 on Build GRCh38
rs886054070 Chr.18: 59435263-59435263 on Build GRCh38
rs936827957 Chr.18: 59435675-59435675 on Build GRCh38
rs537510947 Chr.18: 59435641-59435641 on Build GRCh38
rs563607682 Chr.18: 59435413-59435413 on Build GRCh38
rs1022549527 Chr.18: 59435429-59435429 on Build GRCh38
rs536627089 Chr.18: 59435436-59435436 on Build GRCh38
rs1045406537 Chr.18: 59435457-59435457 on Build GRCh38
rs933354818 Chr.18: 59435593-59435593 on Build GRCh38
rs71357636 Chr.18: 59435628-59435628 on Build GRCh38
rs141921214 Chr.18: 59435530-59435530 on Build GRCh38
rs1043014005 Chr.18: 59435651-59435651 on Build GRCh38
rs957154094 Chr.18: 59435408-59435408 on Build GRCh38
rs781384409 Chr.18: 59435223-59435223 on Build GRCh38
rs1017042549 Chr.18: 59435299-59435299 on Build GRCh38
rs563567883 Chr.18: 59435402-59435402 on Build GRCh38
rs1053920634 Chr.18: 59435680-59435680 on Build GRCh38
rs1026251825 Chr.18: 59435465-59435465 on Build GRCh38

View Full Product Details