Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121566649-121567246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB2IP
Entrez Gene ID
153090
Gene Name
DAB2 interacting protein
Gene Aliases
AF9Q34, AIP-1, AIP1, DIP1/2
UniGene
Hs.522378
Gene Chromosome Location
Chr.9: 121566883-121785530 on Build GRCh38
Transcripts
NM_032552.3 XM_005251721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245314 c.40+7C>T p.? Chr.9: 121567235-121567235 on Build GRCh38
4636789 c.30C>T p.D10D Chr.9: 121567218-121567218 on Build GRCh38
7418434 c.35C>A p.S12Y Chr.9: 121567223-121567223 on Build GRCh38
1292781 c.39C>T p.Y13Y Chr.9: 121567227-121567227 on Build GRCh38
5410304 c.15C>T p.S5S Chr.9: 121567203-121567203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968572492 Chr.9: 121566914-121566914 on Build GRCh38
rs953347463 Chr.9: 121567130-121567130 on Build GRCh38
rs940491819 Chr.9: 121566950-121566950 on Build GRCh38
rs1051724010 Chr.9: 121567080-121567080 on Build GRCh38
rs957528667 Chr.9: 121567091-121567091 on Build GRCh38
rs200496124 Chr.9: 121567198-121567198 on Build GRCh38
rs553586926 Chr.9: 121566752-121566752 on Build GRCh38
rs971858268 Chr.9: 121567159-121567159 on Build GRCh38
rs556348772 Chr.9: 121567029-121567029 on Build GRCh38
rs1013282732 Chr.9: 121567039-121567039 on Build GRCh38
rs779342806 Chr.9: 121567145-121567145 on Build GRCh38
rs369955581 Chr.9: 121567175-121567175 on Build GRCh38
rs765763166 Chr.9: 121567179-121567179 on Build GRCh38
rs778322713 Chr.9: 121566883-121566883 on Build GRCh38
rs1033175179 Chr.9: 121566837-121566837 on Build GRCh38
rs992424611 Chr.9: 121566931-121566931 on Build GRCh38
rs148409496 Chr.9: 121567171-121567171 on Build GRCh38
rs772015259 Chr.9: 121566857-121566857 on Build GRCh38
rs768595008 Chr.9: 121567169-121567169 on Build GRCh38
rs1036646473 Chr.9: 121567010-121567010 on Build GRCh38
rs938377899 Chr.9: 121566929-121566929 on Build GRCh38
rs770115094 Chr.9: 121566851-121566851 on Build GRCh38
rs35128663 Chr.9: 121567126-121567127 on Build GRCh38
rs539656991 Chr.9: 121566727-121566727 on Build GRCh38
rs1045107716 Chr.9: 121567061-121567061 on Build GRCh38
rs1015174951 Chr.9: 121566751-121566752 on Build GRCh38
rs1013646951 Chr.9: 121566711-121566711 on Build GRCh38
rs779450388 Chr.9: 121567187-121567187 on Build GRCh38
rs779359789 Chr.9: 121567197-121567197 on Build GRCh38
rs555712577 Chr.9: 121566876-121566876 on Build GRCh38
rs560588941 Chr.9: 121567114-121567114 on Build GRCh38
rs904980003 Chr.9: 121567079-121567079 on Build GRCh38
rs1000609041 Chr.9: 121567087-121567087 on Build GRCh38
rs757734023 Chr.9: 121567195-121567195 on Build GRCh38
rs976130964 Chr.9: 121567110-121567110 on Build GRCh38
rs932058499 Chr.9: 121566904-121566904 on Build GRCh38
rs898053159 Chr.9: 121567059-121567059 on Build GRCh38
rs966680617 Chr.9: 121567107-121567107 on Build GRCh38
rs942195363 Chr.9: 121566996-121566996 on Build GRCh38
rs58469433 Chr.9: 121566761-121566761 on Build GRCh38
rs553920424 Chr.9: 121566800-121566800 on Build GRCh38
rs922067386 Chr.9: 121566896-121566896 on Build GRCh38
rs896365904 Chr.9: 121567031-121567031 on Build GRCh38
rs976458933 Chr.9: 121566890-121566890 on Build GRCh38
rs1032061041 Chr.9: 121567088-121567088 on Build GRCh38
rs1020546159 Chr.9: 121567095-121567095 on Build GRCh38
rs533976306 Chr.9: 121566766-121566766 on Build GRCh38
rs1048917881 Chr.9: 121566918-121566918 on Build GRCh38
rs201252302 Chr.9: 121567227-121567227 on Build GRCh38
rs968981559 Chr.9: 121567127-121567127 on Build GRCh38
rs142520673 Chr.9: 121567185-121567185 on Build GRCh38
rs757423468 Chr.9: 121566787-121566787 on Build GRCh38
rs745887681 Chr.9: 121566838-121566838 on Build GRCh38
rs915680234 Chr.9: 121566937-121566937 on Build GRCh38
rs969532804 Chr.9: 121566805-121566805 on Build GRCh38
rs984845100 Chr.9: 121567131-121567131 on Build GRCh38
rs909060590 Chr.9: 121566925-121566925 on Build GRCh38
rs966087577 Chr.9: 121566885-121566885 on Build GRCh38
rs909279139 Chr.9: 121567142-121567142 on Build GRCh38
rs1036206042 Chr.9: 121566685-121566685 on Build GRCh38
rs369466458 Chr.9: 121567147-121567147 on Build GRCh38
rs545706056 Chr.9: 121567044-121567044 on Build GRCh38
rs1045601367 Chr.9: 121566692-121566692 on Build GRCh38
rs771262362 Chr.9: 121567144-121567144 on Build GRCh38
rs573844828 Chr.9: 121566951-121566951 on Build GRCh38
rs1036256560 Chr.9: 121567023-121567023 on Build GRCh38
rs977779912 Chr.9: 121566920-121566920 on Build GRCh38
rs186002930 Chr.9: 121567126-121567126 on Build GRCh38
rs772343538 Chr.9: 121567202-121567202 on Build GRCh38
rs372088944 Chr.9: 121567150-121567150 on Build GRCh38
rs749679985 Chr.9: 121567140-121567140 on Build GRCh38
rs145245840 Chr.9: 121566698-121566698 on Build GRCh38
rs778480636 Chr.9: 121567194-121567194 on Build GRCh38
rs182217215 Chr.9: 121567032-121567032 on Build GRCh38
rs747154084 Chr.9: 121567218-121567218 on Build GRCh38
rs199630836 Chr.9: 121567219-121567219 on Build GRCh38
rs1010382787 Chr.9: 121567094-121567094 on Build GRCh38
rs748540060 Chr.9: 121566817-121566817 on Build GRCh38
rs749835580 Chr.9: 121567089-121567089 on Build GRCh38
rs537940499 Chr.9: 121567015-121567015 on Build GRCh38
rs756630816 Chr.9: 121567187-121567187 on Build GRCh38
rs771999384 Chr.9: 121567146-121567146 on Build GRCh38
rs776397734 Chr.9: 121567170-121567170 on Build GRCh38
rs111725535 Chr.9: 121566960-121566960 on Build GRCh38
rs936282742 Chr.9: 121567203-121567203 on Build GRCh38
rs750089842 Chr.9: 121567178-121567178 on Build GRCh38
rs887982588 Chr.9: 121566989-121566989 on Build GRCh38
rs778861773 Chr.9: 121566830-121566830 on Build GRCh38
rs35115829 Chr.9: 121567190-121567191 on Build GRCh38

View Full Product Details