Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110138034-110138626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP2
Entrez Gene ID
11217
Gene Name
A-kinase anchoring protein 2
Gene Aliases
AKAP-2, AKAPKL, MISP2, PRKA2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 110048598-110172512 on Build GRCh38
Transcripts
NM_001004065.4 NM_001136562.2 NM_001198656.1 NM_007203.4 NM_147150.2

Gene Symbol
PALM2-AKAP2
Entrez Gene ID
445815
Gene Name
PALM2-AKAP2 readthrough
Gene Aliases
AKAP2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 109780297-110172512 on Build GRCh38
Transcripts
NM_001004065.4 NM_001136562.2 NM_001198656.1 NM_007203.4 NM_147150.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1683652 c.2530_2531insAAGCTG p.A843_A844insEA Chr.9: 110138074-110138075 on Build GRCh38
5811665 c.2715C>A p.T905T Chr.9: 110138259-110138259 on Build GRCh38
6737109 c.2359G>T p.A787S Chr.9: 110138329-110138329 on Build GRCh38
3902938 c.2101G>T p.A701S Chr.9: 110138071-110138071 on Build GRCh38
5451949 c.2113G>A p.G705S Chr.9: 110138083-110138083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560571546 Chr.9: 110138436-110138436 on Build GRCh38
rs778457092 Chr.9: 110138249-110138249 on Build GRCh38
rs769596408 Chr.9: 110138398-110138398 on Build GRCh38
rs758590924 Chr.9: 110138082-110138082 on Build GRCh38
rs138545173 Chr.9: 110138122-110138122 on Build GRCh38
rs748069822 Chr.9: 110138394-110138394 on Build GRCh38
rs767818036 Chr.9: 110138304-110138304 on Build GRCh38
rs750832165 Chr.9: 110138570-110138570 on Build GRCh38
rs773100302 Chr.9: 110138413-110138413 on Build GRCh38
rs746629234 Chr.9: 110138502-110138502 on Build GRCh38
rs377019694 Chr.9: 110138497-110138497 on Build GRCh38
rs746520941 Chr.9: 110138266-110138266 on Build GRCh38
rs769839334 Chr.9: 110138513-110138513 on Build GRCh38
rs555942483 Chr.9: 110138350-110138350 on Build GRCh38
rs953797464 Chr.9: 110138282-110138282 on Build GRCh38
rs75000652 Chr.9: 110138075-110138075 on Build GRCh38
rs939332416 Chr.9: 110138359-110138359 on Build GRCh38
rs760869185 Chr.9: 110138454-110138454 on Build GRCh38
rs1050484076 Chr.9: 110138193-110138193 on Build GRCh38
rs149797840 Chr.9: 110138420-110138420 on Build GRCh38
rs756751023 Chr.9: 110138211-110138211 on Build GRCh38
rs754329741 Chr.9: 110138334-110138334 on Build GRCh38
rs200087277 Chr.9: 110138148-110138148 on Build GRCh38
rs12338163 Chr.9: 110138187-110138187 on Build GRCh38
rs376606574 Chr.9: 110138475-110138475 on Build GRCh38
rs755252885 Chr.9: 110138583-110138585 on Build GRCh38
rs150240514 Chr.9: 110138328-110138328 on Build GRCh38
rs1027949247 Chr.9: 110138579-110138579 on Build GRCh38
rs202061551 Chr.9: 110138480-110138480 on Build GRCh38
rs139808664 Chr.9: 110138417-110138417 on Build GRCh38
rs770546074 Chr.9: 110138124-110138124 on Build GRCh38
rs376237093 Chr.9: 110138202-110138203 on Build GRCh38
rs61735424 Chr.9: 110138186-110138186 on Build GRCh38
rs137889332 Chr.9: 110138083-110138083 on Build GRCh38
rs542301338 Chr.9: 110138424-110138424 on Build GRCh38
rs777511698 Chr.9: 110138519-110138519 on Build GRCh38
rs765789312 Chr.9: 110138567-110138567 on Build GRCh38
rs1028420537 Chr.9: 110138233-110138233 on Build GRCh38
rs199910947 Chr.9: 110138325-110138325 on Build GRCh38
rs775424696 Chr.9: 110138161-110138161 on Build GRCh38
rs750860393 Chr.9: 110138496-110138496 on Build GRCh38
rs150118041 Chr.9: 110138113-110138113 on Build GRCh38
rs761023069 Chr.9: 110138544-110138544 on Build GRCh38
rs34665027 Chr.9: 110138061-110138061 on Build GRCh38
rs780805317 Chr.9: 110138507-110138507 on Build GRCh38
rs990809211 Chr.9: 110138386-110138386 on Build GRCh38
rs760651898 Chr.9: 110138169-110138169 on Build GRCh38
rs561065144 Chr.9: 110138416-110138416 on Build GRCh38
rs749098354 Chr.9: 110138522-110138522 on Build GRCh38
rs74612901 Chr.9: 110138601-110138601 on Build GRCh38
rs112489156 Chr.9: 110138291-110138291 on Build GRCh38
rs771568417 Chr.9: 110138160-110138160 on Build GRCh38
rs770808786 Chr.9: 110138524-110138524 on Build GRCh38
rs745471428 Chr.9: 110138264-110138264 on Build GRCh38
rs767369093 Chr.9: 110138063-110138063 on Build GRCh38
rs752803948 Chr.9: 110138206-110138206 on Build GRCh38
rs776401771 Chr.9: 110138073-110138074 on Build GRCh38
rs770005256 Chr.9: 110138539-110138539 on Build GRCh38
rs771259884 Chr.9: 110138241-110138241 on Build GRCh38
rs370870911 Chr.9: 110138346-110138346 on Build GRCh38
rs781372720 Chr.9: 110138204-110138204 on Build GRCh38
rs770306162 Chr.9: 110138117-110138117 on Build GRCh38
rs143737736 Chr.9: 110138267-110138267 on Build GRCh38
rs764217285 Chr.9: 110138463-110138463 on Build GRCh38
rs779670894 Chr.9: 110138361-110138361 on Build GRCh38
rs776496960 Chr.9: 110138285-110138285 on Build GRCh38
rs35935397 Chr.9: 110138092-110138092 on Build GRCh38
rs150611744 Chr.9: 110138347-110138347 on Build GRCh38
rs778408995 Chr.9: 110138225-110138225 on Build GRCh38
rs749683953 Chr.9: 110138240-110138240 on Build GRCh38
rs762125148 Chr.9: 110138203-110138203 on Build GRCh38
rs531486809 Chr.9: 110138573-110138573 on Build GRCh38
rs768091773 Chr.9: 110138387-110138387 on Build GRCh38
rs759853090 Chr.9: 110138301-110138301 on Build GRCh38
rs542888642 Chr.9: 110138330-110138330 on Build GRCh38
rs768964636 Chr.9: 110138546-110138546 on Build GRCh38
rs3739456 Chr.9: 110138505-110138505 on Build GRCh38
rs201693389 Chr.9: 110138197-110138197 on Build GRCh38
rs888451902 Chr.9: 110138202-110138202 on Build GRCh38
rs541180913 Chr.9: 110138552-110138552 on Build GRCh38
rs182568604 Chr.9: 110138180-110138180 on Build GRCh38
rs752560049 Chr.9: 110138071-110138071 on Build GRCh38
rs767440591 Chr.9: 110138384-110138384 on Build GRCh38
rs775787860 Chr.9: 110138428-110138428 on Build GRCh38
rs761783014 Chr.9: 110138555-110138555 on Build GRCh38
rs775877745 Chr.9: 110138534-110138534 on Build GRCh38
rs140643568 Chr.9: 110138520-110138520 on Build GRCh38
rs759366058 Chr.9: 110138075-110138076 on Build GRCh38
rs773996995 Chr.9: 110138139-110138139 on Build GRCh38
rs754236658 Chr.9: 110138521-110138523 on Build GRCh38
rs755031122 Chr.9: 110138096-110138096 on Build GRCh38
rs111629628 Chr.9: 110138219-110138219 on Build GRCh38
rs776585237 Chr.9: 110138388-110138388 on Build GRCh38
rs373087862 Chr.9: 110138474-110138474 on Build GRCh38
rs139897209 Chr.9: 110138292-110138292 on Build GRCh38
rs373159646 Chr.9: 110138074-110138075 on Build GRCh38
rs1042669407 Chr.9: 110138069-110138069 on Build GRCh38
rs750589648 Chr.9: 110138450-110138451 on Build GRCh38
rs747582238 Chr.9: 110138508-110138508 on Build GRCh38
rs746539180 Chr.9: 110138364-110138364 on Build GRCh38
rs761260209 Chr.9: 110138465-110138466 on Build GRCh38
rs760490130 Chr.9: 110138072-110138072 on Build GRCh38
rs771598917 Chr.9: 110138265-110138265 on Build GRCh38
rs763985320 Chr.9: 110138173-110138173 on Build GRCh38
rs199516874 Chr.9: 110138060-110138060 on Build GRCh38
rs765655099 Chr.9: 110138494-110138494 on Build GRCh38
rs545952225 Chr.9: 110138322-110138322 on Build GRCh38
rs776835403 Chr.9: 110138551-110138551 on Build GRCh38
rs575100870 Chr.9: 110138353-110138353 on Build GRCh38
rs1018905760 Chr.9: 110138223-110138223 on Build GRCh38
rs76059538 Chr.9: 110138076-110138076 on Build GRCh38
rs755044641 Chr.9: 110138198-110138198 on Build GRCh38
rs773798967 Chr.9: 110138055-110138055 on Build GRCh38
rs767033135 Chr.9: 110138496-110138496 on Build GRCh38
rs150402481 Chr.9: 110138061-110138062 on Build GRCh38

View Full Product Details