Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 25382965-25383492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
UniGene
Hs.152173
Gene Chromosome Location
Chr.4: 25377226-25418498 on Build GRCh38
Transcripts
NM_001286756.1 NM_013367.2 XM_011513838.1 XM_011513839.1 XM_017008138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1054666 c.349G>A p.E117K Chr.4: 25383374-25383374 on Build GRCh38
6545312 c.333C>T p.S111S Chr.4: 25383358-25383358 on Build GRCh38
1319344 c.236-2A>T p.? Chr.4: 25383259-25383259 on Build GRCh38
3603081 c.321G>A p.E107E Chr.4: 25383346-25383346 on Build GRCh38
3428446 c.250C>T p.L84F Chr.4: 25383275-25383275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374830999 Chr.4: 25383279-25383279 on Build GRCh38
rs762596842 Chr.4: 25383307-25383307 on Build GRCh38
rs960691803 Chr.4: 25383031-25383031 on Build GRCh38
rs769312617 Chr.4: 25383293-25383293 on Build GRCh38
rs756223606 Chr.4: 25383362-25383362 on Build GRCh38
rs192962868 Chr.4: 25383184-25383184 on Build GRCh38
rs1034047370 Chr.4: 25383265-25383265 on Build GRCh38
rs771928831 Chr.4: 25383259-25383259 on Build GRCh38
rs762128577 Chr.4: 25383337-25383337 on Build GRCh38
rs748867356 Chr.4: 25383428-25383428 on Build GRCh38
rs561644214 Chr.4: 25383021-25383021 on Build GRCh38
rs750916286 Chr.4: 25383345-25383345 on Build GRCh38
rs773928073 Chr.4: 25383442-25383442 on Build GRCh38
rs746788136 Chr.4: 25383408-25383408 on Build GRCh38
rs183571387 Chr.4: 25383227-25383227 on Build GRCh38
rs145762349 Chr.4: 25382994-25382994 on Build GRCh38
rs377168380 Chr.4: 25383155-25383155 on Build GRCh38
rs141365775 Chr.4: 25383129-25383129 on Build GRCh38
rs767095785 Chr.4: 25383002-25383002 on Build GRCh38
rs188212244 Chr.4: 25383411-25383411 on Build GRCh38
rs759024464 Chr.4: 25383292-25383292 on Build GRCh38
rs750489087 Chr.4: 25383234-25383234 on Build GRCh38
rs891889262 Chr.4: 25383204-25383204 on Build GRCh38
rs533238611 Chr.4: 25383078-25383078 on Build GRCh38
rs908442040 Chr.4: 25383039-25383039 on Build GRCh38
rs201839196 Chr.4: 25383289-25383289 on Build GRCh38
rs753691993 Chr.4: 25383400-25383400 on Build GRCh38
rs939732338 Chr.4: 25383089-25383089 on Build GRCh38
rs941281731 Chr.4: 25383072-25383072 on Build GRCh38
rs991828032 Chr.4: 25383033-25383033 on Build GRCh38
rs756315710 Chr.4: 25383239-25383239 on Build GRCh38
rs147243013 Chr.4: 25383333-25383333 on Build GRCh38
rs376010530 Chr.4: 25383372-25383372 on Build GRCh38
rs745832603 Chr.4: 25383418-25383418 on Build GRCh38
rs1002034579 Chr.4: 25383245-25383245 on Build GRCh38
rs138403729 Chr.4: 25383067-25383067 on Build GRCh38
rs74863363 Chr.4: 25383260-25383260 on Build GRCh38
rs13127336 Chr.4: 25383324-25383324 on Build GRCh38
rs745672751 Chr.4: 25383244-25383245 on Build GRCh38
rs1030385877 Chr.4: 25383422-25383422 on Build GRCh38
rs1011645337 Chr.4: 25383208-25383208 on Build GRCh38
rs148626453 Chr.4: 25383039-25383041 on Build GRCh38
rs1009412704 Chr.4: 25383007-25383007 on Build GRCh38
rs746836321 Chr.4: 25383274-25383274 on Build GRCh38
rs908388400 Chr.4: 25383466-25383466 on Build GRCh38
rs527510679 Chr.4: 25383022-25383022 on Build GRCh38
rs760384142 Chr.4: 25383131-25383131 on Build GRCh38
rs967969597 Chr.4: 25383016-25383016 on Build GRCh38
rs775080525 Chr.4: 25383299-25383299 on Build GRCh38
rs200095852 Chr.4: 25383410-25383410 on Build GRCh38
rs556214229 Chr.4: 25383426-25383426 on Build GRCh38
rs369576271 Chr.4: 25383036-25383036 on Build GRCh38
rs758202065 Chr.4: 25383413-25383414 on Build GRCh38
rs13102409 Chr.4: 25383326-25383326 on Build GRCh38
rs749554010 Chr.4: 25383247-25383247 on Build GRCh38
rs948458256 Chr.4: 25383132-25383132 on Build GRCh38
rs541799233 Chr.4: 25383008-25383008 on Build GRCh38
rs957628521 Chr.4: 25382991-25382991 on Build GRCh38
rs371805695 Chr.4: 25383341-25383341 on Build GRCh38
rs780375315 Chr.4: 25383243-25383243 on Build GRCh38
rs967897206 Chr.4: 25383228-25383228 on Build GRCh38
rs779990618 Chr.4: 25383440-25383441 on Build GRCh38
rs780287319 Chr.4: 25383371-25383371 on Build GRCh38
rs2292405 Chr.4: 25383118-25383118 on Build GRCh38
rs575797971 Chr.4: 25383309-25383309 on Build GRCh38
rs576001123 Chr.4: 25383432-25383432 on Build GRCh38
rs776473600 Chr.4: 25383285-25383285 on Build GRCh38
rs779068339 Chr.4: 25383399-25383399 on Build GRCh38
rs755236879 Chr.4: 25383374-25383374 on Build GRCh38
rs367635861 Chr.4: 25383280-25383280 on Build GRCh38
rs181787523 Chr.4: 25383419-25383419 on Build GRCh38
rs891377222 Chr.4: 25383148-25383148 on Build GRCh38
rs777638838 Chr.4: 25383268-25383268 on Build GRCh38
rs1050457522 Chr.4: 25383138-25383138 on Build GRCh38
rs990111379 Chr.4: 25382993-25382993 on Build GRCh38
rs77497792 Chr.4: 25383219-25383219 on Build GRCh38

View Full Product Details