Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151506991-151507510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT2
Entrez Gene ID
2196
Gene Name
FAT atypical cadherin 2
Gene Aliases
CDHF8, CDHR9, HFAT2, MEGF1
UniGene
Hs.591255
Gene Chromosome Location
Chr.5: 151504092-151594822 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
UniGene
Hs.269004
Gene Chromosome Location
Chr.5: 151411344-151556318 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4523919 c.12414G>A p.R4138R Chr.5: 151507257-151507257 on Build GRCh38
216711 c.12417A>G p.P4139P Chr.5: 151507254-151507254 on Build GRCh38
3761045 c.12350C>T p.P4117L Chr.5: 151507321-151507321 on Build GRCh38
301551 c.12363_12364GG>CT p.K4121>? Chr.5: 151507307-151507308 on Build GRCh38
1718827 c.12164C>T p.A4055V Chr.5: 151507507-151507507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772772158 Chr.5: 151507480-151507480 on Build GRCh38
rs759410331 Chr.5: 151507236-151507236 on Build GRCh38
rs1105168 Chr.5: 151507321-151507321 on Build GRCh38
rs749389986 Chr.5: 151507453-151507453 on Build GRCh38
rs745784430 Chr.5: 151507343-151507343 on Build GRCh38
rs762302252 Chr.5: 151507488-151507488 on Build GRCh38
rs138705803 Chr.5: 151507351-151507351 on Build GRCh38
rs777974723 Chr.5: 151507417-151507417 on Build GRCh38
rs111771262 Chr.5: 151507203-151507203 on Build GRCh38
rs758682292 Chr.5: 151507224-151507224 on Build GRCh38
rs575183690 Chr.5: 151507155-151507155 on Build GRCh38
rs755238824 Chr.5: 151507400-151507400 on Build GRCh38
rs748130207 Chr.5: 151507215-151507215 on Build GRCh38
rs927235826 Chr.5: 151507340-151507340 on Build GRCh38
rs779466326 Chr.5: 151507204-151507204 on Build GRCh38
rs763502422 Chr.5: 151507372-151507372 on Build GRCh38
rs143801705 Chr.5: 151507479-151507479 on Build GRCh38
rs775185995 Chr.5: 151507241-151507241 on Build GRCh38
rs373433514 Chr.5: 151507137-151507137 on Build GRCh38
rs755091164 Chr.5: 151507156-151507156 on Build GRCh38
rs755223311 Chr.5: 151507315-151507315 on Build GRCh38
rs370437664 Chr.5: 151507227-151507227 on Build GRCh38
rs543652342 Chr.5: 151507258-151507258 on Build GRCh38
rs188790055 Chr.5: 151507056-151507056 on Build GRCh38
rs756209489 Chr.5: 151507333-151507333 on Build GRCh38
rs768808587 Chr.5: 151507216-151507216 on Build GRCh38
rs780306307 Chr.5: 151507211-151507211 on Build GRCh38
rs141033551 Chr.5: 151507185-151507185 on Build GRCh38
rs7722798 Chr.5: 151507087-151507087 on Build GRCh38
rs776208786 Chr.5: 151507355-151507355 on Build GRCh38
rs369979058 Chr.5: 151507298-151507298 on Build GRCh38
rs1019364592 Chr.5: 151507218-151507218 on Build GRCh38
rs567269595 Chr.5: 151507044-151507044 on Build GRCh38
rs770489194 Chr.5: 151507490-151507490 on Build GRCh38
rs749663238 Chr.5: 151507409-151507409 on Build GRCh38
rs193215777 Chr.5: 151507075-151507075 on Build GRCh38
rs146563944 Chr.5: 151507231-151507231 on Build GRCh38
rs780255836 Chr.5: 151507119-151507119 on Build GRCh38
rs145466001 Chr.5: 151507207-151507207 on Build GRCh38
rs7723266 Chr.5: 151507164-151507164 on Build GRCh38
rs748391997 Chr.5: 151507322-151507322 on Build GRCh38
rs750934872 Chr.5: 151507205-151507205 on Build GRCh38
rs7723079 Chr.5: 151507055-151507055 on Build GRCh38
rs758862420 Chr.5: 151507295-151507295 on Build GRCh38
rs747212122 Chr.5: 151507127-151507127 on Build GRCh38
rs377061912 Chr.5: 151507194-151507194 on Build GRCh38
rs531281319 Chr.5: 151507403-151507403 on Build GRCh38
rs199556909 Chr.5: 151507457-151507457 on Build GRCh38
rs545597751 Chr.5: 151507348-151507348 on Build GRCh38
rs757751855 Chr.5: 151507197-151507197 on Build GRCh38
rs368309685 Chr.5: 151507233-151507233 on Build GRCh38
rs1040829740 Chr.5: 151507050-151507050 on Build GRCh38
rs747769250 Chr.5: 151507478-151507478 on Build GRCh38
rs748304520 Chr.5: 151507225-151507225 on Build GRCh38
rs768734506 Chr.5: 151507135-151507135 on Build GRCh38
rs751925193 Chr.5: 151507307-151507307 on Build GRCh38
rs904759464 Chr.5: 151507320-151507320 on Build GRCh38
rs201825044 Chr.5: 151507404-151507404 on Build GRCh38
rs760355926 Chr.5: 151507242-151507242 on Build GRCh38
rs746980768 Chr.5: 151507458-151507458 on Build GRCh38
rs1034329298 Chr.5: 151507102-151507102 on Build GRCh38
rs1056462267 Chr.5: 151507052-151507052 on Build GRCh38
rs764741469 Chr.5: 151507282-151507282 on Build GRCh38
rs144848990 Chr.5: 151507259-151507259 on Build GRCh38
rs750882454 Chr.5: 151507289-151507289 on Build GRCh38
rs766743053 Chr.5: 151507297-151507297 on Build GRCh38
rs779535153 Chr.5: 151507407-151507407 on Build GRCh38
rs770681196 Chr.5: 151507337-151507338 on Build GRCh38
rs898716084 Chr.5: 151507128-151507128 on Build GRCh38
rs533240264 Chr.5: 151507337-151507337 on Build GRCh38
rs766860387 Chr.5: 151507386-151507386 on Build GRCh38
rs768938668 Chr.5: 151507447-151507447 on Build GRCh38
rs776442798 Chr.5: 151507462-151507462 on Build GRCh38
rs993306008 Chr.5: 151507141-151507141 on Build GRCh38
rs374289322 Chr.5: 151507456-151507456 on Build GRCh38
rs372485938 Chr.5: 151507089-151507089 on Build GRCh38
rs760273190 Chr.5: 151507171-151507171 on Build GRCh38
rs370614755 Chr.5: 151507273-151507273 on Build GRCh38
rs901329361 Chr.5: 151507054-151507054 on Build GRCh38
rs201787846 Chr.5: 151507136-151507136 on Build GRCh38
rs918406526 Chr.5: 151507414-151507414 on Build GRCh38
rs752111755 Chr.5: 151507391-151507391 on Build GRCh38
rs377690143 Chr.5: 151507146-151507146 on Build GRCh38
rs755763462 Chr.5: 151507392-151507392 on Build GRCh38
rs761476586 Chr.5: 151507359-151507359 on Build GRCh38
rs768447645 Chr.5: 151507353-151507353 on Build GRCh38
rs756442595 Chr.5: 151507416-151507416 on Build GRCh38
rs768392091 Chr.5: 151507460-151507460 on Build GRCh38
rs771858676 Chr.5: 151507238-151507238 on Build GRCh38
rs772181147 Chr.5: 151507114-151507114 on Build GRCh38
rs367689507 Chr.5: 151507367-151507367 on Build GRCh38
rs200773290 Chr.5: 151507360-151507360 on Build GRCh38
rs972570293 Chr.5: 151507193-151507193 on Build GRCh38
rs746293951 Chr.5: 151507103-151507103 on Build GRCh38

View Full Product Details