Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 20481071-20481669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F3
Entrez Gene ID
1871
Gene Name
E2F transcription factor 3
Gene Aliases
E2F-3
UniGene
Hs.269408
Gene Chromosome Location
Chr.6: 20353285-20493714 on Build GRCh38
Transcripts
NM_001243076.2 NM_001949.4 XM_005248865.4 XM_005248866.4 XM_011514324.2 XM_011514328.2 XM_017010330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
290791 c.662T>C p.I221T Chr.6: 20481362-20481362 on Build GRCh38
6453117 c.670G>A p.V224I Chr.6: 20481370-20481370 on Build GRCh38
7160329 c.541A>G p.T181A Chr.6: 20481241-20481241 on Build GRCh38
6470982 c.693T>A p.I231I Chr.6: 20481393-20481393 on Build GRCh38
6470983 c.694A>T p.K232* Chr.6: 20481394-20481394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571679412 Chr.6: 20481506-20481506 on Build GRCh38
rs200064115 Chr.6: 20481378-20481378 on Build GRCh38
rs746801031 Chr.6: 20481288-20481288 on Build GRCh38
rs758115436 Chr.6: 20481464-20481464 on Build GRCh38
rs971582511 Chr.6: 20481142-20481142 on Build GRCh38
rs757990691 Chr.6: 20481243-20481243 on Build GRCh38
rs777604852 Chr.6: 20481246-20481246 on Build GRCh38
rs759628539 Chr.6: 20481173-20481173 on Build GRCh38
rs1016537961 Chr.6: 20481215-20481215 on Build GRCh38
rs573206876 Chr.6: 20481630-20481630 on Build GRCh38
rs777372521 Chr.6: 20481467-20481467 on Build GRCh38
rs757759086 Chr.6: 20481193-20481193 on Build GRCh38
rs1003353273 Chr.6: 20481567-20481567 on Build GRCh38
rs1032803140 Chr.6: 20481591-20481591 on Build GRCh38
rs1012772253 Chr.6: 20481601-20481601 on Build GRCh38
rs1034734922 Chr.6: 20481174-20481174 on Build GRCh38
rs778898372 Chr.6: 20481182-20481182 on Build GRCh38
rs187344631 Chr.6: 20481146-20481146 on Build GRCh38
rs886967275 Chr.6: 20481643-20481643 on Build GRCh38
rs200506712 Chr.6: 20481303-20481303 on Build GRCh38
rs539269037 Chr.6: 20481463-20481463 on Build GRCh38
rs181603569 Chr.6: 20481294-20481294 on Build GRCh38
rs1044954638 Chr.6: 20481480-20481480 on Build GRCh38
rs191815365 Chr.6: 20481179-20481179 on Build GRCh38
rs747303404 Chr.6: 20481519-20481519 on Build GRCh38
rs542718331 Chr.6: 20481518-20481518 on Build GRCh38
rs138776759 Chr.6: 20481242-20481242 on Build GRCh38
rs1023954109 Chr.6: 20481096-20481096 on Build GRCh38
rs770741555 Chr.6: 20481459-20481460 on Build GRCh38
rs376054295 Chr.6: 20481167-20481167 on Build GRCh38
rs769945484 Chr.6: 20481345-20481345 on Build GRCh38
rs1024403243 Chr.6: 20481608-20481609 on Build GRCh38
rs755037452 Chr.6: 20481165-20481165 on Build GRCh38
rs948764509 Chr.6: 20481479-20481479 on Build GRCh38
rs972227003 Chr.6: 20481616-20481616 on Build GRCh38
rs753339943 Chr.6: 20481204-20481204 on Build GRCh38
rs201376251 Chr.6: 20481369-20481369 on Build GRCh38
rs555002825 Chr.6: 20481170-20481170 on Build GRCh38
rs551147614 Chr.6: 20481161-20481161 on Build GRCh38
rs747753211 Chr.6: 20481155-20481155 on Build GRCh38
rs934718343 Chr.6: 20481461-20481461 on Build GRCh38
rs772865381 Chr.6: 20481187-20481188 on Build GRCh38
rs767495188 Chr.6: 20481104-20481104 on Build GRCh38
rs1040134278 Chr.6: 20481439-20481439 on Build GRCh38
rs575713354 Chr.6: 20481511-20481511 on Build GRCh38
rs760198994 Chr.6: 20481199-20481199 on Build GRCh38
rs745743197 Chr.6: 20481333-20481333 on Build GRCh38
rs771676946 Chr.6: 20481173-20481178 on Build GRCh38
rs764395178 Chr.6: 20481428-20481428 on Build GRCh38
rs370434089 Chr.6: 20481166-20481166 on Build GRCh38
rs369637402 Chr.6: 20481218-20481218 on Build GRCh38
rs752957903 Chr.6: 20481222-20481222 on Build GRCh38
rs4134938 Chr.6: 20481447-20481447 on Build GRCh38
rs991153908 Chr.6: 20481094-20481095 on Build GRCh38
rs971063433 Chr.6: 20481612-20481612 on Build GRCh38
rs750974803 Chr.6: 20481449-20481449 on Build GRCh38
rs747804161 Chr.6: 20481232-20481232 on Build GRCh38
rs1000018432 Chr.6: 20481528-20481528 on Build GRCh38
rs4134939 Chr.6: 20481594-20481594 on Build GRCh38
rs776640693 Chr.6: 20481171-20481171 on Build GRCh38
rs781704387 Chr.6: 20481203-20481203 on Build GRCh38
rs1041763444 Chr.6: 20481505-20481505 on Build GRCh38
rs774898549 Chr.6: 20481433-20481433 on Build GRCh38
rs567111741 Chr.6: 20481354-20481354 on Build GRCh38
rs910171089 Chr.6: 20481327-20481327 on Build GRCh38
rs780656274 Chr.6: 20481165-20481165 on Build GRCh38
rs765427245 Chr.6: 20481177-20481177 on Build GRCh38
rs764741445 Chr.6: 20481452-20481452 on Build GRCh38
rs762217609 Chr.6: 20481434-20481434 on Build GRCh38
rs1026029798 Chr.6: 20481639-20481639 on Build GRCh38
rs763278356 Chr.6: 20481414-20481414 on Build GRCh38
rs571959359 Chr.6: 20481459-20481459 on Build GRCh38

View Full Product Details