Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31726901-31727489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDAH2
Entrez Gene ID
23564
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
DDAH, DDAHII, G6a, HEL-S-277, NG30
UniGene
Hs.247362
Gene Chromosome Location
Chr.6: 31727037-31730265 on Build GRCh38
Transcripts
NM_001303007.1 NM_001303008.1 NM_013974.2 XM_011514448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3861789 c.475G>A p.A159T Chr.6: 31727287-31727287 on Build GRCh38
3777495 c.504C>T p.C168C Chr.6: 31727258-31727258 on Build GRCh38
4654087 c.475G>T p.A159S Chr.6: 31727287-31727287 on Build GRCh38
4185953 c.793C>A p.L265M Chr.6: 31727302-31727302 on Build GRCh38
4421540 c.436C>A p.P146T Chr.6: 31727326-31727326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769483429 Chr.6: 31727131-31727131 on Build GRCh38
rs778766347 Chr.6: 31727201-31727201 on Build GRCh38
rs1027995536 Chr.6: 31726923-31726923 on Build GRCh38
rs754647325 Chr.6: 31727196-31727196 on Build GRCh38
rs764969360 Chr.6: 31727199-31727216 on Build GRCh38
rs45565837 Chr.6: 31727082-31727082 on Build GRCh38
rs950564948 Chr.6: 31726986-31726986 on Build GRCh38
rs557920597 Chr.6: 31726968-31726968 on Build GRCh38
rs766150261 Chr.6: 31727173-31727173 on Build GRCh38
rs148908277 Chr.6: 31727277-31727277 on Build GRCh38
rs949481542 Chr.6: 31727075-31727075 on Build GRCh38
rs867163078 Chr.6: 31727432-31727432 on Build GRCh38
rs983305553 Chr.6: 31727009-31727009 on Build GRCh38
rs771369955 Chr.6: 31727236-31727236 on Build GRCh38
rs918364409 Chr.6: 31726927-31726927 on Build GRCh38
rs1054028374 Chr.6: 31727264-31727264 on Build GRCh38
rs908624094 Chr.6: 31727074-31727074 on Build GRCh38
rs760623483 Chr.6: 31727287-31727287 on Build GRCh38
rs373175852 Chr.6: 31727284-31727284 on Build GRCh38
rs1042568862 Chr.6: 31727376-31727376 on Build GRCh38
rs754788450 Chr.6: 31727384-31727384 on Build GRCh38
rs766268044 Chr.6: 31727288-31727288 on Build GRCh38
rs751068012 Chr.6: 31727381-31727381 on Build GRCh38
rs777294461 Chr.6: 31727332-31727332 on Build GRCh38
rs1048071707 Chr.6: 31726984-31726984 on Build GRCh38
rs138036242 Chr.6: 31727327-31727327 on Build GRCh38
rs756022047 Chr.6: 31727171-31727171 on Build GRCh38
rs759470664 Chr.6: 31727297-31727297 on Build GRCh38
rs533470524 Chr.6: 31727132-31727132 on Build GRCh38
rs555664979 Chr.6: 31727036-31727036 on Build GRCh38
rs759204983 Chr.6: 31727188-31727189 on Build GRCh38
rs562967660 Chr.6: 31727127-31727127 on Build GRCh38
rs527712794 Chr.6: 31727388-31727388 on Build GRCh38
rs774318297 Chr.6: 31727285-31727285 on Build GRCh38
rs143584444 Chr.6: 31727299-31727299 on Build GRCh38
rs777160515 Chr.6: 31727221-31727221 on Build GRCh38
rs752457491 Chr.6: 31727254-31727255 on Build GRCh38
rs773004243 Chr.6: 31727258-31727258 on Build GRCh38
rs750822347 Chr.6: 31727162-31727165 on Build GRCh38
rs752337796 Chr.6: 31727311-31727311 on Build GRCh38
rs906417584 Chr.6: 31727151-31727151 on Build GRCh38
rs764719456 Chr.6: 31727111-31727111 on Build GRCh38
rs746230780 Chr.6: 31727395-31727395 on Build GRCh38
rs145447627 Chr.6: 31727266-31727266 on Build GRCh38
rs757913554 Chr.6: 31727215-31727215 on Build GRCh38
rs749902463 Chr.6: 31727115-31727115 on Build GRCh38
rs1008564505 Chr.6: 31727454-31727454 on Build GRCh38
rs888099289 Chr.6: 31727032-31727032 on Build GRCh38
rs573742405 Chr.6: 31727054-31727054 on Build GRCh38
rs777018614 Chr.6: 31727241-31727241 on Build GRCh38
rs754324785 Chr.6: 31727006-31727006 on Build GRCh38
rs541654362 Chr.6: 31727227-31727227 on Build GRCh38
rs747497629 Chr.6: 31727223-31727223 on Build GRCh38
rs758006955 Chr.6: 31727194-31727194 on Build GRCh38
rs150096378 Chr.6: 31727251-31727251 on Build GRCh38
rs747656485 Chr.6: 31727210-31727210 on Build GRCh38

View Full Product Details