Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41777807-41778339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRS3
Entrez Gene ID
10817
Gene Name
fibroblast growth factor receptor substrate 3
Gene Aliases
FRS2-beta, FRS2B, FRS2beta, SNT-2, SNT2
UniGene
Hs.194208
Gene Chromosome Location
Chr.6: 41770176-41779905 on Build GRCh38
Transcripts
NM_006653.4 XM_011514254.2 XM_011514255.2 XM_017010193.1

SNP Detail

SNP ID Chromosome Location
rs146229648 Chr.6: 41778277-41778277 on Build GRCh38
rs185686467 Chr.6: 41778003-41778003 on Build GRCh38
rs555170299 Chr.6: 41778302-41778302 on Build GRCh38
rs9471650 Chr.6: 41778155-41778155 on Build GRCh38
rs182502611 Chr.6: 41778100-41778100 on Build GRCh38
rs549180595 Chr.6: 41778113-41778113 on Build GRCh38
rs922599234 Chr.6: 41777856-41777856 on Build GRCh38
rs760527713 Chr.6: 41777948-41777948 on Build GRCh38
rs985834613 Chr.6: 41778300-41778300 on Build GRCh38
rs541593317 Chr.6: 41777924-41777924 on Build GRCh38
rs182659432 Chr.6: 41778303-41778303 on Build GRCh38
rs776022191 Chr.6: 41778081-41778081 on Build GRCh38
rs1056302994 Chr.6: 41778165-41778165 on Build GRCh38
rs940811876 Chr.6: 41778088-41778088 on Build GRCh38
rs763335247 Chr.6: 41778110-41778110 on Build GRCh38
rs552538218 Chr.6: 41778072-41778072 on Build GRCh38
rs1034610044 Chr.6: 41777999-41777999 on Build GRCh38
rs570101092 Chr.6: 41778059-41778059 on Build GRCh38
rs968500073 Chr.6: 41778307-41778307 on Build GRCh38
rs902408294 Chr.6: 41777949-41777949 on Build GRCh38
rs3804281 Chr.6: 41778251-41778251 on Build GRCh38
rs961527994 Chr.6: 41778274-41778274 on Build GRCh38
rs756200488 Chr.6: 41778093-41778093 on Build GRCh38
rs774562461 Chr.6: 41778150-41778150 on Build GRCh38
rs908777679 Chr.6: 41777828-41777828 on Build GRCh38
rs1029901375 Chr.6: 41778285-41778285 on Build GRCh38
rs1027806508 Chr.6: 41778021-41778021 on Build GRCh38
rs777558279 Chr.6: 41778148-41778148 on Build GRCh38
rs998381328 Chr.6: 41778284-41778284 on Build GRCh38
rs989616149 Chr.6: 41778231-41778231 on Build GRCh38
rs931131391 Chr.6: 41778101-41778101 on Build GRCh38
rs115852548 Chr.6: 41778120-41778120 on Build GRCh38
rs539382190 Chr.6: 41778230-41778230 on Build GRCh38
rs976713829 Chr.6: 41777845-41777845 on Build GRCh38
rs1027343026 Chr.6: 41778249-41778249 on Build GRCh38
rs893919741 Chr.6: 41777903-41777903 on Build GRCh38
rs532706356 Chr.6: 41778068-41778068 on Build GRCh38
rs556544828 Chr.6: 41778217-41778217 on Build GRCh38
rs752664280 Chr.6: 41778118-41778118 on Build GRCh38
rs916808082 Chr.6: 41778013-41778013 on Build GRCh38
rs953736300 Chr.6: 41777912-41777912 on Build GRCh38
rs139301498 Chr.6: 41778283-41778283 on Build GRCh38
rs946523745 Chr.6: 41777910-41777910 on Build GRCh38
rs1012309577 Chr.6: 41778226-41778226 on Build GRCh38
rs948046031 Chr.6: 41778050-41778050 on Build GRCh38
rs112863256 Chr.6: 41778291-41778291 on Build GRCh38
rs41273792 Chr.6: 41778172-41778172 on Build GRCh38
rs1019737497 Chr.6: 41778137-41778137 on Build GRCh38
rs1050262020 Chr.6: 41777888-41777888 on Build GRCh38
rs537256435 Chr.6: 41778146-41778146 on Build GRCh38
rs190147027 Chr.6: 41778060-41778060 on Build GRCh38
rs1003424569 Chr.6: 41777951-41777951 on Build GRCh38
rs186826681 Chr.6: 41778115-41778115 on Build GRCh38
rs369225404 Chr.6: 41778127-41778127 on Build GRCh38
rs758123788 Chr.6: 41778139-41778139 on Build GRCh38
rs961580975 Chr.6: 41778069-41778069 on Build GRCh38
rs570759203 Chr.6: 41778179-41778179 on Build GRCh38
rs1014719948 Chr.6: 41778260-41778260 on Build GRCh38
rs767686148 Chr.6: 41777913-41777913 on Build GRCh38
rs181810095 Chr.6: 41778000-41778000 on Build GRCh38
rs940274853 Chr.6: 41777834-41777834 on Build GRCh38
rs1004637005 Chr.6: 41778254-41778254 on Build GRCh38
rs951854772 Chr.6: 41778045-41778045 on Build GRCh38
rs927990164 Chr.6: 41778292-41778292 on Build GRCh38
rs192151144 Chr.6: 41777862-41777862 on Build GRCh38
rs9471651 Chr.6: 41778166-41778166 on Build GRCh38
rs373962230 Chr.6: 41777927-41777927 on Build GRCh38
rs897010203 Chr.6: 41778095-41778095 on Build GRCh38
rs984673778 Chr.6: 41778048-41778048 on Build GRCh38
rs370116583 Chr.6: 41777915-41777915 on Build GRCh38
rs969590998 Chr.6: 41778259-41778259 on Build GRCh38

View Full Product Details