Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2794721-2795283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA12
Entrez Gene ID
2768
Gene Name
G protein subunit alpha 12
Gene Aliases
NNX3, RMP, gep
UniGene
Hs.487341
Gene Chromosome Location
Chr.7: 2728105-2870656 on Build GRCh38
Transcripts
NM_001282441.1 NM_001293092.1 NM_007353.2 XM_011515288.2 XM_017012008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1568449 c.475A>G p.R159G Chr.7: 2794978-2794978 on Build GRCh38
1635265 c.367G>A p.E123K Chr.7: 2795086-2795086 on Build GRCh38
746284 c.487A>T p.I163F Chr.7: 2794966-2794966 on Build GRCh38
6177720 c.506G>A p.R169Q Chr.7: 2794947-2794947 on Build GRCh38
3637777 c.421C>T p.P141S Chr.7: 2795032-2795032 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770558705 Chr.7: 2795164-2795164 on Build GRCh38
rs746845598 Chr.7: 2795051-2795051 on Build GRCh38
rs576111020 Chr.7: 2794796-2794796 on Build GRCh38
rs2527686 Chr.7: 2794779-2794779 on Build GRCh38
rs778866783 Chr.7: 2795022-2795022 on Build GRCh38
rs754917183 Chr.7: 2795008-2795008 on Build GRCh38
rs955555358 Chr.7: 2795195-2795195 on Build GRCh38
rs1041978667 Chr.7: 2794948-2794948 on Build GRCh38
rs749273521 Chr.7: 2795003-2795003 on Build GRCh38
rs145512130 Chr.7: 2795092-2795092 on Build GRCh38
rs186566532 Chr.7: 2794884-2794884 on Build GRCh38
rs190617612 Chr.7: 2795050-2795050 on Build GRCh38
rs758106371 Chr.7: 2795126-2795126 on Build GRCh38
rs964076664 Chr.7: 2795238-2795238 on Build GRCh38
rs764331463 Chr.7: 2794929-2794929 on Build GRCh38
rs763287777 Chr.7: 2795060-2795060 on Build GRCh38
rs200966565 Chr.7: 2794892-2794892 on Build GRCh38
rs928483371 Chr.7: 2795252-2795252 on Build GRCh38
rs142768832 Chr.7: 2794941-2794941 on Build GRCh38
rs780830669 Chr.7: 2795135-2795135 on Build GRCh38
rs946000505 Chr.7: 2795237-2795237 on Build GRCh38
rs775950051 Chr.7: 2795190-2795190 on Build GRCh38
rs2230877 Chr.7: 2795120-2795120 on Build GRCh38
rs955381925 Chr.7: 2794973-2794973 on Build GRCh38
rs942721556 Chr.7: 2794901-2794901 on Build GRCh38
rs139108346 Chr.7: 2795203-2795203 on Build GRCh38
rs536277438 Chr.7: 2794758-2794758 on Build GRCh38
rs1047972234 Chr.7: 2794765-2794765 on Build GRCh38
rs990109432 Chr.7: 2795224-2795224 on Build GRCh38
rs771918639 Chr.7: 2795030-2795030 on Build GRCh38
rs2230877 Chr.7: 2795120-2795120 on Build GRCh38
rs770378497 Chr.7: 2795039-2795039 on Build GRCh38
rs751704187 Chr.7: 2795074-2795074 on Build GRCh38
rs2533886 Chr.7: 2795235-2795235 on Build GRCh38
rs777924151 Chr.7: 2794914-2794914 on Build GRCh38
rs764422187 Chr.7: 2795072-2795072 on Build GRCh38
rs775323250 Chr.7: 2794873-2794873 on Build GRCh38
rs867168568 Chr.7: 2794875-2794875 on Build GRCh38
rs758327437 Chr.7: 2794814-2794814 on Build GRCh38
rs758572740 Chr.7: 2794913-2794913 on Build GRCh38
rs765610501 Chr.7: 2794997-2794997 on Build GRCh38
rs974310051 Chr.7: 2795250-2795251 on Build GRCh38
rs977350307 Chr.7: 2795247-2795247 on Build GRCh38
rs749152313 Chr.7: 2794942-2794942 on Build GRCh38
rs773978036 Chr.7: 2794988-2794988 on Build GRCh38
rs886094147 Chr.7: 2794778-2794778 on Build GRCh38
rs879083916 Chr.7: 2795042-2795042 on Build GRCh38
rs560890439 Chr.7: 2794985-2794985 on Build GRCh38
rs962929431 Chr.7: 2794785-2794785 on Build GRCh38
rs1047457263 Chr.7: 2795107-2795107 on Build GRCh38
rs572992848 Chr.7: 2794762-2794762 on Build GRCh38
rs760099419 Chr.7: 2794986-2794986 on Build GRCh38
rs894208409 Chr.7: 2795071-2795071 on Build GRCh38
rs2230878 Chr.7: 2794943-2794943 on Build GRCh38
rs749133463 Chr.7: 2794918-2794918 on Build GRCh38
rs372701112 Chr.7: 2795143-2795143 on Build GRCh38
rs576875175 Chr.7: 2794818-2794818 on Build GRCh38
rs918724170 Chr.7: 2794791-2794791 on Build GRCh38
rs756146751 Chr.7: 2794925-2794925 on Build GRCh38
rs757901134 Chr.7: 2794881-2794882 on Build GRCh38
rs775896855 Chr.7: 2795053-2795053 on Build GRCh38
rs754971607 Chr.7: 2795110-2795110 on Build GRCh38
rs181093880 Chr.7: 2794864-2794864 on Build GRCh38
rs777583728 Chr.7: 2795127-2795127 on Build GRCh38
rs140101989 Chr.7: 2795029-2795029 on Build GRCh38
rs12721527 Chr.7: 2795034-2795034 on Build GRCh38
rs12721525 Chr.7: 2794955-2794955 on Build GRCh38
rs12721526 Chr.7: 2794852-2794852 on Build GRCh38
rs994595795 Chr.7: 2794764-2794764 on Build GRCh38
rs373673209 Chr.7: 2795157-2795157 on Build GRCh38
rs751715632 Chr.7: 2795002-2795002 on Build GRCh38
rs2230878 Chr.7: 2794943-2794943 on Build GRCh38
rs531842185 Chr.7: 2795198-2795198 on Build GRCh38
rs778991320 Chr.7: 2795117-2795117 on Build GRCh38
rs955612091 Chr.7: 2795226-2795226 on Build GRCh38
rs768347396 Chr.7: 2795000-2795000 on Build GRCh38
rs745503501 Chr.7: 2795218-2795218 on Build GRCh38
rs367724356 Chr.7: 2794863-2794863 on Build GRCh38
rs972905092 Chr.7: 2794787-2794787 on Build GRCh38
rs746997653 Chr.7: 2794915-2794915 on Build GRCh38
rs368937953 Chr.7: 2795175-2795175 on Build GRCh38
rs1010507934 Chr.7: 2795191-2795191 on Build GRCh38
rs766968121 Chr.7: 2794987-2794987 on Build GRCh38
rs767520343 Chr.7: 2795081-2795081 on Build GRCh38
rs771794152 Chr.7: 2794982-2794982 on Build GRCh38
rs1018246637 Chr.7: 2795236-2795236 on Build GRCh38
rs77711396 Chr.7: 2795183-2795183 on Build GRCh38
rs571763604 Chr.7: 2795213-2795213 on Build GRCh38
rs747827635 Chr.7: 2794951-2794951 on Build GRCh38
rs752999465 Chr.7: 2794999-2794999 on Build GRCh38
rs777305261 Chr.7: 2795154-2795157 on Build GRCh38
rs78999582 Chr.7: 2794808-2794808 on Build GRCh38
rs774244411 Chr.7: 2794947-2794947 on Build GRCh38
rs759995518 Chr.7: 2794989-2794989 on Build GRCh38
rs772726296 Chr.7: 2795036-2795036 on Build GRCh38
rs752948556 Chr.7: 2794896-2794896 on Build GRCh38
rs746569487 Chr.7: 2795037-2795037 on Build GRCh38

View Full Product Details