Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122077695-122078229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASS
Entrez Gene ID
10157
Gene Name
aminoadipate-semialdehyde synthase
Gene Aliases
LKR/SDH, LKRSDH, LORSDH
UniGene
Hs.156738
Gene Chromosome Location
Chr.7: 122073544-122144290 on Build GRCh38
Transcripts
NM_005763.3 XM_011515725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1636983 c.2505G>A p.M835I Chr.7: 122077995-122077995 on Build GRCh38
4153790 c.2620G>A p.V874M Chr.7: 122077880-122077880 on Build GRCh38
598864 c.2565G>A p.T855T Chr.7: 122077935-122077935 on Build GRCh38
4168145 c.2564C>T p.T855M Chr.7: 122077936-122077936 on Build GRCh38
3995323 c.2584G>T p.G862W Chr.7: 122077916-122077916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147476318 Chr.7: 122077865-122077866 on Build GRCh38
rs117573737 Chr.7: 122078137-122078137 on Build GRCh38
rs760663934 Chr.7: 122077808-122077808 on Build GRCh38
rs150593459 Chr.7: 122077758-122077758 on Build GRCh38
rs1043080781 Chr.7: 122077987-122077987 on Build GRCh38
rs768298788 Chr.7: 122077866-122077866 on Build GRCh38
rs961040266 Chr.7: 122077766-122077766 on Build GRCh38
rs531058371 Chr.7: 122077750-122077750 on Build GRCh38
rs993503401 Chr.7: 122077751-122077751 on Build GRCh38
rs777824271 Chr.7: 122077917-122077917 on Build GRCh38
rs905307693 Chr.7: 122077775-122077775 on Build GRCh38
rs761402330 Chr.7: 122078105-122078105 on Build GRCh38
rs771797768 Chr.7: 122077981-122077981 on Build GRCh38
rs1014474544 Chr.7: 122078094-122078094 on Build GRCh38
rs764183023 Chr.7: 122078053-122078053 on Build GRCh38
rs891766148 Chr.7: 122077929-122077929 on Build GRCh38
rs746330018 Chr.7: 122077865-122077865 on Build GRCh38
rs777448610 Chr.7: 122078023-122078032 on Build GRCh38
rs149527213 Chr.7: 122077977-122077977 on Build GRCh38
rs1034498269 Chr.7: 122078032-122078032 on Build GRCh38
rs587777121 Chr.7: 122077833-122077837 on Build GRCh38
rs766283847 Chr.7: 122077809-122077809 on Build GRCh38
rs760432920 Chr.7: 122077872-122077872 on Build GRCh38
rs1010186650 Chr.7: 122077952-122077952 on Build GRCh38
rs746385156 Chr.7: 122077964-122077964 on Build GRCh38
rs530285419 Chr.7: 122077862-122077862 on Build GRCh38
rs1034958206 Chr.7: 122077743-122077743 on Build GRCh38
rs773110350 Chr.7: 122077870-122077870 on Build GRCh38
rs1003460814 Chr.7: 122078103-122078103 on Build GRCh38
rs1025596306 Chr.7: 122077833-122077833 on Build GRCh38
rs367958255 Chr.7: 122077786-122077786 on Build GRCh38
rs751788580 Chr.7: 122077928-122077928 on Build GRCh38
rs771610987 Chr.7: 122077868-122077868 on Build GRCh38
rs566866368 Chr.7: 122077988-122077988 on Build GRCh38
rs994047002 Chr.7: 122078144-122078144 on Build GRCh38
rs762971045 Chr.7: 122078042-122078042 on Build GRCh38
rs765424237 Chr.7: 122077895-122077895 on Build GRCh38
rs373242380 Chr.7: 122077838-122077838 on Build GRCh38
rs1001709486 Chr.7: 122077739-122077739 on Build GRCh38
rs770715430 Chr.7: 122078012-122078012 on Build GRCh38
rs757721535 Chr.7: 122077943-122077943 on Build GRCh38
rs976504512 Chr.7: 122077980-122077980 on Build GRCh38
rs747590381 Chr.7: 122077979-122077979 on Build GRCh38
rs759527383 Chr.7: 122078015-122078015 on Build GRCh38
rs752702037 Chr.7: 122077904-122077904 on Build GRCh38
rs972977767 Chr.7: 122077828-122077828 on Build GRCh38
rs1003600021 Chr.7: 122077781-122077781 on Build GRCh38
rs1035108976 Chr.7: 122078122-122078122 on Build GRCh38
rs960485415 Chr.7: 122078086-122078086 on Build GRCh38
rs943946020 Chr.7: 122077881-122077881 on Build GRCh38
rs779193257 Chr.7: 122077747-122077747 on Build GRCh38
rs769437600 Chr.7: 122077740-122077740 on Build GRCh38
rs764775526 Chr.7: 122078111-122078111 on Build GRCh38
rs116061308 Chr.7: 122077880-122077880 on Build GRCh38
rs765058679 Chr.7: 122078017-122078017 on Build GRCh38
rs760261179 Chr.7: 122077991-122077991 on Build GRCh38
rs1045295677 Chr.7: 122078072-122078072 on Build GRCh38
rs878936449 Chr.7: 122077978-122077978 on Build GRCh38
rs201639775 Chr.7: 122078028-122078028 on Build GRCh38
rs747351819 Chr.7: 122077845-122077845 on Build GRCh38
rs750181686 Chr.7: 122077796-122077796 on Build GRCh38
rs894083594 Chr.7: 122078039-122078039 on Build GRCh38
rs149105850 Chr.7: 122077868-122077869 on Build GRCh38
rs972426537 Chr.7: 122077778-122077778 on Build GRCh38
rs766111824 Chr.7: 122077875-122077875 on Build GRCh38
rs1015177150 Chr.7: 122078097-122078097 on Build GRCh38
rs778255100 Chr.7: 122077841-122077841 on Build GRCh38
rs778798520 Chr.7: 122077818-122077818 on Build GRCh38
rs771603825 Chr.7: 122077850-122077850 on Build GRCh38
rs759400940 Chr.7: 122077891-122077891 on Build GRCh38
rs562175564 Chr.7: 122078059-122078059 on Build GRCh38
rs565209129 Chr.7: 122077727-122077727 on Build GRCh38
rs138726205 Chr.7: 122077906-122077906 on Build GRCh38
rs752968291 Chr.7: 122077821-122077821 on Build GRCh38
rs905214344 Chr.7: 122078095-122078095 on Build GRCh38
rs370656085 Chr.7: 122077963-122077963 on Build GRCh38
rs374098864 Chr.7: 122077962-122077962 on Build GRCh38
rs1025881247 Chr.7: 122078177-122078177 on Build GRCh38
rs770202556 Chr.7: 122077972-122077972 on Build GRCh38
rs776405627 Chr.7: 122078013-122078013 on Build GRCh38
rs371822556 Chr.7: 122077810-122077810 on Build GRCh38
rs758173965 Chr.7: 122078018-122078021 on Build GRCh38
rs761594354 Chr.7: 122077867-122077867 on Build GRCh38
rs545417330 Chr.7: 122077721-122077721 on Build GRCh38

View Full Product Details