Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143397180-143397779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA1
Entrez Gene ID
2041
Gene Name
EPH receptor A1
Gene Aliases
EPH, EPHT, EPHT1
UniGene
Hs.89839
Gene Chromosome Location
Chr.7: 143390813-143408892 on Build GRCh38
Transcripts
NM_005232.4 XM_006715880.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661675 c.1672G>A p.G558S Chr.7: 143397601-143397601 on Build GRCh38
1548900 c.1693A>C p.I565L Chr.7: 143397580-143397580 on Build GRCh38
5439194 c.1747G>C p.D583H Chr.7: 143397328-143397328 on Build GRCh38
364760 c.1651G>T p.V551F Chr.7: 143397622-143397622 on Build GRCh38
3878959 c.1706G>A p.R569Q Chr.7: 143397567-143397567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780288031 Chr.7: 143397352-143397352 on Build GRCh38
rs776663355 Chr.7: 143397659-143397659 on Build GRCh38
rs185706866 Chr.7: 143397545-143397545 on Build GRCh38
rs564687025 Chr.7: 143397490-143397490 on Build GRCh38
rs777514852 Chr.7: 143397411-143397411 on Build GRCh38
rs762715134 Chr.7: 143397235-143397238 on Build GRCh38
rs756443190 Chr.7: 143397350-143397350 on Build GRCh38
rs188438809 Chr.7: 143397233-143397233 on Build GRCh38
rs1046462373 Chr.7: 143397714-143397714 on Build GRCh38
rs111778416 Chr.7: 143397600-143397600 on Build GRCh38
rs766683764 Chr.7: 143397540-143397540 on Build GRCh38
rs752138390 Chr.7: 143397690-143397690 on Build GRCh38
rs572481807 Chr.7: 143397376-143397376 on Build GRCh38
rs925917171 Chr.7: 143397715-143397715 on Build GRCh38
rs775980315 Chr.7: 143397677-143397677 on Build GRCh38
rs45497499 Chr.7: 143397542-143397542 on Build GRCh38
rs555414886 Chr.7: 143397574-143397574 on Build GRCh38
rs201508170 Chr.7: 143397541-143397541 on Build GRCh38
rs760509682 Chr.7: 143397291-143397291 on Build GRCh38
rs555587376 Chr.7: 143397330-143397330 on Build GRCh38
rs1016338802 Chr.7: 143397569-143397569 on Build GRCh38
rs867344192 Chr.7: 143397363-143397363 on Build GRCh38
rs774390706 Chr.7: 143397648-143397648 on Build GRCh38
rs142999380 Chr.7: 143397566-143397566 on Build GRCh38
rs759805828 Chr.7: 143397663-143397663 on Build GRCh38
rs535384858 Chr.7: 143397229-143397229 on Build GRCh38
rs1054227824 Chr.7: 143397273-143397273 on Build GRCh38
rs146724598 Chr.7: 143397235-143397236 on Build GRCh38
rs1009953141 Chr.7: 143397285-143397285 on Build GRCh38
rs760467922 Chr.7: 143397538-143397539 on Build GRCh38
rs779732304 Chr.7: 143397701-143397701 on Build GRCh38
rs776941825 Chr.7: 143397307-143397307 on Build GRCh38
rs910708895 Chr.7: 143397460-143397460 on Build GRCh38
rs1052285437 Chr.7: 143397306-143397306 on Build GRCh38
rs773268230 Chr.7: 143397560-143397560 on Build GRCh38
rs776282653 Chr.7: 143397513-143397513 on Build GRCh38
rs768560395 Chr.7: 143397647-143397647 on Build GRCh38
rs910648566 Chr.7: 143397535-143397535 on Build GRCh38
rs1000673503 Chr.7: 143397247-143397247 on Build GRCh38
rs750667620 Chr.7: 143397346-143397346 on Build GRCh38
rs199981664 Chr.7: 143397297-143397297 on Build GRCh38
rs202068423 Chr.7: 143397538-143397538 on Build GRCh38
rs776947009 Chr.7: 143397567-143397567 on Build GRCh38
rs373850526 Chr.7: 143397526-143397526 on Build GRCh38
rs754801581 Chr.7: 143397704-143397704 on Build GRCh38
rs775727329 Chr.7: 143397271-143397271 on Build GRCh38
rs774012786 Chr.7: 143397538-143397538 on Build GRCh38
rs34178823 Chr.7: 143397322-143397322 on Build GRCh38
rs200563282 Chr.7: 143397537-143397538 on Build GRCh38
rs1002258055 Chr.7: 143397358-143397358 on Build GRCh38
rs200212760 Chr.7: 143397568-143397568 on Build GRCh38
rs367732092 Chr.7: 143397700-143397700 on Build GRCh38
rs780668174 Chr.7: 143397615-143397615 on Build GRCh38
rs371664721 Chr.7: 143397268-143397268 on Build GRCh38
rs764663016 Chr.7: 143397685-143397685 on Build GRCh38
rs138816415 Chr.7: 143397276-143397276 on Build GRCh38
rs372030742 Chr.7: 143397542-143397542 on Build GRCh38
rs919388181 Chr.7: 143397476-143397476 on Build GRCh38
rs138347396 Chr.7: 143397623-143397623 on Build GRCh38
rs1013254568 Chr.7: 143397342-143397342 on Build GRCh38
rs866192433 Chr.7: 143397316-143397316 on Build GRCh38
rs537746418 Chr.7: 143397269-143397269 on Build GRCh38
rs747022455 Chr.7: 143397649-143397649 on Build GRCh38
rs765617275 Chr.7: 143397671-143397671 on Build GRCh38
rs775607178 Chr.7: 143397237-143397238 on Build GRCh38
rs771742971 Chr.7: 143397406-143397406 on Build GRCh38
rs769337411 Chr.7: 143397582-143397599 on Build GRCh38
rs1052212564 Chr.7: 143397338-143397338 on Build GRCh38
rs1030816724 Chr.7: 143397248-143397248 on Build GRCh38
rs201923054 Chr.7: 143397377-143397377 on Build GRCh38
rs763458946 Chr.7: 143397678-143397678 on Build GRCh38
rs765726381 Chr.7: 143397564-143397564 on Build GRCh38
rs919457417 Chr.7: 143397296-143397296 on Build GRCh38
rs770689905 Chr.7: 143397413-143397413 on Build GRCh38
rs780762492 Chr.7: 143397539-143397540 on Build GRCh38
rs976791042 Chr.7: 143397396-143397396 on Build GRCh38
rs372400722 Chr.7: 143397417-143397417 on Build GRCh38
rs1033504857 Chr.7: 143397310-143397310 on Build GRCh38
rs138374184 Chr.7: 143397323-143397323 on Build GRCh38
rs574748801 Chr.7: 143397331-143397331 on Build GRCh38
rs770650418 Chr.7: 143397293-143397293 on Build GRCh38
rs983868627 Chr.7: 143397325-143397325 on Build GRCh38
rs62472725 Chr.7: 143397431-143397431 on Build GRCh38
rs777423438 Chr.7: 143397543-143397543 on Build GRCh38
rs370950145 Chr.7: 143397519-143397519 on Build GRCh38
rs775349109 Chr.7: 143397546-143397546 on Build GRCh38
rs1040036628 Chr.7: 143397232-143397232 on Build GRCh38
rs112594963 Chr.7: 143397708-143397708 on Build GRCh38
rs747635716 Chr.7: 143397390-143397390 on Build GRCh38
rs904925689 Chr.7: 143397548-143397548 on Build GRCh38
rs775445317 Chr.7: 143397528-143397528 on Build GRCh38
rs889157136 Chr.7: 143397225-143397225 on Build GRCh38
rs766091682 Chr.7: 143397730-143397730 on Build GRCh38
rs771069813 Chr.7: 143397655-143397655 on Build GRCh38
rs756672063 Chr.7: 143397696-143397696 on Build GRCh38
rs760702958 Chr.7: 143397288-143397288 on Build GRCh38
rs779820434 Chr.7: 143397624-143397624 on Build GRCh38
rs987993560 Chr.7: 143397752-143397752 on Build GRCh38
rs761841029 Chr.7: 143397531-143397531 on Build GRCh38
rs765284054 Chr.7: 143397324-143397324 on Build GRCh38
rs868237069 Chr.7: 143397581-143397581 on Build GRCh38
rs772324191 Chr.7: 143397448-143397448 on Build GRCh38
rs542197026 Chr.7: 143397341-143397341 on Build GRCh38
rs1013322988 Chr.7: 143397251-143397251 on Build GRCh38
rs936860521 Chr.7: 143397632-143397632 on Build GRCh38
rs79446026 Chr.7: 143397544-143397544 on Build GRCh38
rs961085961 Chr.7: 143397385-143397385 on Build GRCh38
rs909473186 Chr.7: 143397284-143397284 on Build GRCh38
rs542834261 Chr.7: 143397711-143397711 on Build GRCh38
rs750578123 Chr.7: 143397539-143397539 on Build GRCh38
rs560427865 Chr.7: 143397357-143397357 on Build GRCh38
rs749142175 Chr.7: 143397644-143397644 on Build GRCh38
rs181565708 Chr.7: 143397504-143397504 on Build GRCh38
rs535509887 Chr.7: 143397622-143397622 on Build GRCh38
rs758966923 Chr.7: 143397575-143397575 on Build GRCh38
rs35719334 Chr.7: 143397351-143397351 on Build GRCh38
rs755432125 Chr.7: 143397256-143397269 on Build GRCh38
rs45453497 Chr.7: 143397722-143397722 on Build GRCh38
rs954869349 Chr.7: 143397494-143397494 on Build GRCh38
rs754945454 Chr.7: 143397526-143397526 on Build GRCh38
rs760861491 Chr.7: 143397563-143397563 on Build GRCh38
rs978908441 Chr.7: 143397712-143397712 on Build GRCh38
rs763454805 Chr.7: 143397702-143397702 on Build GRCh38
rs112960467 Chr.7: 143397215-143397215 on Build GRCh38
rs565761713 Chr.7: 143397249-143397249 on Build GRCh38

View Full Product Details