Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89709783-89710175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF20B
Entrez Gene ID
9585
Gene Name
kinesin family member 20B
Gene Aliases
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1
UniGene
Hs.240
Gene Chromosome Location
Chr.10: 89701590-89774943 on Build GRCh38
Transcripts
NM_001284259.1 NM_016195.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6255952 c.427G>T p.G143* Chr.10: 89710002-89710002 on Build GRCh38
921303 c.359G>A p.G120D Chr.10: 89709934-89709934 on Build GRCh38
6695033 c.437G>A p.R146H Chr.10: 89710012-89710012 on Build GRCh38
3441425 c.356T>C p.F119S Chr.10: 89709931-89709931 on Build GRCh38
921305 c.455G>A p.G152E Chr.10: 89710030-89710030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766306284 Chr.10: 89710092-89710092 on Build GRCh38
rs771970019 Chr.10: 89709949-89709949 on Build GRCh38
rs775411838 Chr.10: 89710031-89710031 on Build GRCh38
rs866463802 Chr.10: 89710060-89710060 on Build GRCh38
rs764627008 Chr.10: 89710067-89710067 on Build GRCh38
rs746740786 Chr.10: 89710038-89710038 on Build GRCh38
rs761829678 Chr.10: 89710064-89710064 on Build GRCh38
rs541129006 Chr.10: 89709942-89709942 on Build GRCh38
rs995424668 Chr.10: 89710052-89710053 on Build GRCh38
rs116079640 Chr.10: 89709988-89709988 on Build GRCh38
rs376662889 Chr.10: 89710028-89710028 on Build GRCh38
rs764542928 Chr.10: 89710110-89710110 on Build GRCh38
rs866641676 Chr.10: 89709837-89709837 on Build GRCh38
rs190543605 Chr.10: 89710006-89710006 on Build GRCh38
rs368756216 Chr.10: 89709905-89709905 on Build GRCh38
rs149777607 Chr.10: 89709972-89709972 on Build GRCh38
rs764831714 Chr.10: 89709993-89709993 on Build GRCh38
rs532340876 Chr.10: 89710117-89710117 on Build GRCh38
rs1013134298 Chr.10: 89710132-89710132 on Build GRCh38
rs886474438 Chr.10: 89710096-89710096 on Build GRCh38
rs372770283 Chr.10: 89710029-89710029 on Build GRCh38
rs991773364 Chr.10: 89709836-89709836 on Build GRCh38
rs769419161 Chr.10: 89709986-89709986 on Build GRCh38
rs746134088 Chr.10: 89709849-89709849 on Build GRCh38
rs773327403 Chr.10: 89710012-89710012 on Build GRCh38
rs775465092 Chr.10: 89709956-89709956 on Build GRCh38
rs115619054 Chr.10: 89709922-89709922 on Build GRCh38
rs1005385996 Chr.10: 89709921-89709921 on Build GRCh38
rs903288308 Chr.10: 89710036-89710036 on Build GRCh38
rs762584646 Chr.10: 89710100-89710100 on Build GRCh38
rs776389826 Chr.10: 89710057-89710057 on Build GRCh38
rs200906639 Chr.10: 89710059-89710059 on Build GRCh38
rs775299231 Chr.10: 89710091-89710091 on Build GRCh38
rs368664005 Chr.10: 89710011-89710011 on Build GRCh38
rs895857763 Chr.10: 89710121-89710121 on Build GRCh38
rs767867106 Chr.10: 89709999-89709999 on Build GRCh38
rs995393526 Chr.10: 89709833-89709833 on Build GRCh38
rs769714206 Chr.10: 89709980-89709980 on Build GRCh38
rs200805454 Chr.10: 89710004-89710004 on Build GRCh38
rs745500106 Chr.10: 89709905-89709905 on Build GRCh38
rs140764762 Chr.10: 89709940-89709940 on Build GRCh38
rs768501568 Chr.10: 89710055-89710055 on Build GRCh38
rs749990301 Chr.10: 89709994-89709994 on Build GRCh38
rs151174992 Chr.10: 89710145-89710145 on Build GRCh38
rs143233819 Chr.10: 89710107-89710107 on Build GRCh38
rs770357459 Chr.10: 89709909-89709909 on Build GRCh38
rs974192993 Chr.10: 89709904-89709904 on Build GRCh38
rs758091272 Chr.10: 89709997-89709997 on Build GRCh38
rs372714100 Chr.10: 89709936-89709936 on Build GRCh38
rs943951834 Chr.10: 89709986-89709986 on Build GRCh38
rs564911496 Chr.10: 89709875-89709875 on Build GRCh38
rs10881631 Chr.10: 89709819-89709819 on Build GRCh38
rs777899436 Chr.10: 89709917-89709917 on Build GRCh38
rs1046942620 Chr.10: 89710095-89710095 on Build GRCh38
rs1023228587 Chr.10: 89710135-89710135 on Build GRCh38
rs374626048 Chr.10: 89709975-89709975 on Build GRCh38
rs558842087 Chr.10: 89710149-89710149 on Build GRCh38
rs778078682 Chr.10: 89710009-89710009 on Build GRCh38
rs544973225 Chr.10: 89709850-89709850 on Build GRCh38
rs942538562 Chr.10: 89710021-89710021 on Build GRCh38

View Full Product Details