Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51061307-51061898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP2
Entrez Gene ID
81566
Gene Name
cysteine and serine rich nuclear protein 2
Gene Aliases
C12orf2, C12orf22, FAM130A1, PPP1R72, TAIP-12
UniGene
Hs.524425
Gene Chromosome Location
Chr.12: 51061205-51083671 on Build GRCh38
Transcripts
NM_030809.2 NR_045072.1 XM_006719621.2 XM_017019991.1 XM_011538163.2 XM_017019168.1

Gene Symbol
LETMD1
Entrez Gene ID
25875
Gene Name
LETM1 domain containing 1
Gene Aliases
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
UniGene
Hs.655272
Gene Chromosome Location
Chr.12: 51048230-51068984 on Build GRCh38
Transcripts
NM_030809.2 NR_045072.1 XM_006719621.2 XM_017019991.1 XM_011538163.2 XM_017019168.1

SNP Detail

SNP ID Chromosome Location
rs987118652 Chr.12: 51061349-51061349 on Build GRCh38
rs911519497 Chr.12: 51061354-51061354 on Build GRCh38
rs889549665 Chr.12: 51061396-51061396 on Build GRCh38
rs920479689 Chr.12: 51061391-51061391 on Build GRCh38
rs953105589 Chr.12: 51061338-51061341 on Build GRCh38
rs935823338 Chr.12: 51061831-51061831 on Build GRCh38
rs1050743723 Chr.12: 51061382-51061382 on Build GRCh38
rs7722 Chr.12: 51061621-51061621 on Build GRCh38
rs868709991 Chr.12: 51061446-51061446 on Build GRCh38
rs917277412 Chr.12: 51061798-51061798 on Build GRCh38
rs930458616 Chr.12: 51061393-51061393 on Build GRCh38
rs538506926 Chr.12: 51061778-51061778 on Build GRCh38
rs998267271 Chr.12: 51061377-51061377 on Build GRCh38
rs961944055 Chr.12: 51061375-51061375 on Build GRCh38
rs748209242 Chr.12: 51061551-51061552 on Build GRCh38
rs993258452 Chr.12: 51061633-51061633 on Build GRCh38
rs895810274 Chr.12: 51061493-51061493 on Build GRCh38
rs1006583845 Chr.12: 51061474-51061474 on Build GRCh38
rs571319882 Chr.12: 51061573-51061588 on Build GRCh38
rs568126113 Chr.12: 51061545-51061545 on Build GRCh38
rs1018599903 Chr.12: 51061574-51061574 on Build GRCh38
rs368406143 Chr.12: 51061352-51061352 on Build GRCh38
rs1027997087 Chr.12: 51061700-51061700 on Build GRCh38
rs946356309 Chr.12: 51061497-51061500 on Build GRCh38
rs779049916 Chr.12: 51061539-51061540 on Build GRCh38
rs925813311 Chr.12: 51061786-51061786 on Build GRCh38
rs376432410 Chr.12: 51061543-51061543 on Build GRCh38
rs995133956 Chr.12: 51061538-51061538 on Build GRCh38
rs937369935 Chr.12: 51061433-51061433 on Build GRCh38
rs545789091 Chr.12: 51061374-51061376 on Build GRCh38
rs888869689 Chr.12: 51061572-51061573 on Build GRCh38
rs546755375 Chr.12: 51061594-51061594 on Build GRCh38
rs574914569 Chr.12: 51061832-51061832 on Build GRCh38
rs775891222 Chr.12: 51061734-51061734 on Build GRCh38
rs746515000 Chr.12: 51061491-51061491 on Build GRCh38
rs548371527 Chr.12: 51061341-51061341 on Build GRCh38
rs980161786 Chr.12: 51061753-51061753 on Build GRCh38
rs1017753125 Chr.12: 51061510-51061510 on Build GRCh38
rs763461192 Chr.12: 51061802-51061802 on Build GRCh38
rs1042342661 Chr.12: 51061526-51061526 on Build GRCh38
rs149767308 Chr.12: 51061788-51061788 on Build GRCh38
rs970157151 Chr.12: 51061674-51061674 on Build GRCh38
rs372953324 Chr.12: 51061376-51061376 on Build GRCh38
rs559338795 Chr.12: 51061717-51061717 on Build GRCh38
rs917227109 Chr.12: 51061672-51061672 on Build GRCh38
rs992907754 Chr.12: 51061785-51061785 on Build GRCh38
rs540360883 Chr.12: 51061834-51061834 on Build GRCh38
rs937440382 Chr.12: 51061760-51061760 on Build GRCh38
rs913142833 Chr.12: 51061854-51061854 on Build GRCh38
rs904719957 Chr.12: 51061531-51061531 on Build GRCh38
rs376346945 Chr.12: 51061607-51061608 on Build GRCh38
rs569581719 Chr.12: 51061741-51061741 on Build GRCh38
rs770357610 Chr.12: 51061515-51061515 on Build GRCh38
rs777834044 Chr.12: 51061568-51061569 on Build GRCh38
rs776014460 Chr.12: 51061534-51061534 on Build GRCh38
rs191151666 Chr.12: 51061336-51061336 on Build GRCh38
rs777457778 Chr.12: 51061422-51061422 on Build GRCh38
rs1044875651 Chr.12: 51061407-51061414 on Build GRCh38
rs530230641 Chr.12: 51061589-51061589 on Build GRCh38
rs746336604 Chr.12: 51061546-51061546 on Build GRCh38
rs972030440 Chr.12: 51061641-51061645 on Build GRCh38
rs566892447 Chr.12: 51061615-51061615 on Build GRCh38

View Full Product Details