Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110040732-110041321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf76
Entrez Gene ID
400073
Gene Name
chromosome 12 open reading frame 76
Gene Aliases
-
UniGene
Hs.44817
Gene Chromosome Location
Chr.12: 110041177-110067695 on Build GRCh38
Transcripts
NM_207435.2 XM_017019297.1

SNP Detail

SNP ID Chromosome Location
rs559668326 Chr.12: 110041085-110041085 on Build GRCh38
rs768426266 Chr.12: 110041091-110041091 on Build GRCh38
rs952440434 Chr.12: 110041047-110041047 on Build GRCh38
rs747829059 Chr.12: 110041203-110041203 on Build GRCh38
rs1053218995 Chr.12: 110041204-110041204 on Build GRCh38
rs867347910 Chr.12: 110041170-110041170 on Build GRCh38
rs767574110 Chr.12: 110040983-110040983 on Build GRCh38
rs555294963 Chr.12: 110041258-110041261 on Build GRCh38
rs189145301 Chr.12: 110040816-110040816 on Build GRCh38
rs985438348 Chr.12: 110041050-110041050 on Build GRCh38
rs1053122948 Chr.12: 110040766-110040766 on Build GRCh38
rs572884888 Chr.12: 110040917-110040917 on Build GRCh38
rs898152941 Chr.12: 110041013-110041013 on Build GRCh38
rs114547587 Chr.12: 110041099-110041099 on Build GRCh38
rs1045029240 Chr.12: 110040982-110040982 on Build GRCh38
rs1044592347 Chr.12: 110041097-110041097 on Build GRCh38
rs558035208 Chr.12: 110040849-110040849 on Build GRCh38
rs972348500 Chr.12: 110040989-110040989 on Build GRCh38
rs577965874 Chr.12: 110041278-110041278 on Build GRCh38
rs947951075 Chr.12: 110040977-110040977 on Build GRCh38
rs544513006 Chr.12: 110041167-110041167 on Build GRCh38
rs1011613192 Chr.12: 110041277-110041277 on Build GRCh38
rs750539098 Chr.12: 110041062-110041062 on Build GRCh38
rs1019770948 Chr.12: 110041130-110041130 on Build GRCh38
rs771070972 Chr.12: 110041097-110041098 on Build GRCh38
rs375071961 Chr.12: 110041095-110041098 on Build GRCh38
rs975618227 Chr.12: 110041191-110041191 on Build GRCh38
rs995187368 Chr.12: 110041028-110041028 on Build GRCh38
rs533745946 Chr.12: 110040920-110040920 on Build GRCh38
rs947413573 Chr.12: 110040790-110040790 on Build GRCh38
rs918234049 Chr.12: 110040991-110040991 on Build GRCh38
rs555190931 Chr.12: 110041063-110041063 on Build GRCh38
rs1019286991 Chr.12: 110040866-110040866 on Build GRCh38
rs546663519 Chr.12: 110040761-110040761 on Build GRCh38
rs1056300788 Chr.12: 110040895-110040895 on Build GRCh38
rs954009952 Chr.12: 110040836-110040836 on Build GRCh38
rs552358449 Chr.12: 110040940-110040940 on Build GRCh38
rs909506879 Chr.12: 110041055-110041055 on Build GRCh38
rs756176184 Chr.12: 110040771-110040771 on Build GRCh38
rs569190474 Chr.12: 110041100-110041100 on Build GRCh38
rs538828814 Chr.12: 110040821-110040821 on Build GRCh38
rs891779644 Chr.12: 110041271-110041271 on Build GRCh38
rs369005366 Chr.12: 110040841-110040841 on Build GRCh38
rs74824676 Chr.12: 110041179-110041179 on Build GRCh38
rs977895101 Chr.12: 110040807-110040807 on Build GRCh38
rs1011891845 Chr.12: 110040802-110040802 on Build GRCh38

View Full Product Details