Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48961469-48962009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARIH2
Entrez Gene ID
10425
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases
ARI2, TRIAD1
UniGene
Hs.633601
Gene Chromosome Location
Chr.3: 48918820-48985541 on Build GRCh38
Transcripts
NM_001317333.1 NM_001317334.1 NM_006321.3 XM_011533265.1 XM_011533266.1 XM_011533267.1 XM_011533268.1 XM_011533270.1 XM_011533271.1 XM_011533273.1 XM_011533275.2 XM_017005531.1 XM_017005532.1 XM_017005533.1 XM_017005534.1 XM_017005535.1 XM_017005536.1 XM_017005537.1 XM_017005538.1 XM_017005539.1 XM_017005540.1 XM_017005541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3719950 c.323+1G>A p.? Chr.3: 48961680-48961680 on Build GRCh38
5864762 c.308C>T p.S103L Chr.3: 48961664-48961664 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745522148 Chr.3: 48961686-48961686 on Build GRCh38
rs916983666 Chr.3: 48961555-48961555 on Build GRCh38
rs984476867 Chr.3: 48961810-48961810 on Build GRCh38
rs4974082 Chr.3: 48961890-48961890 on Build GRCh38
rs950188287 Chr.3: 48961578-48961578 on Build GRCh38
rs773832423 Chr.3: 48961709-48961709 on Build GRCh38
rs944736111 Chr.3: 48961826-48961826 on Build GRCh38
rs113007676 Chr.3: 48961772-48961772 on Build GRCh38
rs201120422 Chr.3: 48961698-48961698 on Build GRCh38
rs781166831 Chr.3: 48961601-48961601 on Build GRCh38
rs534794366 Chr.3: 48961865-48961865 on Build GRCh38
rs961638614 Chr.3: 48961838-48961838 on Build GRCh38
rs917305913 Chr.3: 48961894-48961894 on Build GRCh38
rs958710060 Chr.3: 48961539-48961539 on Build GRCh38
rs563909254 Chr.3: 48961550-48961550 on Build GRCh38
rs377524338 Chr.3: 48961730-48961730 on Build GRCh38
rs1054293102 Chr.3: 48961748-48961748 on Build GRCh38
rs754780534 Chr.3: 48961589-48961589 on Build GRCh38
rs180762235 Chr.3: 48961729-48961729 on Build GRCh38
rs751509219 Chr.3: 48961588-48961589 on Build GRCh38
rs763130779 Chr.3: 48961586-48961586 on Build GRCh38
rs570508737 Chr.3: 48961776-48961776 on Build GRCh38
rs751653276 Chr.3: 48961722-48961722 on Build GRCh38
rs558009558 Chr.3: 48961966-48961966 on Build GRCh38
rs777806461 Chr.3: 48961618-48961618 on Build GRCh38
rs755186223 Chr.3: 48961597-48961597 on Build GRCh38
rs898086957 Chr.3: 48961870-48961870 on Build GRCh38
rs900934777 Chr.3: 48961753-48961753 on Build GRCh38
rs369493966 Chr.3: 48961574-48961574 on Build GRCh38
rs771671771 Chr.3: 48961695-48961695 on Build GRCh38
rs894682347 Chr.3: 48961972-48961972 on Build GRCh38
rs757489619 Chr.3: 48961653-48961653 on Build GRCh38
rs561875846 Chr.3: 48961591-48961591 on Build GRCh38
rs751885858 Chr.3: 48961596-48961596 on Build GRCh38
rs763353636 Chr.3: 48961714-48961714 on Build GRCh38
rs750869060 Chr.3: 48961581-48961581 on Build GRCh38
rs1027785948 Chr.3: 48961523-48961523 on Build GRCh38
rs752495075 Chr.3: 48961604-48961604 on Build GRCh38
rs1028316099 Chr.3: 48961905-48961905 on Build GRCh38
rs781000402 Chr.3: 48961594-48961597 on Build GRCh38
rs754016241 Chr.3: 48961639-48961642 on Build GRCh38
rs1016934980 Chr.3: 48961817-48961817 on Build GRCh38
rs1028182322 Chr.3: 48961765-48961765 on Build GRCh38
rs1036299141 Chr.3: 48961858-48961858 on Build GRCh38
rs770534282 Chr.3: 48961636-48961636 on Build GRCh38
rs994986992 Chr.3: 48961519-48961519 on Build GRCh38
rs762286886 Chr.3: 48961706-48961706 on Build GRCh38
rs921071769 Chr.3: 48961763-48961763 on Build GRCh38
rs368954794 Chr.3: 48961624-48961624 on Build GRCh38
rs1044557813 Chr.3: 48961638-48961638 on Build GRCh38
rs529068228 Chr.3: 48961697-48961697 on Build GRCh38
rs368801554 Chr.3: 48961554-48961554 on Build GRCh38
rs752831036 Chr.3: 48961731-48961731 on Build GRCh38
rs766574090 Chr.3: 48961721-48961721 on Build GRCh38
rs762257343 Chr.3: 48961569-48961569 on Build GRCh38
rs893501840 Chr.3: 48961719-48961719 on Build GRCh38
rs778339677 Chr.3: 48961672-48961672 on Build GRCh38
rs552582294 Chr.3: 48961756-48961756 on Build GRCh38
rs778904017 Chr.3: 48961708-48961709 on Build GRCh38
rs932467275 Chr.3: 48961775-48961775 on Build GRCh38
rs773592380 Chr.3: 48961900-48961900 on Build GRCh38
rs201600501 Chr.3: 48961708-48961708 on Build GRCh38
rs144199771 Chr.3: 48961617-48961617 on Build GRCh38
rs1013483303 Chr.3: 48961974-48961974 on Build GRCh38

View Full Product Details