Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24815328-24815881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPO
Entrez Gene ID
79172
Gene Name
centromere protein O
Gene Aliases
CENP-O, ICEN-36, MCM21R
UniGene
Hs.731569
Gene Chromosome Location
Chr.2: 24793115-24822376 on Build GRCh38
Transcripts
NM_001199803.2 NM_001322101.1 NM_024322.3 NR_136182.1 NR_136183.1 NR_136184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6900616 c.480A>G p.A160A Chr.2: 24815660-24815660 on Build GRCh38
6626676 c.431T>C p.I144T Chr.2: 24815611-24815611 on Build GRCh38
4312106 c.572T>C p.L191P Chr.2: 24815752-24815752 on Build GRCh38
3702167 c.375C>T p.C125C Chr.2: 24815537-24815537 on Build GRCh38
7167120 c.461C>G p.P154R Chr.2: 24815641-24815641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201818285 Chr.2: 24815603-24815603 on Build GRCh38
rs201667046 Chr.2: 24815781-24815781 on Build GRCh38
rs1047852366 Chr.2: 24815616-24815616 on Build GRCh38
rs780282930 Chr.2: 24815717-24815718 on Build GRCh38
rs767176362 Chr.2: 24815772-24815772 on Build GRCh38
rs749161909 Chr.2: 24815529-24815529 on Build GRCh38
rs372578088 Chr.2: 24815705-24815705 on Build GRCh38
rs954584693 Chr.2: 24815468-24815468 on Build GRCh38
rs545448123 Chr.2: 24815553-24815553 on Build GRCh38
rs941815101 Chr.2: 24815845-24815845 on Build GRCh38
rs928805515 Chr.2: 24815356-24815356 on Build GRCh38
rs960248524 Chr.2: 24815565-24815565 on Build GRCh38
rs750011914 Chr.2: 24815476-24815476 on Build GRCh38
rs963038038 Chr.2: 24815491-24815491 on Build GRCh38
rs758420677 Chr.2: 24815592-24815609 on Build GRCh38
rs747271055 Chr.2: 24815653-24815653 on Build GRCh38
rs745732110 Chr.2: 24815425-24815425 on Build GRCh38
rs780948304 Chr.2: 24815505-24815505 on Build GRCh38
rs762313736 Chr.2: 24815573-24815573 on Build GRCh38
rs776946111 Chr.2: 24815564-24815564 on Build GRCh38
rs571329299 Chr.2: 24815789-24815789 on Build GRCh38
rs753857158 Chr.2: 24815711-24815711 on Build GRCh38
rs200386691 Chr.2: 24815666-24815666 on Build GRCh38
rs762036539 Chr.2: 24815466-24815466 on Build GRCh38
rs148113144 Chr.2: 24815752-24815752 on Build GRCh38
rs780764531 Chr.2: 24815613-24815613 on Build GRCh38
rs775321967 Chr.2: 24815783-24815783 on Build GRCh38
rs145335606 Chr.2: 24815521-24815521 on Build GRCh38
rs773751203 Chr.2: 24815678-24815678 on Build GRCh38
rs907664496 Chr.2: 24815618-24815618 on Build GRCh38
rs372623684 Chr.2: 24815787-24815787 on Build GRCh38
rs770888598 Chr.2: 24815638-24815638 on Build GRCh38
rs938819932 Chr.2: 24815374-24815374 on Build GRCh38
rs765600312 Chr.2: 24815470-24815470 on Build GRCh38
rs771683572 Chr.2: 24815850-24815853 on Build GRCh38
rs970102472 Chr.2: 24815758-24815758 on Build GRCh38
rs765920012 Chr.2: 24815608-24815608 on Build GRCh38
rs763267429 Chr.2: 24815684-24815684 on Build GRCh38
rs370086084 Chr.2: 24815723-24815723 on Build GRCh38
rs770267734 Chr.2: 24815677-24815677 on Build GRCh38
rs980392846 Chr.2: 24815586-24815586 on Build GRCh38
rs779742790 Chr.2: 24815486-24815486 on Build GRCh38
rs757216982 Chr.2: 24815795-24815795 on Build GRCh38
rs201226760 Chr.2: 24815630-24815630 on Build GRCh38
rs1031767373 Chr.2: 24815490-24815490 on Build GRCh38
rs978598087 Chr.2: 24815835-24815835 on Build GRCh38
rs967162131 Chr.2: 24815825-24815825 on Build GRCh38
rs956099930 Chr.2: 24815519-24815519 on Build GRCh38
rs771130957 Chr.2: 24815759-24815759 on Build GRCh38
rs756004056 Chr.2: 24815433-24815433 on Build GRCh38
rs1047492166 Chr.2: 24815375-24815375 on Build GRCh38
rs185944020 Chr.2: 24815410-24815410 on Build GRCh38
rs2082881 Chr.2: 24815399-24815399 on Build GRCh38
rs781321310 Chr.2: 24815731-24815731 on Build GRCh38
rs908033641 Chr.2: 24815819-24815819 on Build GRCh38
rs116574275 Chr.2: 24815427-24815427 on Build GRCh38
rs375613704 Chr.2: 24815706-24815706 on Build GRCh38
rs769651735 Chr.2: 24815517-24815517 on Build GRCh38
rs149340371 Chr.2: 24815748-24815748 on Build GRCh38
rs764090580 Chr.2: 24815708-24815708 on Build GRCh38
rs749068844 Chr.2: 24815633-24815633 on Build GRCh38
rs376462033 Chr.2: 24815563-24815563 on Build GRCh38
rs369252068 Chr.2: 24815607-24815607 on Build GRCh38
rs373113971 Chr.2: 24815679-24815679 on Build GRCh38
rs779847843 Chr.2: 24815463-24815463 on Build GRCh38
rs748289031 Chr.2: 24815673-24815673 on Build GRCh38
rs144007254 Chr.2: 24815547-24815547 on Build GRCh38
rs768804391 Chr.2: 24815556-24815556 on Build GRCh38
rs946106810 Chr.2: 24815593-24815593 on Build GRCh38
rs765146249 Chr.2: 24815803-24815803 on Build GRCh38
rs748135467 Chr.2: 24815516-24815516 on Build GRCh38
rs773815481 Chr.2: 24815688-24815688 on Build GRCh38
rs146897343 Chr.2: 24815745-24815745 on Build GRCh38
rs141799599 Chr.2: 24815645-24815645 on Build GRCh38
rs374134677 Chr.2: 24815725-24815725 on Build GRCh38
rs753762959 Chr.2: 24815791-24815791 on Build GRCh38
rs762768053 Chr.2: 24815696-24815696 on Build GRCh38
rs775639711 Chr.2: 24815434-24815434 on Build GRCh38
rs149023980 Chr.2: 24815604-24815604 on Build GRCh38
rs577755886 Chr.2: 24815420-24815420 on Build GRCh38
rs1040119942 Chr.2: 24815609-24815609 on Build GRCh38
rs950451603 Chr.2: 24815455-24815455 on Build GRCh38
rs774768704 Chr.2: 24815763-24815763 on Build GRCh38
rs200630774 Chr.2: 24815764-24815764 on Build GRCh38
rs879523023 Chr.2: 24815390-24815390 on Build GRCh38
rs547353937 Chr.2: 24815619-24815619 on Build GRCh38
rs763882377 Chr.2: 24815741-24815741 on Build GRCh38
rs1007549567 Chr.2: 24815395-24815395 on Build GRCh38
rs759227850 Chr.2: 24815693-24815693 on Build GRCh38
rs777393299 Chr.2: 24815825-24815826 on Build GRCh38
rs748199199 Chr.2: 24815738-24815738 on Build GRCh38
rs61732141 Chr.2: 24815612-24815612 on Build GRCh38
rs765800513 Chr.2: 24815580-24815580 on Build GRCh38
rs376487868 Chr.2: 24815728-24815728 on Build GRCh38
rs199499885 Chr.2: 24815788-24815788 on Build GRCh38
rs147672058 Chr.2: 24815695-24815695 on Build GRCh38
rs540115327 Chr.2: 24815537-24815537 on Build GRCh38
rs914557567 Chr.2: 24815540-24815540 on Build GRCh38
rs200842981 Chr.2: 24815534-24815534 on Build GRCh38
rs202236473 Chr.2: 24815687-24815687 on Build GRCh38
rs754559773 Chr.2: 24815492-24815492 on Build GRCh38

View Full Product Details