Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47160016-47160570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM2
Entrez Gene ID
805
Gene Name
calmodulin 2
Gene Aliases
CAMII, LQT15, PHKD, PHKD2, caM
UniGene
Hs.468442
Gene Chromosome Location
Chr.2: 47160082-47176936 on Build GRCh38
Transcripts
NM_001305624.1 NM_001305625.1 NM_001305626.1 NM_001743.5

SNP Detail

SNP ID Chromosome Location
rs999088772 Chr.2: 47160229-47160229 on Build GRCh38
rs1049555769 Chr.2: 47160185-47160185 on Build GRCh38
rs79550517 Chr.2: 47160230-47160230 on Build GRCh38
rs187511042 Chr.2: 47160187-47160187 on Build GRCh38
rs909810069 Chr.2: 47160467-47160467 on Build GRCh38
rs768674199 Chr.2: 47160244-47160244 on Build GRCh38
rs139812365 Chr.2: 47160151-47160151 on Build GRCh38
rs997925923 Chr.2: 47160426-47160426 on Build GRCh38
rs576376968 Chr.2: 47160380-47160380 on Build GRCh38
rs1019082576 Chr.2: 47160408-47160408 on Build GRCh38
rs562287437 Chr.2: 47160305-47160305 on Build GRCh38
rs74959150 Chr.2: 47160207-47160207 on Build GRCh38
rs562996983 Chr.2: 47160319-47160320 on Build GRCh38
rs915672191 Chr.2: 47160078-47160078 on Build GRCh38
rs1803209 Chr.2: 47160430-47160430 on Build GRCh38
rs998823939 Chr.2: 47160544-47160544 on Build GRCh38
rs919818636 Chr.2: 47160157-47160157 on Build GRCh38
rs1803207 Chr.2: 47160361-47160361 on Build GRCh38
rs912378943 Chr.2: 47160428-47160428 on Build GRCh38
rs116421442 Chr.2: 47160114-47160114 on Build GRCh38
rs146630528 Chr.2: 47160409-47160409 on Build GRCh38
rs193256689 Chr.2: 47160277-47160277 on Build GRCh38
rs973705705 Chr.2: 47160356-47160356 on Build GRCh38
rs11551428 Chr.2: 47160534-47160534 on Build GRCh38
rs17036320 Chr.2: 47160048-47160048 on Build GRCh38
rs971518658 Chr.2: 47160107-47160107 on Build GRCh38
rs1803206 Chr.2: 47160494-47160494 on Build GRCh38
rs981590320 Chr.2: 47160337-47160337 on Build GRCh38
rs144539242 Chr.2: 47160265-47160265 on Build GRCh38
rs932517589 Chr.2: 47160392-47160392 on Build GRCh38
rs15176 Chr.2: 47160284-47160284 on Build GRCh38
rs771735051 Chr.2: 47160060-47160060 on Build GRCh38
rs903242208 Chr.2: 47160528-47160528 on Build GRCh38
rs1025341795 Chr.2: 47160431-47160433 on Build GRCh38
rs971209145 Chr.2: 47160335-47160335 on Build GRCh38
rs919849831 Chr.2: 47160384-47160384 on Build GRCh38
rs976836709 Chr.2: 47160104-47160108 on Build GRCh38
rs965122764 Chr.2: 47160061-47160061 on Build GRCh38
rs951016454 Chr.2: 47160435-47160435 on Build GRCh38
rs543893798 Chr.2: 47160180-47160180 on Build GRCh38
rs889326779 Chr.2: 47160334-47160334 on Build GRCh38
rs185411626 Chr.2: 47160522-47160522 on Build GRCh38
rs908229242 Chr.2: 47160346-47160346 on Build GRCh38
rs908201068 Chr.2: 47160133-47160133 on Build GRCh38
rs1803208 Chr.2: 47160479-47160479 on Build GRCh38
rs1023513541 Chr.2: 47160307-47160307 on Build GRCh38
rs939986303 Chr.2: 47160135-47160135 on Build GRCh38
rs879396732 Chr.2: 47160488-47160488 on Build GRCh38
rs944840309 Chr.2: 47160495-47160495 on Build GRCh38
rs937152565 Chr.2: 47160179-47160182 on Build GRCh38
rs1013044680 Chr.2: 47160294-47160294 on Build GRCh38
rs1040411940 Chr.2: 47160208-47160208 on Build GRCh38
rs11551467 Chr.2: 47160299-47160299 on Build GRCh38
rs575041831 Chr.2: 47160437-47160437 on Build GRCh38
rs1018443498 Chr.2: 47160049-47160049 on Build GRCh38
rs1040842601 Chr.2: 47160510-47160510 on Build GRCh38
rs774217808 Chr.2: 47160434-47160434 on Build GRCh38
rs961314876 Chr.2: 47160353-47160353 on Build GRCh38
rs768275353 Chr.2: 47160458-47160458 on Build GRCh38
rs957587402 Chr.2: 47160287-47160287 on Build GRCh38
rs984074987 Chr.2: 47160456-47160456 on Build GRCh38
rs749206055 Chr.2: 47160224-47160224 on Build GRCh38
rs9581 Chr.2: 47160289-47160289 on Build GRCh38
rs1054633786 Chr.2: 47160327-47160327 on Build GRCh38
rs565042586 Chr.2: 47160456-47160456 on Build GRCh38
rs541631852 Chr.2: 47160164-47160164 on Build GRCh38
rs909749244 Chr.2: 47160170-47160170 on Build GRCh38
rs950142023 Chr.2: 47160253-47160253 on Build GRCh38
rs556314561 Chr.2: 47160103-47160103 on Build GRCh38
rs368324983 Chr.2: 47160043-47160043 on Build GRCh38

View Full Product Details