Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219420388-219420916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DES
Entrez Gene ID
1674
Gene Name
desmin
Gene Aliases
CSM1, CSM2, LGMD2R
UniGene
Hs.594952
Gene Chromosome Location
Chr.2: 219418377-219426739 on Build GRCh38
Transcripts
NM_001927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7364169 c.885G>A p.W295* Chr.2: 219420644-219420644 on Build GRCh38
5547428 c.928_930delAAC p.N311delN Chr.2: 219420858-219420860 on Build GRCh38
5584992 c.738G>A p.E246E Chr.2: 219420497-219420497 on Build GRCh38
5686074 c.915G>A p.Q305Q Chr.2: 219420845-219420845 on Build GRCh38
6510625 c.777C>A p.V259V Chr.2: 219420536-219420536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945223899 Chr.2: 219420674-219420674 on Build GRCh38
rs371053066 Chr.2: 219420823-219420823 on Build GRCh38
rs372532465 Chr.2: 219420596-219420596 on Build GRCh38
rs578191306 Chr.2: 219420854-219420854 on Build GRCh38
rs397516699 Chr.2: 219420660-219420661 on Build GRCh38
rs34337334 Chr.2: 219420864-219420864 on Build GRCh38
rs776181610 Chr.2: 219420436-219420436 on Build GRCh38
rs751348358 Chr.2: 219420888-219420888 on Build GRCh38
rs748742357 Chr.2: 219420873-219420873 on Build GRCh38
rs150370918 Chr.2: 219420551-219420551 on Build GRCh38
rs369144706 Chr.2: 219420584-219420584 on Build GRCh38
rs773937521 Chr.2: 219420693-219420693 on Build GRCh38
rs752188244 Chr.2: 219420446-219420446 on Build GRCh38
rs761475402 Chr.2: 219420592-219420592 on Build GRCh38
rs756017313 Chr.2: 219420481-219420481 on Build GRCh38
rs763599850 Chr.2: 219420581-219420581 on Build GRCh38
rs2669118 Chr.2: 219420767-219420767 on Build GRCh38
rs2666108 Chr.2: 219420766-219420766 on Build GRCh38
rs756434148 Chr.2: 219420863-219420863 on Build GRCh38
rs770258461 Chr.2: 219420576-219420576 on Build GRCh38
rs794728986 Chr.2: 219420642-219420642 on Build GRCh38
rs893676371 Chr.2: 219420423-219420423 on Build GRCh38
rs776035165 Chr.2: 219420676-219420676 on Build GRCh38
rs998215582 Chr.2: 219420470-219420470 on Build GRCh38
rs769213907 Chr.2: 219420866-219420866 on Build GRCh38
rs776995850 Chr.2: 219420571-219420571 on Build GRCh38
rs989384566 Chr.2: 219420740-219420740 on Build GRCh38
rs754852794 Chr.2: 219420839-219420839 on Build GRCh38
rs776125412 Chr.2: 219420578-219420578 on Build GRCh38
rs146755676 Chr.2: 219420644-219420644 on Build GRCh38
rs137945254 Chr.2: 219420673-219420673 on Build GRCh38
rs769305726 Chr.2: 219420689-219420689 on Build GRCh38
rs148947510 Chr.2: 219420865-219420865 on Build GRCh38
rs754941710 Chr.2: 219420650-219420650 on Build GRCh38
rs771116995 Chr.2: 219420668-219420668 on Build GRCh38
rs876657771 Chr.2: 219420612-219420612 on Build GRCh38
rs766252091 Chr.2: 219420867-219420867 on Build GRCh38
rs779875721 Chr.2: 219420622-219420622 on Build GRCh38
rs945046007 Chr.2: 219420786-219420786 on Build GRCh38
rs778826152 Chr.2: 219420842-219420842 on Build GRCh38
rs375906682 Chr.2: 219420502-219420502 on Build GRCh38
rs762634257 Chr.2: 219420661-219420662 on Build GRCh38
rs2632582 Chr.2: 219420682-219420682 on Build GRCh38
rs750160975 Chr.2: 219420598-219420598 on Build GRCh38
rs753745620 Chr.2: 219420634-219420634 on Build GRCh38
rs745667526 Chr.2: 219420476-219420476 on Build GRCh38
rs370385097 Chr.2: 219420672-219420672 on Build GRCh38
rs761229058 Chr.2: 219420701-219420701 on Build GRCh38
rs140375681 Chr.2: 219420487-219420487 on Build GRCh38
rs763753066 Chr.2: 219420454-219420454 on Build GRCh38
rs200300865 Chr.2: 219420688-219420688 on Build GRCh38
rs113252016 Chr.2: 219420492-219420492 on Build GRCh38
rs753655637 Chr.2: 219420834-219420834 on Build GRCh38
rs752747277 Chr.2: 219420846-219420846 on Build GRCh38
rs933928168 Chr.2: 219420760-219420760 on Build GRCh38
rs267607494 Chr.2: 219420580-219420580 on Build GRCh38
rs757975162 Chr.2: 219420451-219420451 on Build GRCh38
rs772117708 Chr.2: 219420501-219420501 on Build GRCh38
rs1053865406 Chr.2: 219420414-219420414 on Build GRCh38
rs747073500 Chr.2: 219420653-219420653 on Build GRCh38
rs759823001 Chr.2: 219420575-219420575 on Build GRCh38
rs1007681126 Chr.2: 219420519-219420519 on Build GRCh38
rs780995749 Chr.2: 219420469-219420469 on Build GRCh38
rs35419667 Chr.2: 219420499-219420500 on Build GRCh38
rs767115272 Chr.2: 219420781-219420781 on Build GRCh38
rs2666107 Chr.2: 219420449-219420449 on Build GRCh38
rs760197212 Chr.2: 219420892-219420892 on Build GRCh38
rs771455648 Chr.2: 219420874-219420874 on Build GRCh38
rs766123980 Chr.2: 219420791-219420791 on Build GRCh38
rs147327878 Chr.2: 219420544-219420544 on Build GRCh38
rs794728985 Chr.2: 219420591-219420591 on Build GRCh38
rs375709017 Chr.2: 219420853-219420853 on Build GRCh38
rs981782522 Chr.2: 219420628-219420628 on Build GRCh38
rs922526158 Chr.2: 219420732-219420732 on Build GRCh38
rs750024872 Chr.2: 219420782-219420782 on Build GRCh38
rs756897013 Chr.2: 219420465-219420465 on Build GRCh38
rs1058261 Chr.2: 219420587-219420587 on Build GRCh38
rs926406647 Chr.2: 219420685-219420685 on Build GRCh38
rs547425842 Chr.2: 219420440-219420440 on Build GRCh38
rs886044420 Chr.2: 219420527-219420527 on Build GRCh38
rs773192272 Chr.2: 219420761-219420761 on Build GRCh38
rs1042314538 Chr.2: 219420444-219420444 on Build GRCh38
rs139818514 Chr.2: 219420614-219420614 on Build GRCh38
rs994389035 Chr.2: 219420582-219420582 on Build GRCh38
rs201433470 Chr.2: 219420669-219420669 on Build GRCh38
rs62636491 Chr.2: 219420652-219420652 on Build GRCh38
rs889828698 Chr.2: 219420774-219420774 on Build GRCh38
rs144057476 Chr.2: 219420495-219420495 on Build GRCh38
rs41272701 Chr.2: 219420460-219420460 on Build GRCh38

View Full Product Details