Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216674804-216675400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP5
Entrez Gene ID
3488
Gene Name
insulin like growth factor binding protein 5
Gene Aliases
IBP5
UniGene
Hs.607212
Gene Chromosome Location
Chr.2: 216672105-216695549 on Build GRCh38
Transcripts
NM_000599.3

SNP Detail

SNP ID Chromosome Location
rs866148550 Chr.2: 216675126-216675126 on Build GRCh38
rs1057110093 Chr.2: 216674876-216674876 on Build GRCh38
rs753974375 Chr.2: 216675042-216675042 on Build GRCh38
rs148962226 Chr.2: 216675230-216675230 on Build GRCh38
rs575562095 Chr.2: 216675025-216675025 on Build GRCh38
rs557229910 Chr.2: 216675024-216675024 on Build GRCh38
rs898331368 Chr.2: 216674909-216674909 on Build GRCh38
rs1046085204 Chr.2: 216675258-216675258 on Build GRCh38
rs927367394 Chr.2: 216674852-216674852 on Build GRCh38
rs937953288 Chr.2: 216675352-216675352 on Build GRCh38
rs187844839 Chr.2: 216674915-216674915 on Build GRCh38
rs941805730 Chr.2: 216674930-216674930 on Build GRCh38
rs574494647 Chr.2: 216675015-216675015 on Build GRCh38
rs529952121 Chr.2: 216675067-216675067 on Build GRCh38
rs753897564 Chr.2: 216674966-216674969 on Build GRCh38
rs939960611 Chr.2: 216675174-216675174 on Build GRCh38
rs777017754 Chr.2: 216675309-216675309 on Build GRCh38
rs11575211 Chr.2: 216674916-216674916 on Build GRCh38
rs968973815 Chr.2: 216675346-216675346 on Build GRCh38
rs553062494 Chr.2: 216674987-216674987 on Build GRCh38
rs11575213 Chr.2: 216674853-216674853 on Build GRCh38
rs956313703 Chr.2: 216674955-216674955 on Build GRCh38
rs1021068751 Chr.2: 216675152-216675152 on Build GRCh38
rs569216878 Chr.2: 216674910-216674910 on Build GRCh38
rs1036891797 Chr.2: 216674906-216674906 on Build GRCh38
rs576873932 Chr.2: 216675218-216675218 on Build GRCh38
rs572825268 Chr.2: 216675195-216675195 on Build GRCh38
rs1020995227 Chr.2: 216674836-216674836 on Build GRCh38
rs918902038 Chr.2: 216674958-216674958 on Build GRCh38
rs771383222 Chr.2: 216675281-216675281 on Build GRCh38
rs779126397 Chr.2: 216675184-216675184 on Build GRCh38
rs910295074 Chr.2: 216674929-216674929 on Build GRCh38
rs1036945765 Chr.2: 216675183-216675183 on Build GRCh38
rs1008320267 Chr.2: 216674931-216674931 on Build GRCh38
rs145753482 Chr.2: 216675225-216675225 on Build GRCh38
rs562747121 Chr.2: 216675273-216675273 on Build GRCh38
rs867242935 Chr.2: 216675326-216675326 on Build GRCh38
rs143720329 Chr.2: 216675251-216675251 on Build GRCh38
rs1034511561 Chr.2: 216675373-216675373 on Build GRCh38
rs746765988 Chr.2: 216675328-216675328 on Build GRCh38
rs1010897699 Chr.2: 216674835-216674835 on Build GRCh38
rs980691042 Chr.2: 216675372-216675372 on Build GRCh38
rs377099243 Chr.2: 216674972-216674972 on Build GRCh38
rs922856764 Chr.2: 216674890-216674890 on Build GRCh38
rs1057183447 Chr.2: 216675149-216675149 on Build GRCh38
rs540328333 Chr.2: 216675196-216675196 on Build GRCh38
rs962930176 Chr.2: 216675228-216675228 on Build GRCh38
rs1029917719 Chr.2: 216675194-216675194 on Build GRCh38
rs966672627 Chr.2: 216675179-216675179 on Build GRCh38
rs985634539 Chr.2: 216675206-216675206 on Build GRCh38
rs2241194 Chr.2: 216675055-216675055 on Build GRCh38
rs1019225128 Chr.2: 216674934-216674934 on Build GRCh38
rs748641187 Chr.2: 216675010-216675010 on Build GRCh38
rs570839770 Chr.2: 216674956-216674956 on Build GRCh38
rs147605063 Chr.2: 216675274-216675274 on Build GRCh38
rs1002039731 Chr.2: 216675026-216675026 on Build GRCh38
rs11575210 Chr.2: 216674986-216674986 on Build GRCh38
rs11575212 Chr.2: 216674881-216674881 on Build GRCh38
rs1004121203 Chr.2: 216674885-216674885 on Build GRCh38
rs1011430913 Chr.2: 216675119-216675119 on Build GRCh38
rs758321394 Chr.2: 216675255-216675255 on Build GRCh38
rs777474746 Chr.2: 216675299-216675299 on Build GRCh38
rs535710310 Chr.2: 216675017-216675017 on Build GRCh38

View Full Product Details