Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34090156-34090730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC7
Entrez Gene ID
56851
Gene Name
ER membrane protein complex subunit 7
Gene Aliases
C11orf3, C15orf24, HT022, ORF1-FL1
UniGene
Hs.160565
Gene Chromosome Location
Chr.15: 34084022-34101852 on Build GRCh38
Transcripts
NM_020154.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817609 c.492A>G p.P164P Chr.15: 34090320-34090320 on Build GRCh38
4494479 c.433C>T p.P145S Chr.15: 34090379-34090379 on Build GRCh38
6871151 c.391G>A p.V131I Chr.15: 34090421-34090421 on Build GRCh38
2267835 c.462G>A p.S154S Chr.15: 34090350-34090350 on Build GRCh38
2267837 c.404C>T p.P135L Chr.15: 34090408-34090408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950544086 Chr.15: 34090501-34090501 on Build GRCh38
rs767261000 Chr.15: 34090290-34090290 on Build GRCh38
rs1011716370 Chr.15: 34090687-34090687 on Build GRCh38
rs11557327 Chr.15: 34090378-34090378 on Build GRCh38
rs1038703261 Chr.15: 34090608-34090608 on Build GRCh38
rs140273811 Chr.15: 34090375-34090375 on Build GRCh38
rs770602135 Chr.15: 34090380-34090380 on Build GRCh38
rs376147512 Chr.15: 34090383-34090383 on Build GRCh38
rs372034094 Chr.15: 34090626-34090626 on Build GRCh38
rs960108847 Chr.15: 34090388-34090388 on Build GRCh38
rs922970872 Chr.15: 34090611-34090611 on Build GRCh38
rs1022590901 Chr.15: 34090675-34090675 on Build GRCh38
rs764094450 Chr.15: 34090319-34090319 on Build GRCh38
rs776617754 Chr.15: 34090447-34090447 on Build GRCh38
rs577926379 Chr.15: 34090496-34090496 on Build GRCh38
rs140847906 Chr.15: 34090691-34090691 on Build GRCh38
rs750533108 Chr.15: 34090466-34090466 on Build GRCh38
rs866005480 Chr.15: 34090429-34090429 on Build GRCh38
rs751716052 Chr.15: 34090482-34090482 on Build GRCh38
rs190604325 Chr.15: 34090351-34090351 on Build GRCh38
rs1053053920 Chr.15: 34090547-34090547 on Build GRCh38
rs145380983 Chr.15: 34090491-34090491 on Build GRCh38
rs761708257 Chr.15: 34090449-34090449 on Build GRCh38
rs763784272 Chr.15: 34090437-34090437 on Build GRCh38
rs768843263 Chr.15: 34090356-34090356 on Build GRCh38
rs111443866 Chr.15: 34090663-34090663 on Build GRCh38
rs775211063 Chr.15: 34090261-34090261 on Build GRCh38
rs577896270 Chr.15: 34090255-34090255 on Build GRCh38
rs758341598 Chr.15: 34090470-34090470 on Build GRCh38
rs758668112 Chr.15: 34090350-34090350 on Build GRCh38
rs760344541 Chr.15: 34090304-34090304 on Build GRCh38
rs906105449 Chr.15: 34090704-34090704 on Build GRCh38
rs373701824 Chr.15: 34090420-34090420 on Build GRCh38
rs781778397 Chr.15: 34090368-34090368 on Build GRCh38
rs774940926 Chr.15: 34090419-34090419 on Build GRCh38
rs760533840 Chr.15: 34090428-34090428 on Build GRCh38
rs778959248 Chr.15: 34090345-34090345 on Build GRCh38
rs765110916 Chr.15: 34090452-34090452 on Build GRCh38
rs972824055 Chr.15: 34090438-34090438 on Build GRCh38
rs892725500 Chr.15: 34090211-34090211 on Build GRCh38
rs753715035 Chr.15: 34090335-34090335 on Build GRCh38
rs759237658 Chr.15: 34090277-34090277 on Build GRCh38
rs542747769 Chr.15: 34090628-34090628 on Build GRCh38
rs535294114 Chr.15: 34090414-34090414 on Build GRCh38
rs142173787 Chr.15: 34090203-34090203 on Build GRCh38
rs76849149 Chr.15: 34090665-34090665 on Build GRCh38
rs7165231 Chr.15: 34090530-34090530 on Build GRCh38
rs376375922 Chr.15: 34090526-34090526 on Build GRCh38
rs943296879 Chr.15: 34090582-34090582 on Build GRCh38
rs750692480 Chr.15: 34090346-34090346 on Build GRCh38
rs1022310494 Chr.15: 34090240-34090241 on Build GRCh38
rs770555284 Chr.15: 34090272-34090272 on Build GRCh38
rs762568508 Chr.15: 34090268-34090268 on Build GRCh38
rs368384136 Chr.15: 34090573-34090573 on Build GRCh38
rs569772326 Chr.15: 34090204-34090204 on Build GRCh38
rs939507862 Chr.15: 34090541-34090541 on Build GRCh38
rs752555548 Chr.15: 34090294-34090294 on Build GRCh38
rs150305850 Chr.15: 34090341-34090341 on Build GRCh38
rs781659794 Chr.15: 34090493-34090493 on Build GRCh38
rs3751642 Chr.15: 34090276-34090276 on Build GRCh38
rs1045439201 Chr.15: 34090525-34090525 on Build GRCh38
rs112390189 Chr.15: 34090314-34090314 on Build GRCh38
rs953410742 Chr.15: 34090263-34090263 on Build GRCh38
rs575324849 Chr.15: 34090538-34090538 on Build GRCh38
rs370272260 Chr.15: 34090415-34090415 on Build GRCh38
rs761488152 Chr.15: 34090678-34090678 on Build GRCh38
rs1043645721 Chr.15: 34090697-34090697 on Build GRCh38
rs766635045 Chr.15: 34090471-34090471 on Build GRCh38
rs1053694032 Chr.15: 34090228-34090228 on Build GRCh38
rs146450132 Chr.15: 34090518-34090518 on Build GRCh38
rs747402157 Chr.15: 34090353-34090353 on Build GRCh38

View Full Product Details