Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67176788-67177160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0895L
Entrez Gene ID
653319
Gene Name
KIAA0895 like
Gene Aliases
-
UniGene
Hs.651201
Gene Chromosome Location
Chr.16: 67175602-67183980 on Build GRCh38
Transcripts
NM_001040715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6693426 c.1303A>G p.T435A Chr.16: 67176924-67176924 on Build GRCh38
6305019 c.935G>T p.R312L Chr.16: 67176827-67176827 on Build GRCh38
6693428 c.871C>T p.R291C Chr.16: 67176891-67176891 on Build GRCh38
4756793 c.1315C>T p.H439Y Chr.16: 67176912-67176912 on Build GRCh38
6693427 c.838A>G p.T280A Chr.16: 67176924-67176924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958004104 Chr.16: 67177110-67177112 on Build GRCh38
rs200496423 Chr.16: 67176948-67176948 on Build GRCh38
rs774426608 Chr.16: 67176921-67176921 on Build GRCh38
rs570890803 Chr.16: 67177072-67177072 on Build GRCh38
rs760617969 Chr.16: 67176922-67176922 on Build GRCh38
rs781483998 Chr.16: 67176984-67176984 on Build GRCh38
rs189018091 Chr.16: 67177044-67177044 on Build GRCh38
rs1032576421 Chr.16: 67177088-67177088 on Build GRCh38
rs199656814 Chr.16: 67176944-67176944 on Build GRCh38
rs944145100 Chr.16: 67176832-67176832 on Build GRCh38
rs751181428 Chr.16: 67176981-67176981 on Build GRCh38
rs758031278 Chr.16: 67176828-67176828 on Build GRCh38
rs768863183 Chr.16: 67176920-67176920 on Build GRCh38
rs202158207 Chr.16: 67176891-67176891 on Build GRCh38
rs542248302 Chr.16: 67176890-67176890 on Build GRCh38
rs771851220 Chr.16: 67177132-67177132 on Build GRCh38
rs766249942 Chr.16: 67176935-67176935 on Build GRCh38
rs199684727 Chr.16: 67176851-67176851 on Build GRCh38
rs760095954 Chr.16: 67177124-67177124 on Build GRCh38
rs985319533 Chr.16: 67177127-67177127 on Build GRCh38
rs375177109 Chr.16: 67176991-67176991 on Build GRCh38
rs780558144 Chr.16: 67176919-67176920 on Build GRCh38
rs749446714 Chr.16: 67177011-67177011 on Build GRCh38
rs144066300 Chr.16: 67177000-67177000 on Build GRCh38
rs530734976 Chr.16: 67176916-67176916 on Build GRCh38
rs1024689729 Chr.16: 67177052-67177052 on Build GRCh38
rs918764196 Chr.16: 67176884-67176884 on Build GRCh38
rs780090209 Chr.16: 67176838-67176838 on Build GRCh38
rs752315601 Chr.16: 67176827-67176827 on Build GRCh38
rs759497506 Chr.16: 67176947-67176947 on Build GRCh38
rs201718288 Chr.16: 67176885-67176885 on Build GRCh38
rs757666120 Chr.16: 67176982-67176982 on Build GRCh38
rs999399629 Chr.16: 67177083-67177083 on Build GRCh38
rs753875942 Chr.16: 67176940-67176940 on Build GRCh38
rs753826814 Chr.16: 67176816-67176816 on Build GRCh38
rs61733789 Chr.16: 67176952-67176952 on Build GRCh38
rs781694026 Chr.16: 67176863-67176863 on Build GRCh38
rs866995622 Chr.16: 67176893-67176893 on Build GRCh38
rs763871564 Chr.16: 67176968-67176968 on Build GRCh38
rs747430185 Chr.16: 67176839-67176839 on Build GRCh38
rs146229206 Chr.16: 67176817-67176817 on Build GRCh38
rs746400628 Chr.16: 67176990-67176990 on Build GRCh38

View Full Product Details