Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48855893-48856311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCOCO2
Entrez Gene ID
10241
Gene Name
calcium binding and coiled-coil domain 2
Gene Aliases
NDP52
UniGene
Hs.514920
Gene Chromosome Location
Chr.17: 48830988-48865245 on Build GRCh38
Transcripts
NM_001261390.1 NM_001261391.1 NM_001261393.1 NM_001261395.1 NM_005831.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2697942 c.929T>C p.L310P Chr.17: 48856108-48856108 on Build GRCh38
460306 c.1001A>G p.E334G Chr.17: 48856180-48856180 on Build GRCh38
980673 c.1006G>A p.D336N Chr.17: 48856185-48856185 on Build GRCh38
2697943 c.951C>T p.N317N Chr.17: 48856130-48856130 on Build GRCh38
1582349 c.979C>G p.Q327E Chr.17: 48856158-48856158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753162024 Chr.17: 48856113-48856113 on Build GRCh38
rs749205137 Chr.17: 48856171-48856171 on Build GRCh38
rs199933709 Chr.17: 48856130-48856130 on Build GRCh38
rs746013537 Chr.17: 48856147-48856147 on Build GRCh38
rs571475167 Chr.17: 48855977-48855977 on Build GRCh38
rs774309869 Chr.17: 48856072-48856073 on Build GRCh38
rs769777241 Chr.17: 48856049-48856049 on Build GRCh38
rs748010619 Chr.17: 48856279-48856279 on Build GRCh38
rs762882033 Chr.17: 48856055-48856055 on Build GRCh38
rs976544575 Chr.17: 48856268-48856268 on Build GRCh38
rs1013382891 Chr.17: 48856145-48856145 on Build GRCh38
rs767258030 Chr.17: 48856075-48856075 on Build GRCh38
rs533835841 Chr.17: 48856001-48856001 on Build GRCh38
rs755682584 Chr.17: 48856085-48856085 on Build GRCh38
rs773952651 Chr.17: 48856039-48856039 on Build GRCh38
rs28367230 Chr.17: 48855994-48855994 on Build GRCh38
rs761689483 Chr.17: 48856219-48856219 on Build GRCh38
rs1004222641 Chr.17: 48856020-48856020 on Build GRCh38
rs752450000 Chr.17: 48856188-48856188 on Build GRCh38
rs943907301 Chr.17: 48856260-48856261 on Build GRCh38
rs911121866 Chr.17: 48856257-48856257 on Build GRCh38
rs763808058 Chr.17: 48856066-48856066 on Build GRCh38
rs775582461 Chr.17: 48856170-48856170 on Build GRCh38
rs934144828 Chr.17: 48855930-48855930 on Build GRCh38
rs147129075 Chr.17: 48855974-48855974 on Build GRCh38
rs761599572 Chr.17: 48856072-48856072 on Build GRCh38
rs140191996 Chr.17: 48856035-48856035 on Build GRCh38
rs1023878627 Chr.17: 48855988-48855988 on Build GRCh38
rs370175978 Chr.17: 48856081-48856081 on Build GRCh38
rs555768493 Chr.17: 48856282-48856282 on Build GRCh38
rs768608004 Chr.17: 48856203-48856203 on Build GRCh38
rs781375670 Chr.17: 48856131-48856131 on Build GRCh38
rs765852493 Chr.17: 48856089-48856089 on Build GRCh38
rs960841030 Chr.17: 48856100-48856100 on Build GRCh38
rs769766376 Chr.17: 48856167-48856167 on Build GRCh38
rs11539410 Chr.17: 48856138-48856138 on Build GRCh38
rs759227719 Chr.17: 48856120-48856123 on Build GRCh38
rs569159955 Chr.17: 48855914-48855914 on Build GRCh38
rs372990912 Chr.17: 48856083-48856083 on Build GRCh38
rs1051362447 Chr.17: 48855943-48855943 on Build GRCh38
rs775284599 Chr.17: 48856053-48856053 on Build GRCh38
rs375296166 Chr.17: 48855942-48855942 on Build GRCh38
rs759244453 Chr.17: 48856212-48856212 on Build GRCh38
rs112959796 Chr.17: 48856158-48856158 on Build GRCh38
rs758893046 Chr.17: 48856120-48856120 on Build GRCh38
rs985808435 Chr.17: 48856217-48856217 on Build GRCh38
rs767251591 Chr.17: 48856163-48856164 on Build GRCh38
rs144063772 Chr.17: 48856069-48856069 on Build GRCh38
rs534940711 Chr.17: 48855973-48855973 on Build GRCh38
rs369037244 Chr.17: 48856074-48856074 on Build GRCh38
rs778033786 Chr.17: 48856121-48856121 on Build GRCh38
rs774338066 Chr.17: 48856206-48856206 on Build GRCh38

View Full Product Details