Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 67509925-67510523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSEL
Entrez Gene ID
92126
Gene Name
dermatan sulfate epimerase-like
Gene Aliases
C18orf4
UniGene
Hs.124673
Gene Chromosome Location
Chr.18: 67506582-67516730 on Build GRCh38
Transcripts
NM_032160.2

SNP Detail

SNP ID Chromosome Location
rs138365283 Chr.18: 67509975-67509976 on Build GRCh38
rs920410365 Chr.18: 67510426-67510426 on Build GRCh38
rs1052474753 Chr.18: 67510250-67510250 on Build GRCh38
rs984702031 Chr.18: 67510077-67510077 on Build GRCh38
rs774256629 Chr.18: 67510173-67510173 on Build GRCh38
rs906592763 Chr.18: 67510271-67510271 on Build GRCh38
rs911612578 Chr.18: 67510120-67510120 on Build GRCh38
rs370859868 Chr.18: 67510147-67510147 on Build GRCh38
rs765748663 Chr.18: 67510443-67510443 on Build GRCh38
rs12606012 Chr.18: 67510347-67510347 on Build GRCh38
rs528667088 Chr.18: 67510111-67510111 on Build GRCh38
rs547516471 Chr.18: 67510128-67510128 on Build GRCh38
rs915790205 Chr.18: 67510462-67510462 on Build GRCh38
rs865877290 Chr.18: 67510182-67510182 on Build GRCh38
rs948237183 Chr.18: 67510186-67510186 on Build GRCh38
rs868769659 Chr.18: 67510176-67510176 on Build GRCh38
rs563875263 Chr.18: 67509989-67509989 on Build GRCh38
rs115695859 Chr.18: 67510453-67510453 on Build GRCh38
rs991209308 Chr.18: 67510129-67510129 on Build GRCh38
rs989757816 Chr.18: 67510093-67510094 on Build GRCh38
rs961741842 Chr.18: 67510357-67510357 on Build GRCh38
rs953768629 Chr.18: 67510336-67510336 on Build GRCh38
rs961977667 Chr.18: 67510338-67510338 on Build GRCh38
rs540196049 Chr.18: 67510392-67510392 on Build GRCh38
rs746029080 Chr.18: 67510019-67510019 on Build GRCh38
rs1043952901 Chr.18: 67510211-67510211 on Build GRCh38
rs141175176 Chr.18: 67510458-67510458 on Build GRCh38
rs531217009 Chr.18: 67510078-67510078 on Build GRCh38
rs920442950 Chr.18: 67510196-67510196 on Build GRCh38
rs371708528 Chr.18: 67510281-67510281 on Build GRCh38
rs185192869 Chr.18: 67510100-67510100 on Build GRCh38
rs978215069 Chr.18: 67510054-67510059 on Build GRCh38
rs888927191 Chr.18: 67510284-67510284 on Build GRCh38
rs1056041764 Chr.18: 67510457-67510457 on Build GRCh38
rs999622575 Chr.18: 67510274-67510274 on Build GRCh38
rs376134016 Chr.18: 67510210-67510210 on Build GRCh38
rs113200073 Chr.18: 67510304-67510304 on Build GRCh38
rs535908126 Chr.18: 67510190-67510190 on Build GRCh38
rs548163653 Chr.18: 67510195-67510195 on Build GRCh38
rs1029282608 Chr.18: 67510312-67510312 on Build GRCh38
rs1031079471 Chr.18: 67509972-67509972 on Build GRCh38
rs953338155 Chr.18: 67510074-67510074 on Build GRCh38
rs1046361656 Chr.18: 67510472-67510472 on Build GRCh38
rs376850127 Chr.18: 67510423-67510423 on Build GRCh38
rs149504211 Chr.18: 67510104-67510104 on Build GRCh38
rs971226360 Chr.18: 67510009-67510009 on Build GRCh38
rs770088574 Chr.18: 67510125-67510125 on Build GRCh38
rs1006496605 Chr.18: 67510289-67510289 on Build GRCh38
rs569878619 Chr.18: 67510219-67510219 on Build GRCh38
rs748006052 Chr.18: 67510220-67510220 on Build GRCh38
rs1025090713 Chr.18: 67510008-67510008 on Build GRCh38
rs147339502 Chr.18: 67510480-67510480 on Build GRCh38
rs72954696 Chr.18: 67510256-67510256 on Build GRCh38
rs987621247 Chr.18: 67510119-67510119 on Build GRCh38
rs987117712 Chr.18: 67510339-67510339 on Build GRCh38
rs904128621 Chr.18: 67510215-67510215 on Build GRCh38
rs991229434 Chr.18: 67510433-67510433 on Build GRCh38
rs997566235 Chr.18: 67510272-67510272 on Build GRCh38
rs1035287228 Chr.18: 67510358-67510358 on Build GRCh38
rs1055678408 Chr.18: 67510207-67510207 on Build GRCh38
rs532486492 Chr.18: 67510373-67510373 on Build GRCh38
rs368293295 Chr.18: 67509976-67509977 on Build GRCh38
rs961723943 Chr.18: 67510092-67510092 on Build GRCh38
rs1046832008 Chr.18: 67510260-67510260 on Build GRCh38
rs137988179 Chr.18: 67509983-67509983 on Build GRCh38
rs373888080 Chr.18: 67510389-67510389 on Build GRCh38
rs948230869 Chr.18: 67510464-67510464 on Build GRCh38
rs1028858801 Chr.18: 67509996-67509996 on Build GRCh38
rs1026914657 Chr.18: 67509973-67509973 on Build GRCh38
rs1011162922 Chr.18: 67510311-67510311 on Build GRCh38

View Full Product Details