Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74291574-74292166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5A
Entrez Gene ID
1528
Gene Name
cytochrome b5 type A
Gene Aliases
CYB5, MCB5
UniGene
Hs.465413
Gene Chromosome Location
Chr.18: 74253292-74292016 on Build GRCh38
Transcripts
NM_001190807.2 NM_001914.3 NM_148923.3 XM_011525835.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280245 c.48G>T p.E16D Chr.18: 74291828-74291828 on Build GRCh38
438375 c.45G>A p.E15E Chr.18: 74291831-74291831 on Build GRCh38
1630666 c.117A>T p.K39N Chr.18: 74291759-74291759 on Build GRCh38
4073214 c.22G>A p.A8T Chr.18: 74291854-74291854 on Build GRCh38
4883744 c.100G>T p.V34L Chr.18: 74291776-74291776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552690326 Chr.18: 74292104-74292104 on Build GRCh38
rs775088410 Chr.18: 74291710-74291710 on Build GRCh38
rs547120179 Chr.18: 74291918-74291918 on Build GRCh38
rs536934912 Chr.18: 74291631-74291631 on Build GRCh38
rs77442034 Chr.18: 74292089-74292089 on Build GRCh38
rs1051236 Chr.18: 74291840-74291840 on Build GRCh38
rs541876209 Chr.18: 74291628-74291629 on Build GRCh38
rs768388551 Chr.18: 74291720-74291720 on Build GRCh38
rs745678645 Chr.18: 74291780-74291780 on Build GRCh38
rs770210317 Chr.18: 74291896-74291896 on Build GRCh38
rs372498253 Chr.18: 74291883-74291883 on Build GRCh38
rs563217660 Chr.18: 74292041-74292041 on Build GRCh38
rs756073398 Chr.18: 74291786-74291786 on Build GRCh38
rs752735519 Chr.18: 74291755-74291755 on Build GRCh38
rs970058836 Chr.18: 74291837-74291837 on Build GRCh38
rs201644903 Chr.18: 74291851-74291851 on Build GRCh38
rs901639694 Chr.18: 74291637-74291637 on Build GRCh38
rs529361433 Chr.18: 74291984-74291984 on Build GRCh38
rs769865116 Chr.18: 74291696-74291696 on Build GRCh38
rs536967986 Chr.18: 74291991-74291991 on Build GRCh38
rs766259647 Chr.18: 74291909-74291909 on Build GRCh38
rs758514419 Chr.18: 74291757-74291757 on Build GRCh38
rs533326035 Chr.18: 74291911-74291911 on Build GRCh38
rs200020793 Chr.18: 74291803-74291803 on Build GRCh38
rs780177077 Chr.18: 74291788-74291788 on Build GRCh38
rs750209113 Chr.18: 74291852-74291852 on Build GRCh38
rs768103280 Chr.18: 74291996-74291996 on Build GRCh38
rs981515640 Chr.18: 74291838-74291838 on Build GRCh38
rs1022948598 Chr.18: 74291740-74291740 on Build GRCh38
rs901623806 Chr.18: 74291985-74291985 on Build GRCh38
rs780130050 Chr.18: 74291862-74291862 on Build GRCh38
rs1010427639 Chr.18: 74291778-74291778 on Build GRCh38
rs766837154 Chr.18: 74291738-74291738 on Build GRCh38
rs763016491 Chr.18: 74291834-74291834 on Build GRCh38
rs951972793 Chr.18: 74291685-74291685 on Build GRCh38
rs754878118 Chr.18: 74291869-74291869 on Build GRCh38
rs780981148 Chr.18: 74291771-74291771 on Build GRCh38
rs531270678 Chr.18: 74291843-74291843 on Build GRCh38
rs993777788 Chr.18: 74291659-74291659 on Build GRCh38
rs944501713 Chr.18: 74291972-74291972 on Build GRCh38
rs771717839 Chr.18: 74291884-74291884 on Build GRCh38
rs777443430 Chr.18: 74291885-74291885 on Build GRCh38
rs956228194 Chr.18: 74291854-74291854 on Build GRCh38
rs773874716 Chr.18: 74291800-74291800 on Build GRCh38
rs150530087 Chr.18: 74291736-74291736 on Build GRCh38
rs748835589 Chr.18: 74291711-74291711 on Build GRCh38
rs548402150 Chr.18: 74291986-74291986 on Build GRCh38
rs749578614 Chr.18: 74291716-74291716 on Build GRCh38
rs1007236332 Chr.18: 74291629-74291629 on Build GRCh38
rs568385916 Chr.18: 74291622-74291622 on Build GRCh38
rs776025791 Chr.18: 74291828-74291828 on Build GRCh38
rs369980435 Chr.18: 74291984-74291984 on Build GRCh38
rs1040122670 Chr.18: 74291954-74291954 on Build GRCh38
rs906953652 Chr.18: 74292069-74292069 on Build GRCh38
rs747772941 Chr.18: 74291801-74291801 on Build GRCh38
rs748780741 Chr.18: 74291797-74291797 on Build GRCh38
rs573359213 Chr.18: 74291766-74291766 on Build GRCh38
rs201724841 Chr.18: 74291922-74291922 on Build GRCh38
rs1006112969 Chr.18: 74292058-74292058 on Build GRCh38
rs943034746 Chr.18: 74291953-74291953 on Build GRCh38
rs751875031 Chr.18: 74291764-74291764 on Build GRCh38
rs550423813 Chr.18: 74292007-74292007 on Build GRCh38
rs1003722479 Chr.18: 74291807-74291807 on Build GRCh38
rs150039280 Chr.18: 74291634-74291634 on Build GRCh38
rs878970654 Chr.18: 74291956-74291956 on Build GRCh38
rs770255525 Chr.18: 74291816-74291816 on Build GRCh38
rs746210306 Chr.18: 74291888-74291888 on Build GRCh38
rs1041826039 Chr.18: 74292026-74292026 on Build GRCh38
rs58450389 Chr.18: 74291668-74291668 on Build GRCh38
rs576400124 Chr.18: 74291655-74291655 on Build GRCh38
rs777008023 Chr.18: 74291808-74291808 on Build GRCh38
rs907690903 Chr.18: 74291966-74291966 on Build GRCh38
rs560684215 Chr.18: 74291943-74291943 on Build GRCh38
rs36082929 Chr.18: 74292081-74292081 on Build GRCh38
rs756018585 Chr.18: 74291861-74291861 on Build GRCh38
rs944635331 Chr.18: 74291679-74291679 on Build GRCh38
rs1021122191 Chr.18: 74291794-74291794 on Build GRCh38
rs764208370 Chr.18: 74291760-74291760 on Build GRCh38
rs371426321 Chr.18: 74291989-74291989 on Build GRCh38
rs952111960 Chr.18: 74291910-74291910 on Build GRCh38
rs759981028 Chr.18: 74291750-74291750 on Build GRCh38
rs200736659 Chr.18: 74291879-74291879 on Build GRCh38
rs759926243 Chr.18: 74291814-74291814 on Build GRCh38
rs756479808 Chr.18: 74292115-74292115 on Build GRCh38
rs75160992 Chr.18: 74291863-74291863 on Build GRCh38
rs774107052 Chr.18: 74291729-74291729 on Build GRCh38
rs376151180 Chr.18: 74291741-74291741 on Build GRCh38
rs368287511 Chr.18: 74291753-74291753 on Build GRCh38
rs763457593 Chr.18: 74291914-74291914 on Build GRCh38

View Full Product Details