Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136485473-136485933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf163
Entrez Gene ID
158055
Gene Name
chromosome 9 open reading frame 163
Gene Aliases
-
UniGene
Hs.212613
Gene Chromosome Location
Chr.9: 136483495-136486067 on Build GRCh38
Transcripts
NM_152571.2

SNP Detail

SNP ID Chromosome Location
rs752213878 Chr.9: 136485828-136485828 on Build GRCh38
rs879319956 Chr.9: 136485646-136485646 on Build GRCh38
rs184261618 Chr.9: 136485660-136485660 on Build GRCh38
rs576766112 Chr.9: 136485893-136485893 on Build GRCh38
rs769685479 Chr.9: 136485592-136485592 on Build GRCh38
rs1057249205 Chr.9: 136485651-136485651 on Build GRCh38
rs371154981 Chr.9: 136485754-136485754 on Build GRCh38
rs546562460 Chr.9: 136485860-136485860 on Build GRCh38
rs111938365 Chr.9: 136485827-136485827 on Build GRCh38
rs894536918 Chr.9: 136485654-136485654 on Build GRCh38
rs889094597 Chr.9: 136485522-136485522 on Build GRCh38
rs924608028 Chr.9: 136485623-136485623 on Build GRCh38
rs1008505568 Chr.9: 136485709-136485709 on Build GRCh38
rs938639121 Chr.9: 136485636-136485636 on Build GRCh38
rs899077507 Chr.9: 136485492-136485492 on Build GRCh38
rs1055374544 Chr.9: 136485907-136485907 on Build GRCh38
rs987200666 Chr.9: 136485512-136485512 on Build GRCh38
rs551258638 Chr.9: 136485505-136485505 on Build GRCh38
rs368099659 Chr.9: 136485871-136485871 on Build GRCh38
rs534701641 Chr.9: 136485756-136485756 on Build GRCh38
rs1040408883 Chr.9: 136485615-136485615 on Build GRCh38
rs948602370 Chr.9: 136485657-136485657 on Build GRCh38
rs536861355 Chr.9: 136485634-136485634 on Build GRCh38
rs1020930948 Chr.9: 136485790-136485790 on Build GRCh38
rs977686985 Chr.9: 136485775-136485775 on Build GRCh38
rs993379039 Chr.9: 136485675-136485675 on Build GRCh38
rs759555067 Chr.9: 136485706-136485706 on Build GRCh38
rs569555463 Chr.9: 136485519-136485519 on Build GRCh38
rs577709568 Chr.9: 136485816-136485816 on Build GRCh38
rs1139057 Chr.9: 136485868-136485868 on Build GRCh38
rs139360987 Chr.9: 136485728-136485728 on Build GRCh38
rs993158995 Chr.9: 136485498-136485498 on Build GRCh38
rs78824080 Chr.9: 136485908-136485908 on Build GRCh38
rs897708281 Chr.9: 136485638-136485638 on Build GRCh38
rs929114923 Chr.9: 136485872-136485872 on Build GRCh38
rs898966397 Chr.9: 136485672-136485672 on Build GRCh38
rs964273607 Chr.9: 136485525-136485525 on Build GRCh38
rs967600007 Chr.9: 136485783-136485783 on Build GRCh38
rs1040397846 Chr.9: 136485852-136485852 on Build GRCh38
rs997058838 Chr.9: 136485748-136485748 on Build GRCh38
rs964814224 Chr.9: 136485741-136485741 on Build GRCh38
rs780030741 Chr.9: 136485833-136485833 on Build GRCh38
rs919294571 Chr.9: 136485865-136485865 on Build GRCh38
rs566889809 Chr.9: 136485568-136485568 on Build GRCh38
rs887710650 Chr.9: 136485878-136485878 on Build GRCh38
rs1047615123 Chr.9: 136485702-136485702 on Build GRCh38
rs1002013792 Chr.9: 136485881-136485881 on Build GRCh38
rs944697598 Chr.9: 136485594-136485594 on Build GRCh38
rs926099680 Chr.9: 136485806-136485806 on Build GRCh38
rs887647543 Chr.9: 136485698-136485698 on Build GRCh38
rs192762599 Chr.9: 136485504-136485504 on Build GRCh38
rs890354931 Chr.9: 136485914-136485914 on Build GRCh38
rs914867670 Chr.9: 136485565-136485565 on Build GRCh38
rs960054825 Chr.9: 136485814-136485814 on Build GRCh38
rs968832855 Chr.9: 136485771-136485771 on Build GRCh38
rs765111838 Chr.9: 136485774-136485774 on Build GRCh38
rs960750369 Chr.9: 136485764-136485764 on Build GRCh38
rs924826718 Chr.9: 136485795-136485795 on Build GRCh38
rs992807379 Chr.9: 136485818-136485818 on Build GRCh38
rs977636267 Chr.9: 136485606-136485606 on Build GRCh38
rs553032377 Chr.9: 136485765-136485765 on Build GRCh38
rs944792496 Chr.9: 136485834-136485834 on Build GRCh38
rs977676372 Chr.9: 136485791-136485791 on Build GRCh38
rs759491141 Chr.9: 136485569-136485569 on Build GRCh38
rs1025945568 Chr.9: 136485499-136485499 on Build GRCh38
rs975968289 Chr.9: 136485526-136485526 on Build GRCh38

View Full Product Details