Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184164118-184164675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL3
Entrez Gene ID
1857
Gene Name
dishevelled segment polarity protein 3
Gene Aliases
DRS3
UniGene
Hs.388116
Gene Chromosome Location
Chr.3: 184155311-184173614 on Build GRCh38
Transcripts
NM_004423.3 XM_005247172.2 XM_011512513.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4679767 c.443G>A p.R148Q Chr.3: 184164581-184164581 on Build GRCh38
5008783 c.410C>A p.S137Y Chr.3: 184164548-184164548 on Build GRCh38
3330593 c.423C>A p.A141A Chr.3: 184164561-184164561 on Build GRCh38
5398744 c.268C>T p.P90S Chr.3: 184164303-184164303 on Build GRCh38
3330592 c.411T>C p.S137S Chr.3: 184164549-184164549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758521439 Chr.3: 184164509-184164509 on Build GRCh38
rs934221378 Chr.3: 184164348-184164348 on Build GRCh38
rs971267212 Chr.3: 184164191-184164191 on Build GRCh38
rs1687251 Chr.3: 184164543-184164543 on Build GRCh38
rs370231258 Chr.3: 184164391-184164391 on Build GRCh38
rs779260249 Chr.3: 184164443-184164443 on Build GRCh38
rs563100903 Chr.3: 184164417-184164417 on Build GRCh38
rs551101221 Chr.3: 184164138-184164138 on Build GRCh38
rs370614420 Chr.3: 184164632-184164632 on Build GRCh38
rs771353602 Chr.3: 184164641-184164641 on Build GRCh38
rs781289076 Chr.3: 184164622-184164622 on Build GRCh38
rs185415845 Chr.3: 184164418-184164418 on Build GRCh38
rs776921430 Chr.3: 184164449-184164449 on Build GRCh38
rs984215644 Chr.3: 184164250-184164251 on Build GRCh38
rs747798722 Chr.3: 184164156-184164156 on Build GRCh38
rs764021343 Chr.3: 184164581-184164581 on Build GRCh38
rs756458717 Chr.3: 184164352-184164352 on Build GRCh38
rs199666189 Chr.3: 184164221-184164221 on Build GRCh38
rs746071512 Chr.3: 184164338-184164338 on Build GRCh38
rs777876561 Chr.3: 184164498-184164498 on Build GRCh38
rs779764964 Chr.3: 184164314-184164314 on Build GRCh38
rs34900840 Chr.3: 184164443-184164444 on Build GRCh38
rs545608557 Chr.3: 184164305-184164305 on Build GRCh38
rs773790865 Chr.3: 184164570-184164570 on Build GRCh38
rs147153174 Chr.3: 184164320-184164320 on Build GRCh38
rs183681464 Chr.3: 184164572-184164572 on Build GRCh38
rs1709640 Chr.3: 184164629-184164629 on Build GRCh38
rs529577038 Chr.3: 184164371-184164371 on Build GRCh38
rs1709641 Chr.3: 184164611-184164611 on Build GRCh38
rs770233144 Chr.3: 184164540-184164540 on Build GRCh38
rs775378980 Chr.3: 184164577-184164577 on Build GRCh38
rs766428325 Chr.3: 184164617-184164617 on Build GRCh38
rs762252652 Chr.3: 184164451-184164451 on Build GRCh38
rs139649907 Chr.3: 184164579-184164579 on Build GRCh38
rs774137893 Chr.3: 184164238-184164239 on Build GRCh38
rs1687252 Chr.3: 184164595-184164595 on Build GRCh38
rs143835194 Chr.3: 184164533-184164533 on Build GRCh38
rs200959046 Chr.3: 184164246-184164246 on Build GRCh38
rs751192253 Chr.3: 184164479-184164479 on Build GRCh38
rs752446459 Chr.3: 184164493-184164493 on Build GRCh38
rs180953456 Chr.3: 184164373-184164373 on Build GRCh38
rs759168364 Chr.3: 184164571-184164571 on Build GRCh38
rs772430635 Chr.3: 184164340-184164340 on Build GRCh38
rs142016012 Chr.3: 184164341-184164341 on Build GRCh38
rs745394194 Chr.3: 184164645-184164645 on Build GRCh38
rs771079724 Chr.3: 184164218-184164220 on Build GRCh38
rs747244742 Chr.3: 184164525-184164525 on Build GRCh38
rs972777147 Chr.3: 184164559-184164559 on Build GRCh38
rs753876000 Chr.3: 184164505-184164505 on Build GRCh38
rs1049906925 Chr.3: 184164349-184164349 on Build GRCh38
rs146812695 Chr.3: 184164578-184164578 on Build GRCh38
rs567691554 Chr.3: 184164166-184164166 on Build GRCh38
rs751597815 Chr.3: 184164619-184164619 on Build GRCh38
rs748415570 Chr.3: 184164626-184164626 on Build GRCh38
rs1014702318 Chr.3: 184164462-184164462 on Build GRCh38
rs771100860 Chr.3: 184164332-184164332 on Build GRCh38
rs371456196 Chr.3: 184164565-184164565 on Build GRCh38
rs200551245 Chr.3: 184164436-184164436 on Build GRCh38
rs778291939 Chr.3: 184164258-184164258 on Build GRCh38
rs1006370814 Chr.3: 184164441-184164441 on Build GRCh38
rs754780569 Chr.3: 184164407-184164407 on Build GRCh38
rs143084905 Chr.3: 184164158-184164158 on Build GRCh38
rs750073787 Chr.3: 184164302-184164302 on Build GRCh38
rs775253752 Chr.3: 184164649-184164649 on Build GRCh38
rs772666279 Chr.3: 184164464-184164464 on Build GRCh38
rs776053502 Chr.3: 184164230-184164230 on Build GRCh38
rs760470856 Chr.3: 184164652-184164652 on Build GRCh38
rs753241787 Chr.3: 184164251-184164251 on Build GRCh38
rs373066030 Chr.3: 184164395-184164395 on Build GRCh38
rs541068803 Chr.3: 184164290-184164290 on Build GRCh38
rs767986181 Chr.3: 184164223-184164223 on Build GRCh38
rs768776179 Chr.3: 184164526-184164526 on Build GRCh38
rs777165493 Chr.3: 184164363-184164363 on Build GRCh38
rs377048880 Chr.3: 184164465-184164465 on Build GRCh38
rs749781542 Chr.3: 184164635-184164635 on Build GRCh38
rs761022844 Chr.3: 184164237-184164237 on Build GRCh38
rs113730702 Chr.3: 184164633-184164633 on Build GRCh38
rs567121022 Chr.3: 184164555-184164555 on Build GRCh38
rs374043813 Chr.3: 184164643-184164643 on Build GRCh38
rs764658625 Chr.3: 184164157-184164157 on Build GRCh38
rs756773300 Chr.3: 184164256-184164256 on Build GRCh38
rs566955081 Chr.3: 184164566-184164566 on Build GRCh38
rs556815339 Chr.3: 184164648-184164648 on Build GRCh38
rs150288196 Chr.3: 184164326-184164326 on Build GRCh38
rs981673986 Chr.3: 184164171-184164171 on Build GRCh38
rs1024225999 Chr.3: 184164184-184164184 on Build GRCh38
rs770288075 Chr.3: 184164460-184164460 on Build GRCh38
rs759177048 Chr.3: 184164484-184164484 on Build GRCh38
rs371894225 Chr.3: 184164420-184164420 on Build GRCh38
rs749950896 Chr.3: 184164377-184164377 on Build GRCh38
rs780884904 Chr.3: 184164409-184164409 on Build GRCh38
rs780069118 Chr.3: 184164513-184164513 on Build GRCh38
rs756080778 Chr.3: 184164497-184164497 on Build GRCh38
rs748540521 Chr.3: 184164534-184164534 on Build GRCh38
rs150659339 Chr.3: 184164362-184164362 on Build GRCh38
rs780300833 Chr.3: 184164424-184164424 on Build GRCh38
rs754830767 Chr.3: 184164321-184164321 on Build GRCh38
rs373870236 Chr.3: 184164335-184164335 on Build GRCh38
rs765919453 Chr.3: 184164472-184164472 on Build GRCh38
rs751262267 Chr.3: 184164400-184164400 on Build GRCh38
rs985707313 Chr.3: 184164438-184164438 on Build GRCh38
rs112889179 Chr.3: 184164265-184164265 on Build GRCh38
rs201100826 Chr.3: 184164353-184164353 on Build GRCh38
rs34033424 Chr.3: 184164436-184164436 on Build GRCh38
rs772372510 Chr.3: 184164423-184164423 on Build GRCh38
rs144177739 Chr.3: 184164599-184164599 on Build GRCh38
rs368403444 Chr.3: 184164620-184164620 on Build GRCh38
rs764768182 Chr.3: 184164269-184164269 on Build GRCh38
rs757326449 Chr.3: 184164596-184164596 on Build GRCh38
rs779075428 Chr.3: 184164419-184164419 on Build GRCh38
rs139857613 Chr.3: 184164492-184164492 on Build GRCh38
rs868653763 Chr.3: 184164631-184164631 on Build GRCh38
rs781210884 Chr.3: 184164325-184164325 on Build GRCh38
rs368464179 Chr.3: 184164192-184164192 on Build GRCh38
rs968936338 Chr.3: 184164637-184164637 on Build GRCh38
rs563732436 Chr.3: 184164213-184164213 on Build GRCh38
rs1001390194 Chr.3: 184164214-184164214 on Build GRCh38

View Full Product Details