Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 22735743-22736270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA3
Entrez Gene ID
57733
Gene Name
glucosylceramidase beta 3 (gene/pseudogene)
Gene Aliases
CBG, CBGL1, GLUC, KLRP
UniGene
Hs.653107
Gene Chromosome Location
Chr.4: 22692914-22819575 on Build GRCh38
Transcripts
NM_001128432.2 NM_001277225.1 NM_020973.4 NR_102355.1 NR_102356.1 NR_102357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5471874 c.224G>A p.R75H Chr.4: 22736146-22736146 on Build GRCh38
3602947 c.77G>A p.G26E Chr.4: 22735999-22735999 on Build GRCh38
6054376 c.180G>T p.E60D Chr.4: 22736102-22736102 on Build GRCh38
4446083 c.230C>T p.S77F Chr.4: 22736152-22736152 on Build GRCh38
3602946 c.77G>A p.G26E Chr.4: 22735999-22735999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984195248 Chr.4: 22735804-22735804 on Build GRCh38
rs773332525 Chr.4: 22736114-22736114 on Build GRCh38
rs77900119 Chr.4: 22735826-22735826 on Build GRCh38
rs761352071 Chr.4: 22736015-22736015 on Build GRCh38
rs759397504 Chr.4: 22736233-22736233 on Build GRCh38
rs113740191 Chr.4: 22735854-22735854 on Build GRCh38
rs747002355 Chr.4: 22736192-22736192 on Build GRCh38
rs750350152 Chr.4: 22736022-22736022 on Build GRCh38
rs886694506 Chr.4: 22735832-22735832 on Build GRCh38
rs754733226 Chr.4: 22735974-22735974 on Build GRCh38
rs781668553 Chr.4: 22736126-22736126 on Build GRCh38
rs1033858812 Chr.4: 22735799-22735799 on Build GRCh38
rs747487923 Chr.4: 22736078-22736078 on Build GRCh38
rs868709721 Chr.4: 22736050-22736050 on Build GRCh38
rs201138960 Chr.4: 22736145-22736145 on Build GRCh38
rs961445223 Chr.4: 22736106-22736106 on Build GRCh38
rs762632155 Chr.4: 22735999-22735999 on Build GRCh38
rs544339352 Chr.4: 22736080-22736080 on Build GRCh38
rs7688408 Chr.4: 22735803-22735803 on Build GRCh38
rs746652732 Chr.4: 22735916-22735916 on Build GRCh38
rs754048075 Chr.4: 22736031-22736031 on Build GRCh38
rs974369967 Chr.4: 22735812-22735812 on Build GRCh38
rs557206784 Chr.4: 22735825-22735825 on Build GRCh38
rs987428778 Chr.4: 22736140-22736140 on Build GRCh38
rs764123065 Chr.4: 22736159-22736159 on Build GRCh38
rs750260393 Chr.4: 22735936-22735936 on Build GRCh38
rs72130208 Chr.4: 22735851-22735852 on Build GRCh38
rs369914396 Chr.4: 22735978-22735978 on Build GRCh38
rs867652087 Chr.4: 22735809-22735809 on Build GRCh38
rs916788334 Chr.4: 22736207-22736207 on Build GRCh38
rs747405317 Chr.4: 22735980-22735980 on Build GRCh38
rs1017035222 Chr.4: 22736100-22736100 on Build GRCh38
rs746343429 Chr.4: 22736173-22736174 on Build GRCh38
rs377691778 Chr.4: 22735964-22735964 on Build GRCh38
rs376552395 Chr.4: 22735853-22735855 on Build GRCh38
rs893572971 Chr.4: 22735914-22735914 on Build GRCh38
rs78166175 Chr.4: 22735823-22735823 on Build GRCh38
rs779712309 Chr.4: 22735882-22735882 on Build GRCh38
rs781490954 Chr.4: 22736168-22736168 on Build GRCh38
rs373432928 Chr.4: 22736069-22736069 on Build GRCh38
rs529839966 Chr.4: 22736185-22736185 on Build GRCh38
rs370588320 Chr.4: 22736133-22736133 on Build GRCh38
rs73801182 Chr.4: 22735770-22735770 on Build GRCh38
rs759184981 Chr.4: 22736135-22736135 on Build GRCh38
rs369674650 Chr.4: 22735949-22735949 on Build GRCh38
rs757863507 Chr.4: 22735879-22735879 on Build GRCh38
rs770916647 Chr.4: 22736204-22736204 on Build GRCh38
rs915036728 Chr.4: 22736205-22736205 on Build GRCh38
rs764144043 Chr.4: 22736092-22736092 on Build GRCh38
rs765526849 Chr.4: 22736141-22736141 on Build GRCh38
rs755910063 Chr.4: 22735939-22735939 on Build GRCh38
rs766174909 Chr.4: 22736030-22736030 on Build GRCh38
rs1053561707 Chr.4: 22735900-22735900 on Build GRCh38
rs767480722 Chr.4: 22735934-22735934 on Build GRCh38
rs755225301 Chr.4: 22736060-22736060 on Build GRCh38
rs766155658 Chr.4: 22736132-22736132 on Build GRCh38
rs780459054 Chr.4: 22735951-22735951 on Build GRCh38
rs772845187 Chr.4: 22736191-22736191 on Build GRCh38
rs543250101 Chr.4: 22735764-22735764 on Build GRCh38
rs748949559 Chr.4: 22736089-22736089 on Build GRCh38
rs776391787 Chr.4: 22736227-22736227 on Build GRCh38
rs771748925 Chr.4: 22736190-22736190 on Build GRCh38
rs201187423 Chr.4: 22735862-22735862 on Build GRCh38
rs532386957 Chr.4: 22735911-22735911 on Build GRCh38
rs1029699178 Chr.4: 22736125-22736125 on Build GRCh38
rs1042076536 Chr.4: 22735921-22735921 on Build GRCh38
rs367674331 Chr.4: 22736095-22736095 on Build GRCh38
rs10007404 Chr.4: 22735862-22735862 on Build GRCh38
rs771236423 Chr.4: 22735991-22735991 on Build GRCh38
rs758521024 Chr.4: 22736146-22736146 on Build GRCh38
rs910424265 Chr.4: 22735880-22735880 on Build GRCh38
rs775049284 Chr.4: 22735998-22735998 on Build GRCh38
rs964412814 Chr.4: 22736077-22736077 on Build GRCh38
rs747696603 Chr.4: 22736094-22736094 on Build GRCh38
rs865814554 Chr.4: 22736124-22736124 on Build GRCh38
rs202201230 Chr.4: 22736184-22736184 on Build GRCh38
rs773895851 Chr.4: 22736008-22736008 on Build GRCh38
rs778274609 Chr.4: 22736072-22736072 on Build GRCh38
rs755062877 Chr.4: 22735959-22735959 on Build GRCh38
rs1003840177 Chr.4: 22736037-22736037 on Build GRCh38
rs752122778 Chr.4: 22736165-22736165 on Build GRCh38
rs752776815 Chr.4: 22736070-22736070 on Build GRCh38
rs768114012 Chr.4: 22736007-22736007 on Build GRCh38
rs987788084 Chr.4: 22735877-22735877 on Build GRCh38
rs750949225 Chr.4: 22736190-22736190 on Build GRCh38
rs767570401 Chr.4: 22736017-22736017 on Build GRCh38
rs747784699 Chr.4: 22736189-22736189 on Build GRCh38
rs760573009 Chr.4: 22736029-22736029 on Build GRCh38
rs781612582 Chr.4: 22736087-22736087 on Build GRCh38
rs746250725 Chr.4: 22736175-22736175 on Build GRCh38
rs865950874 Chr.4: 22735859-22735859 on Build GRCh38
rs72094820 Chr.4: 22735854-22735855 on Build GRCh38
rs933261393 Chr.4: 22735827-22735827 on Build GRCh38
rs370990153 Chr.4: 22736235-22736235 on Build GRCh38
rs377374628 Chr.4: 22736107-22736107 on Build GRCh38
rs187359066 Chr.4: 22736166-22736166 on Build GRCh38
rs779021493 Chr.4: 22735960-22735960 on Build GRCh38
rs1037885959 Chr.4: 22735856-22735856 on Build GRCh38
rs1034330569 Chr.4: 22736046-22736046 on Build GRCh38
rs760658989 Chr.4: 22736127-22736127 on Build GRCh38
rs373458485 Chr.4: 22735955-22735955 on Build GRCh38

View Full Product Details