Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127662994-127663532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTU
Entrez Gene ID
27152
Gene Name
inturned planar cell polarity protein
Gene Aliases
INT, PDZD6, PDZK6
UniGene
Hs.391481
Gene Chromosome Location
Chr.4: 127623271-127716779 on Build GRCh38
Transcripts
NM_015693.3 XM_011531844.2 XM_011531845.2 XM_011531846.1 XM_011531847.1 XM_011531849.2 XM_011531850.2 XM_011531851.2 XM_017008026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3714807 c.824G>C p.R275T Chr.4: 127663436-127663436 on Build GRCh38
6435353 c.826C>A p.Q276K Chr.4: 127663438-127663438 on Build GRCh38
3714806 c.824G>C p.R275T Chr.4: 127663436-127663436 on Build GRCh38
4002937 c.813G>A p.T271T Chr.4: 127663425-127663425 on Build GRCh38
123767 c.920A>T p.H307L Chr.4: 127663532-127663532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754416293 Chr.4: 127663344-127663344 on Build GRCh38
rs373900644 Chr.4: 127663438-127663438 on Build GRCh38
rs561534751 Chr.4: 127663404-127663404 on Build GRCh38
rs749238632 Chr.4: 127663410-127663410 on Build GRCh38
rs764225744 Chr.4: 127663342-127663342 on Build GRCh38
rs1056488338 Chr.4: 127663278-127663278 on Build GRCh38
rs747864923 Chr.4: 127663024-127663024 on Build GRCh38
rs777217347 Chr.4: 127663408-127663408 on Build GRCh38
rs750799029 Chr.4: 127663369-127663369 on Build GRCh38
rs866497801 Chr.4: 127663450-127663450 on Build GRCh38
rs763015102 Chr.4: 127663167-127663167 on Build GRCh38
rs373411625 Chr.4: 127663317-127663317 on Build GRCh38
rs949834140 Chr.4: 127663413-127663413 on Build GRCh38
rs189213213 Chr.4: 127663046-127663046 on Build GRCh38
rs377653543 Chr.4: 127663041-127663041 on Build GRCh38
rs199825572 Chr.4: 127663494-127663494 on Build GRCh38
rs772758293 Chr.4: 127663214-127663214 on Build GRCh38
rs192407858 Chr.4: 127663230-127663230 on Build GRCh38
rs35147318 Chr.4: 127663506-127663506 on Build GRCh38
rs376868883 Chr.4: 127663381-127663381 on Build GRCh38
rs973047885 Chr.4: 127663025-127663025 on Build GRCh38
rs72616924 Chr.4: 127663349-127663349 on Build GRCh38
rs951584142 Chr.4: 127663307-127663307 on Build GRCh38
rs901289489 Chr.4: 127663279-127663279 on Build GRCh38
rs1025651370 Chr.4: 127663294-127663294 on Build GRCh38
rs917028788 Chr.4: 127663120-127663120 on Build GRCh38
rs903070121 Chr.4: 127663206-127663206 on Build GRCh38
rs184818825 Chr.4: 127663362-127663362 on Build GRCh38
rs760928366 Chr.4: 127663464-127663464 on Build GRCh38
rs111403080 Chr.4: 127663340-127663340 on Build GRCh38
rs774911378 Chr.4: 127663285-127663285 on Build GRCh38
rs766075926 Chr.4: 127663207-127663207 on Build GRCh38
rs377681633 Chr.4: 127663273-127663273 on Build GRCh38
rs370397959 Chr.4: 127663406-127663406 on Build GRCh38
rs756227798 Chr.4: 127663341-127663341 on Build GRCh38
rs4833380 Chr.4: 127663425-127663425 on Build GRCh38
rs766940420 Chr.4: 127663231-127663231 on Build GRCh38
rs753900288 Chr.4: 127663501-127663501 on Build GRCh38
rs1007418304 Chr.4: 127663241-127663241 on Build GRCh38
rs949743616 Chr.4: 127663147-127663147 on Build GRCh38
rs370476930 Chr.4: 127663333-127663333 on Build GRCh38
rs774070949 Chr.4: 127663420-127663420 on Build GRCh38
rs566538315 Chr.4: 127663178-127663178 on Build GRCh38
rs946937209 Chr.4: 127663252-127663252 on Build GRCh38
rs113854559 Chr.4: 127663443-127663443 on Build GRCh38
rs766461133 Chr.4: 127663432-127663432 on Build GRCh38
rs550383122 Chr.4: 127663111-127663111 on Build GRCh38
rs902786244 Chr.4: 127663260-127663260 on Build GRCh38
rs571968544 Chr.4: 127663291-127663291 on Build GRCh38
rs140355383 Chr.4: 127663412-127663412 on Build GRCh38
rs1043004881 Chr.4: 127663259-127663259 on Build GRCh38
rs1041372958 Chr.4: 127663179-127663179 on Build GRCh38
rs942446596 Chr.4: 127663201-127663201 on Build GRCh38
rs149047407 Chr.4: 127663076-127663076 on Build GRCh38
rs759352629 Chr.4: 127663423-127663423 on Build GRCh38
rs918520922 Chr.4: 127663026-127663026 on Build GRCh38
rs780804144 Chr.4: 127663400-127663400 on Build GRCh38
rs143078322 Chr.4: 127663145-127663145 on Build GRCh38
rs910069180 Chr.4: 127663042-127663042 on Build GRCh38

View Full Product Details