Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 123545594-123546136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1G3
Entrez Gene ID
1456
Gene Name
casein kinase 1 gamma 3
Gene Aliases
CKI-gamma 3, CSNK1G3L
UniGene
Hs.129206
Gene Chromosome Location
Chr.5: 123512099-123617045 on Build GRCh38
Transcripts
NM_001031812.3 NM_001044723.2 NM_001270572.1 NM_001270573.1 NM_001270574.1 NM_004384.4 XM_005271891.1 XM_005271892.2 XM_005271894.2 XM_005271895.2 XM_005271896.1 XM_017009057.1 XM_017009058.1 XM_017009059.1 XM_017009060.1 XM_017009061.1 XM_017009062.1 XM_017009063.1 XM_017009064.1 XM_017009065.1 XM_017009066.1 XM_017009067.1 XM_017009068.1 XM_017009069.1 XM_017009070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1633896 c.43A>G p.M15V Chr.5: 123545706-123545706 on Build GRCh38
6168876 c.154G>T p.G52C Chr.5: 123545817-123545817 on Build GRCh38
3373630 c.55A>G p.S19G Chr.5: 123545718-123545718 on Build GRCh38
1619366 c.157A>G p.N53D Chr.5: 123545820-123545820 on Build GRCh38
7330229 c.50G>A p.R17Q Chr.5: 123545713-123545713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759907767 Chr.5: 123545864-123545864 on Build GRCh38
rs1003917806 Chr.5: 123545646-123545646 on Build GRCh38
rs745440820 Chr.5: 123545724-123545724 on Build GRCh38
rs776967678 Chr.5: 123545850-123545850 on Build GRCh38
rs762552881 Chr.5: 123545895-123545895 on Build GRCh38
rs1045628051 Chr.5: 123545967-123545967 on Build GRCh38
rs377249938 Chr.5: 123545698-123545698 on Build GRCh38
rs747767162 Chr.5: 123545832-123545832 on Build GRCh38
rs752765883 Chr.5: 123545741-123545741 on Build GRCh38
rs773905338 Chr.5: 123545716-123545716 on Build GRCh38
rs540419871 Chr.5: 123546078-123546078 on Build GRCh38
rs554875810 Chr.5: 123545946-123545946 on Build GRCh38
rs1034577040 Chr.5: 123546033-123546033 on Build GRCh38
rs536917070 Chr.5: 123545849-123545849 on Build GRCh38
rs1054317511 Chr.5: 123545874-123545874 on Build GRCh38
rs920743729 Chr.5: 123545700-123545700 on Build GRCh38
rs370475468 Chr.5: 123545706-123545706 on Build GRCh38
rs891646744 Chr.5: 123545914-123545914 on Build GRCh38
rs34742615 Chr.5: 123545665-123545665 on Build GRCh38
rs1034218332 Chr.5: 123545662-123545662 on Build GRCh38
rs544688341 Chr.5: 123546083-123546087 on Build GRCh38
rs139353656 Chr.5: 123545740-123545740 on Build GRCh38
rs1009993133 Chr.5: 123545954-123545954 on Build GRCh38
rs760032416 Chr.5: 123545737-123545737 on Build GRCh38
rs959509062 Chr.5: 123546056-123546056 on Build GRCh38
rs758415272 Chr.5: 123545742-123545742 on Build GRCh38
rs572469054 Chr.5: 123545768-123545768 on Build GRCh38
rs759468532 Chr.5: 123545734-123545734 on Build GRCh38
rs748789328 Chr.5: 123545813-123545813 on Build GRCh38
rs374469630 Chr.5: 123545752-123545752 on Build GRCh38
rs973977516 Chr.5: 123545860-123545860 on Build GRCh38
rs781230041 Chr.5: 123545685-123545685 on Build GRCh38
rs749511098 Chr.5: 123545991-123545991 on Build GRCh38
rs573342310 Chr.5: 123546062-123546062 on Build GRCh38
rs935733993 Chr.5: 123545835-123545835 on Build GRCh38
rs1003581576 Chr.5: 123546004-123546004 on Build GRCh38
rs1020805974 Chr.5: 123545631-123545631 on Build GRCh38
rs950868481 Chr.5: 123546070-123546070 on Build GRCh38
rs765667569 Chr.5: 123545883-123545883 on Build GRCh38
rs1016640157 Chr.5: 123546114-123546114 on Build GRCh38
rs755527872 Chr.5: 123545652-123545652 on Build GRCh38
rs751755170 Chr.5: 123545759-123545759 on Build GRCh38
rs377657356 Chr.5: 123545653-123545653 on Build GRCh38
rs946040536 Chr.5: 123545962-123545962 on Build GRCh38
rs746753488 Chr.5: 123545670-123545670 on Build GRCh38
rs146622169 Chr.5: 123545762-123545762 on Build GRCh38
rs922119340 Chr.5: 123545870-123545870 on Build GRCh38
rs754241218 Chr.5: 123545732-123545732 on Build GRCh38
rs982848981 Chr.5: 123546093-123546093 on Build GRCh38
rs775129818 Chr.5: 123545801-123545801 on Build GRCh38
rs745781220 Chr.5: 123545692-123545692 on Build GRCh38
rs569593775 Chr.5: 123545828-123545828 on Build GRCh38
rs761050634 Chr.5: 123545718-123545718 on Build GRCh38
rs769380038 Chr.5: 123545757-123545759 on Build GRCh38
rs766707433 Chr.5: 123545728-123545728 on Build GRCh38
rs756054991 Chr.5: 123545792-123545792 on Build GRCh38
rs916887204 Chr.5: 123545705-123545705 on Build GRCh38
rs1034084789 Chr.5: 123546014-123546014 on Build GRCh38
rs181108291 Chr.5: 123545661-123545661 on Build GRCh38
rs995350529 Chr.5: 123546057-123546057 on Build GRCh38
rs752786063 Chr.5: 123545654-123545654 on Build GRCh38
rs781535738 Chr.5: 123545924-123545927 on Build GRCh38
rs766830197 Chr.5: 123545641-123545641 on Build GRCh38
rs200913320 Chr.5: 123545761-123545761 on Build GRCh38
rs779856749 Chr.5: 123545804-123545804 on Build GRCh38
rs144555645 Chr.5: 123545753-123545753 on Build GRCh38
rs1000323256 Chr.5: 123546027-123546027 on Build GRCh38
rs769050454 Chr.5: 123546088-123546088 on Build GRCh38
rs374355054 Chr.5: 123545644-123545644 on Build GRCh38
rs779964383 Chr.5: 123545952-123545952 on Build GRCh38
rs770882598 Chr.5: 123545673-123545673 on Build GRCh38
rs1041768514 Chr.5: 123545971-123545971 on Build GRCh38
rs745618803 Chr.5: 123545709-123545709 on Build GRCh38
rs745711542 Chr.5: 123545777-123545777 on Build GRCh38
rs1027710887 Chr.5: 123546069-123546069 on Build GRCh38
rs775893997 Chr.5: 123545887-123545887 on Build GRCh38
rs376102229 Chr.5: 123545819-123545819 on Build GRCh38
rs768285137 Chr.5: 123545710-123545710 on Build GRCh38

View Full Product Details