Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31728732-31729306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDAH2
Entrez Gene ID
23564
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
DDAH, DDAHII, G6a, HEL-S-277, NG30
UniGene
Hs.247362
Gene Chromosome Location
Chr.6: 31727037-31730265 on Build GRCh38
Transcripts
NM_001303007.1 NM_001303008.1 NM_013974.2 XM_011514448.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624718 c.275G>A p.W92* Chr.6: 31728887-31728887 on Build GRCh38
5959800 c.203C>T p.S68L Chr.6: 31728959-31728959 on Build GRCh38
3624717 c.309C>T p.V103V Chr.6: 31728734-31728734 on Build GRCh38
6359645 c.5G>T p.G2V Chr.6: 31729157-31729157 on Build GRCh38
3624719 c.101C>T p.P34L Chr.6: 31729061-31729061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778449032 Chr.6: 31729190-31729190 on Build GRCh38
rs751453578 Chr.6: 31729020-31729020 on Build GRCh38
rs1055732783 Chr.6: 31728768-31728768 on Build GRCh38
rs371435542 Chr.6: 31728957-31728957 on Build GRCh38
rs760617963 Chr.6: 31729166-31729166 on Build GRCh38
rs766636105 Chr.6: 31728968-31728968 on Build GRCh38
rs747390096 Chr.6: 31729090-31729090 on Build GRCh38
rs565274260 Chr.6: 31729071-31729071 on Build GRCh38
rs376853159 Chr.6: 31728927-31728927 on Build GRCh38
rs909262756 Chr.6: 31729287-31729287 on Build GRCh38
rs750222340 Chr.6: 31729159-31729159 on Build GRCh38
rs751346909 Chr.6: 31728870-31728870 on Build GRCh38
rs754471815 Chr.6: 31729076-31729076 on Build GRCh38
rs773419342 Chr.6: 31728819-31728819 on Build GRCh38
rs376727018 Chr.6: 31729189-31729189 on Build GRCh38
rs765301432 Chr.6: 31729018-31729018 on Build GRCh38
rs761780700 Chr.6: 31729149-31729149 on Build GRCh38
rs775065727 Chr.6: 31728945-31728945 on Build GRCh38
rs750228711 Chr.6: 31729060-31729060 on Build GRCh38
rs752195343 Chr.6: 31729191-31729191 on Build GRCh38
rs748642109 Chr.6: 31729083-31729083 on Build GRCh38
rs766404608 Chr.6: 31728849-31728849 on Build GRCh38
rs755755817 Chr.6: 31728901-31728901 on Build GRCh38
rs745473395 Chr.6: 31728893-31728893 on Build GRCh38
rs762297044 Chr.6: 31728770-31728770 on Build GRCh38
rs772425050 Chr.6: 31728955-31728955 on Build GRCh38
rs561348708 Chr.6: 31728836-31728836 on Build GRCh38
rs73728976 Chr.6: 31729151-31729151 on Build GRCh38
rs749567402 Chr.6: 31729198-31729204 on Build GRCh38
rs753483978 Chr.6: 31729180-31729180 on Build GRCh38
rs753958371 Chr.6: 31728851-31728851 on Build GRCh38
rs928260663 Chr.6: 31729178-31729178 on Build GRCh38
rs975377875 Chr.6: 31729079-31729079 on Build GRCh38
rs767628982 Chr.6: 31728782-31728782 on Build GRCh38
rs767307185 Chr.6: 31729027-31729027 on Build GRCh38
rs778580785 Chr.6: 31729081-31729081 on Build GRCh38
rs1017529394 Chr.6: 31729109-31729109 on Build GRCh38
rs757006320 Chr.6: 31728876-31728876 on Build GRCh38
rs532560994 Chr.6: 31729087-31729087 on Build GRCh38
rs576302601 Chr.6: 31728868-31728868 on Build GRCh38
rs138128995 Chr.6: 31729074-31729074 on Build GRCh38
rs769330680 Chr.6: 31728757-31728757 on Build GRCh38
rs1009304954 Chr.6: 31728993-31728993 on Build GRCh38
rs375373527 Chr.6: 31729246-31729246 on Build GRCh38
rs907616563 Chr.6: 31729124-31729124 on Build GRCh38
rs776679194 Chr.6: 31728977-31728977 on Build GRCh38
rs766281056 Chr.6: 31729171-31729171 on Build GRCh38
rs774837244 Chr.6: 31728764-31728764 on Build GRCh38
rs368529782 Chr.6: 31728882-31728882 on Build GRCh38
rs371472036 Chr.6: 31728943-31728943 on Build GRCh38
rs1044656908 Chr.6: 31728856-31728856 on Build GRCh38
rs940386874 Chr.6: 31729127-31729127 on Build GRCh38
rs760918705 Chr.6: 31728966-31728966 on Build GRCh38
rs757162385 Chr.6: 31729026-31729026 on Build GRCh38
rs1006155349 Chr.6: 31728842-31728842 on Build GRCh38
rs759764327 Chr.6: 31729120-31729120 on Build GRCh38
rs983519679 Chr.6: 31729240-31729241 on Build GRCh38
rs1056102883 Chr.6: 31729203-31729203 on Build GRCh38
rs542613838 Chr.6: 31728784-31728784 on Build GRCh38
rs753394138 Chr.6: 31729073-31729073 on Build GRCh38
rs772494896 Chr.6: 31729086-31729086 on Build GRCh38
rs775460540 Chr.6: 31729141-31729141 on Build GRCh38
rs776763038 Chr.6: 31729112-31729112 on Build GRCh38
rs368696409 Chr.6: 31728947-31728947 on Build GRCh38
rs765882021 Chr.6: 31729255-31729256 on Build GRCh38
rs1047750710 Chr.6: 31728781-31728781 on Build GRCh38
rs754733474 Chr.6: 31729111-31729111 on Build GRCh38
rs375654675 Chr.6: 31728860-31728860 on Build GRCh38
rs548017157 Chr.6: 31729114-31729114 on Build GRCh38
rs371970217 Chr.6: 31728903-31728903 on Build GRCh38
rs149644767 Chr.6: 31729122-31729122 on Build GRCh38
rs759674784 Chr.6: 31728995-31728995 on Build GRCh38
rs758309508 Chr.6: 31728852-31728852 on Build GRCh38
rs771138152 Chr.6: 31729108-31729108 on Build GRCh38
rs750940851 Chr.6: 31729266-31729266 on Build GRCh38
rs373488867 Chr.6: 31729274-31729274 on Build GRCh38

View Full Product Details