Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 88794136-88794646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf62
Entrez Gene ID
219557
Gene Name
chromosome 7 open reading frame 62
Gene Aliases
-
UniGene
Hs.112877
Gene Chromosome Location
Chr.7: 88794106-88797937 on Build GRCh38
Transcripts
NM_152706.3 XM_011515904.2 NM_181646.3

Gene Symbol
ZNF804B
Entrez Gene ID
219578
Gene Name
zinc finger protein 804B
Gene Aliases
-
UniGene
Hs.684410
Gene Chromosome Location
Chr.7: 88759368-89337057 on Build GRCh38
Transcripts
NM_152706.3 XM_011515904.2 NM_181646.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
747680 c.611C>T p.A204V Chr.7: 88794332-88794332 on Build GRCh38
5615890 c.475G>T p.D159Y Chr.7: 88794468-88794468 on Build GRCh38
271856 c.337G>T p.D113Y Chr.7: 88794606-88794606 on Build GRCh38
3883141 c.475G>A p.D159N Chr.7: 88794468-88794468 on Build GRCh38
270657 c.429_430insT p.M144fs*37 Chr.7: 88794513-88794514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55769925 Chr.7: 88794175-88794175 on Build GRCh38
rs776842699 Chr.7: 88794464-88794464 on Build GRCh38
rs867402496 Chr.7: 88794556-88794556 on Build GRCh38
rs1012225539 Chr.7: 88794227-88794227 on Build GRCh38
rs769803138 Chr.7: 88794577-88794577 on Build GRCh38
rs148216190 Chr.7: 88794339-88794339 on Build GRCh38
rs372317539 Chr.7: 88794468-88794468 on Build GRCh38
rs747661550 Chr.7: 88794196-88794196 on Build GRCh38
rs1030508962 Chr.7: 88794334-88794334 on Build GRCh38
rs780646965 Chr.7: 88794562-88794562 on Build GRCh38
rs139432346 Chr.7: 88794435-88794435 on Build GRCh38
rs745333051 Chr.7: 88794519-88794519 on Build GRCh38
rs758993492 Chr.7: 88794549-88794549 on Build GRCh38
rs377395892 Chr.7: 88794583-88794583 on Build GRCh38
rs759884778 Chr.7: 88794352-88794352 on Build GRCh38
rs752504765 Chr.7: 88794474-88794474 on Build GRCh38
rs755787489 Chr.7: 88794423-88794423 on Build GRCh38
rs754731598 Chr.7: 88794328-88794328 on Build GRCh38
rs202025178 Chr.7: 88794390-88794390 on Build GRCh38
rs767449422 Chr.7: 88794591-88794591 on Build GRCh38
rs200976271 Chr.7: 88794578-88794578 on Build GRCh38
rs966166733 Chr.7: 88794619-88794619 on Build GRCh38
rs755840841 Chr.7: 88794476-88794476 on Build GRCh38
rs747785460 Chr.7: 88794335-88794335 on Build GRCh38
rs780889651 Chr.7: 88794331-88794331 on Build GRCh38
rs771431580 Chr.7: 88794216-88794216 on Build GRCh38
rs778458931 Chr.7: 88794413-88794416 on Build GRCh38
rs774603331 Chr.7: 88794218-88794218 on Build GRCh38
rs769288219 Chr.7: 88794201-88794201 on Build GRCh38
rs777349674 Chr.7: 88794425-88794425 on Build GRCh38
rs2373398 Chr.7: 88794281-88794281 on Build GRCh38
rs923810672 Chr.7: 88794432-88794432 on Build GRCh38
rs377269794 Chr.7: 88794469-88794469 on Build GRCh38
rs776031300 Chr.7: 88794450-88794450 on Build GRCh38
rs910936574 Chr.7: 88794161-88794161 on Build GRCh38
rs758264497 Chr.7: 88794172-88794172 on Build GRCh38
rs965834952 Chr.7: 88794263-88794263 on Build GRCh38
rs143068564 Chr.7: 88794448-88794448 on Build GRCh38
rs561576701 Chr.7: 88794308-88794308 on Build GRCh38
rs768211942 Chr.7: 88794189-88794189 on Build GRCh38
rs773765403 Chr.7: 88794207-88794207 on Build GRCh38
rs201260308 Chr.7: 88794626-88794626 on Build GRCh38
rs760951078 Chr.7: 88794357-88794357 on Build GRCh38
rs10232200 Chr.7: 88794163-88794163 on Build GRCh38
rs1006291790 Chr.7: 88794271-88794271 on Build GRCh38
rs533500155 Chr.7: 88794565-88794565 on Build GRCh38
rs997612987 Chr.7: 88794611-88794611 on Build GRCh38
rs747747087 Chr.7: 88794519-88794519 on Build GRCh38
rs143550892 Chr.7: 88794602-88794602 on Build GRCh38
rs141537750 Chr.7: 88794354-88794354 on Build GRCh38
rs375981756 Chr.7: 88794419-88794419 on Build GRCh38
rs199906096 Chr.7: 88794162-88794162 on Build GRCh38
rs781359459 Chr.7: 88794574-88794574 on Build GRCh38
rs753986544 Chr.7: 88794368-88794368 on Build GRCh38
rs2373397 Chr.7: 88794282-88794282 on Build GRCh38
rs979197936 Chr.7: 88794430-88794430 on Build GRCh38
rs774856236 Chr.7: 88794351-88794351 on Build GRCh38
rs984405852 Chr.7: 88794353-88794353 on Build GRCh38
rs780941076 Chr.7: 88794414-88794414 on Build GRCh38
rs997723662 Chr.7: 88794309-88794309 on Build GRCh38
rs752967398 Chr.7: 88794237-88794237 on Build GRCh38
rs368156278 Chr.7: 88794268-88794268 on Build GRCh38
rs756770295 Chr.7: 88794505-88794505 on Build GRCh38
rs143392475 Chr.7: 88794229-88794229 on Build GRCh38
rs779350706 Chr.7: 88794445-88794445 on Build GRCh38
rs750972928 Chr.7: 88794433-88794433 on Build GRCh38
rs758809917 Chr.7: 88794592-88794600 on Build GRCh38
rs753478549 Chr.7: 88794609-88794609 on Build GRCh38
rs34781240 Chr.7: 88794465-88794465 on Build GRCh38
rs201329404 Chr.7: 88794459-88794459 on Build GRCh38
rs748785233 Chr.7: 88794340-88794340 on Build GRCh38
rs147171457 Chr.7: 88794283-88794283 on Build GRCh38
rs529731262 Chr.7: 88794398-88794398 on Build GRCh38
rs763426089 Chr.7: 88794213-88794213 on Build GRCh38
rs1054263128 Chr.7: 88794528-88794528 on Build GRCh38
rs747327078 Chr.7: 88794455-88794455 on Build GRCh38
rs764250079 Chr.7: 88794265-88794265 on Build GRCh38
rs756967541 Chr.7: 88794612-88794612 on Build GRCh38
rs775402208 Chr.7: 88794593-88794593 on Build GRCh38
rs763858589 Chr.7: 88794608-88794608 on Build GRCh38
rs751321326 Chr.7: 88794300-88794300 on Build GRCh38
rs764368815 Chr.7: 88794360-88794360 on Build GRCh38
rs111812085 Chr.7: 88794190-88794190 on Build GRCh38
rs34276371 Chr.7: 88794386-88794386 on Build GRCh38
rs745791103 Chr.7: 88794165-88794165 on Build GRCh38
rs764751270 Chr.7: 88794625-88794625 on Build GRCh38
rs758766386 Chr.7: 88794446-88794446 on Build GRCh38
rs2293583 Chr.7: 88794567-88794567 on Build GRCh38
rs1043978912 Chr.7: 88794170-88794170 on Build GRCh38
rs185826530 Chr.7: 88794188-88794188 on Build GRCh38
rs369625195 Chr.7: 88794466-88794466 on Build GRCh38
rs767638947 Chr.7: 88794232-88794232 on Build GRCh38
rs369839861 Chr.7: 88794501-88794501 on Build GRCh38
rs769341253 Chr.7: 88794337-88794337 on Build GRCh38
rs867089376 Chr.7: 88794426-88794426 on Build GRCh38
rs771357449 Chr.7: 88794342-88794342 on Build GRCh38
rs371691831 Chr.7: 88794391-88794391 on Build GRCh38
rs760899849 Chr.7: 88794264-88794264 on Build GRCh38

View Full Product Details