Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66518070-66518656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf46
Entrez Gene ID
254778
Gene Name
chromosome 8 open reading frame 46
Gene Aliases
-
UniGene
Hs.268869
Gene Chromosome Location
Chr.8: 66493256-66518524 on Build GRCh38
Transcripts
NM_152765.3

SNP Detail

SNP ID Chromosome Location
rs138439727 Chr.8: 66518542-66518542 on Build GRCh38
rs901085087 Chr.8: 66518390-66518390 on Build GRCh38
rs1003742011 Chr.8: 66518571-66518571 on Build GRCh38
rs966650928 Chr.8: 66518338-66518338 on Build GRCh38
rs903026751 Chr.8: 66518140-66518140 on Build GRCh38
rs888088980 Chr.8: 66518224-66518224 on Build GRCh38
rs548830895 Chr.8: 66518592-66518592 on Build GRCh38
rs866689717 Chr.8: 66518487-66518487 on Build GRCh38
rs911961934 Chr.8: 66518368-66518368 on Build GRCh38
rs959498232 Chr.8: 66518536-66518536 on Build GRCh38
rs545556922 Chr.8: 66518535-66518535 on Build GRCh38
rs1041006684 Chr.8: 66518154-66518154 on Build GRCh38
rs560033356 Chr.8: 66518582-66518582 on Build GRCh38
rs1022466141 Chr.8: 66518593-66518593 on Build GRCh38
rs970921273 Chr.8: 66518265-66518265 on Build GRCh38
rs1005237526 Chr.8: 66518490-66518490 on Build GRCh38
rs534197095 Chr.8: 66518312-66518312 on Build GRCh38
rs949702843 Chr.8: 66518201-66518201 on Build GRCh38
rs932573178 Chr.8: 66518516-66518516 on Build GRCh38
rs923913086 Chr.8: 66518570-66518570 on Build GRCh38
rs527932815 Chr.8: 66518562-66518562 on Build GRCh38
rs149442590 Chr.8: 66518127-66518127 on Build GRCh38
rs751718058 Chr.8: 66518525-66518525 on Build GRCh38
rs117779661 Chr.8: 66518569-66518569 on Build GRCh38
rs62512590 Chr.8: 66518589-66518589 on Build GRCh38
rs375724067 Chr.8: 66518452-66518452 on Build GRCh38
rs756419452 Chr.8: 66518621-66518621 on Build GRCh38
rs915351655 Chr.8: 66518100-66518100 on Build GRCh38
rs560607843 Chr.8: 66518545-66518545 on Build GRCh38
rs779966925 Chr.8: 66518486-66518486 on Build GRCh38
rs556954298 Chr.8: 66518221-66518221 on Build GRCh38
rs966775739 Chr.8: 66518603-66518603 on Build GRCh38
rs978204193 Chr.8: 66518355-66518355 on Build GRCh38
rs542795731 Chr.8: 66518357-66518357 on Build GRCh38
rs921205075 Chr.8: 66518507-66518507 on Build GRCh38
rs879117648 Chr.8: 66518380-66518380 on Build GRCh38
rs1042839578 Chr.8: 66518129-66518129 on Build GRCh38
rs576589781 Chr.8: 66518532-66518532 on Build GRCh38
rs550249875 Chr.8: 66518218-66518218 on Build GRCh38
rs146389699 Chr.8: 66518172-66518172 on Build GRCh38
rs1048313492 Chr.8: 66518531-66518531 on Build GRCh38
rs899744032 Chr.8: 66518163-66518163 on Build GRCh38
rs1033829997 Chr.8: 66518579-66518579 on Build GRCh38
rs116593118 Chr.8: 66518271-66518271 on Build GRCh38
rs994513145 Chr.8: 66518432-66518432 on Build GRCh38
rs556605332 Chr.8: 66518521-66518521 on Build GRCh38
rs959599096 Chr.8: 66518296-66518296 on Build GRCh38
rs1003979220 Chr.8: 66518232-66518232 on Build GRCh38
rs758696588 Chr.8: 66518436-66518436 on Build GRCh38
rs1033968293 Chr.8: 66518289-66518289 on Build GRCh38
rs559753953 Chr.8: 66518585-66518585 on Build GRCh38
rs943830488 Chr.8: 66518146-66518146 on Build GRCh38
rs377216806 Chr.8: 66518613-66518613 on Build GRCh38
rs989669543 Chr.8: 66518321-66518321 on Build GRCh38
rs966508527 Chr.8: 66518626-66518626 on Build GRCh38
rs574083238 Chr.8: 66518346-66518346 on Build GRCh38
rs549050847 Chr.8: 66518115-66518115 on Build GRCh38
rs918183818 Chr.8: 66518187-66518187 on Build GRCh38
rs996543840 Chr.8: 66518164-66518164 on Build GRCh38
rs750626250 Chr.8: 66518360-66518360 on Build GRCh38
rs945484033 Chr.8: 66518114-66518114 on Build GRCh38
rs1015725035 Chr.8: 66518245-66518245 on Build GRCh38
rs543386997 Chr.8: 66518145-66518145 on Build GRCh38
rs538021479 Chr.8: 66518170-66518170 on Build GRCh38
rs1042390152 Chr.8: 66518226-66518226 on Build GRCh38
rs554164629 Chr.8: 66518326-66518326 on Build GRCh38

View Full Product Details