Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196910018-196910543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR4
Entrez Gene ID
10877
Gene Name
complement factor H related 4
Gene Aliases
CFHL4, FHR-4, FHR4
UniGene
Hs.710100
Gene Chromosome Location
Chr.1: 196888014-196918911 on Build GRCh38
Transcripts
NM_001201550.2 NM_001201551.1 NM_006684.4 XM_006711129.3 XM_017000110.1 XM_017000111.1 XM_017000112.1 XM_017000113.1 XM_017000114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3480882 c.932G>A p.G311E Chr.1: 196910416-196910416 on Build GRCh38
6024853 c.969G>C p.G323G Chr.1: 196910453-196910453 on Build GRCh38
3976771 c.867C>A p.Y289* Chr.1: 196910351-196910351 on Build GRCh38
4903450 c.817C>T p.P273S Chr.1: 196910301-196910301 on Build GRCh38
3687836 c.857G>A p.R286H Chr.1: 196910341-196910341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028406979 Chr.1: 196910102-196910102 on Build GRCh38
rs905279177 Chr.1: 196910250-196910250 on Build GRCh38
rs756020612 Chr.1: 196910319-196910319 on Build GRCh38
rs375197314 Chr.1: 196910107-196910107 on Build GRCh38
rs192533579 Chr.1: 196910462-196910462 on Build GRCh38
rs575564104 Chr.1: 196910175-196910175 on Build GRCh38
rs368112020 Chr.1: 196910151-196910151 on Build GRCh38
rs74701062 Chr.1: 196910181-196910181 on Build GRCh38
rs959359717 Chr.1: 196910503-196910503 on Build GRCh38
rs753643477 Chr.1: 196910333-196910333 on Build GRCh38
rs764444563 Chr.1: 196910316-196910316 on Build GRCh38
rs552461407 Chr.1: 196910399-196910399 on Build GRCh38
rs751607261 Chr.1: 196910445-196910445 on Build GRCh38
rs775856781 Chr.1: 196910404-196910404 on Build GRCh38
rs1022530373 Chr.1: 196910297-196910297 on Build GRCh38
rs777524466 Chr.1: 196910371-196910371 on Build GRCh38
rs1005565320 Chr.1: 196910216-196910216 on Build GRCh38
rs772730258 Chr.1: 196910277-196910277 on Build GRCh38
rs540816872 Chr.1: 196910263-196910263 on Build GRCh38
rs770523337 Chr.1: 196910383-196910383 on Build GRCh38
rs780767022 Chr.1: 196910496-196910496 on Build GRCh38
rs780179186 Chr.1: 196910247-196910247 on Build GRCh38
rs184562589 Chr.1: 196910515-196910515 on Build GRCh38
rs771482209 Chr.1: 196910350-196910350 on Build GRCh38
rs752827321 Chr.1: 196910308-196910308 on Build GRCh38
rs187464658 Chr.1: 196910042-196910042 on Build GRCh38
rs747541418 Chr.1: 196910268-196910268 on Build GRCh38
rs1020056178 Chr.1: 196910171-196910171 on Build GRCh38
rs570663018 Chr.1: 196910106-196910106 on Build GRCh38
rs778653315 Chr.1: 196910339-196910339 on Build GRCh38
rs1042577306 Chr.1: 196910424-196910424 on Build GRCh38
rs552795605 Chr.1: 196910354-196910354 on Build GRCh38
rs770576374 Chr.1: 196910495-196910495 on Build GRCh38
rs745884323 Chr.1: 196910411-196910411 on Build GRCh38
rs753695580 Chr.1: 196910461-196910461 on Build GRCh38
rs41265219 Chr.1: 196910303-196910303 on Build GRCh38
rs200497324 Chr.1: 196910464-196910464 on Build GRCh38
rs774519151 Chr.1: 196910403-196910414 on Build GRCh38
rs867140793 Chr.1: 196910302-196910302 on Build GRCh38
rs372959206 Chr.1: 196910087-196910087 on Build GRCh38
rs1045097650 Chr.1: 196910434-196910434 on Build GRCh38
rs564570704 Chr.1: 196910182-196910182 on Build GRCh38
rs770991524 Chr.1: 196910337-196910344 on Build GRCh38
rs866833472 Chr.1: 196910166-196910166 on Build GRCh38
rs188298928 Chr.1: 196910341-196910341 on Build GRCh38
rs552747636 Chr.1: 196910045-196910045 on Build GRCh38
rs371367197 Chr.1: 196910172-196910172 on Build GRCh38
rs569248104 Chr.1: 196910338-196910338 on Build GRCh38
rs139989231 Chr.1: 196910189-196910189 on Build GRCh38
rs777174191 Chr.1: 196910293-196910293 on Build GRCh38
rs746484307 Chr.1: 196910477-196910477 on Build GRCh38
rs199583121 Chr.1: 196910307-196910307 on Build GRCh38
rs772680118 Chr.1: 196910471-196910472 on Build GRCh38
rs773940527 Chr.1: 196910266-196910266 on Build GRCh38
rs910856197 Chr.1: 196910324-196910324 on Build GRCh38
rs995889356 Chr.1: 196910068-196910068 on Build GRCh38
rs1003932736 Chr.1: 196910202-196910202 on Build GRCh38
rs552117202 Chr.1: 196910037-196910037 on Build GRCh38
rs1003594535 Chr.1: 196910258-196910258 on Build GRCh38
rs1014153194 Chr.1: 196910167-196910167 on Build GRCh38
rs777012805 Chr.1: 196910385-196910385 on Build GRCh38
rs753844236 Chr.1: 196910495-196910497 on Build GRCh38
rs574473542 Chr.1: 196910084-196910084 on Build GRCh38
rs534869827 Chr.1: 196910267-196910267 on Build GRCh38
rs746290034 Chr.1: 196910510-196910510 on Build GRCh38
rs780015139 Chr.1: 196910331-196910331 on Build GRCh38
rs921604716 Chr.1: 196910368-196910368 on Build GRCh38
rs184131375 Chr.1: 196910223-196910223 on Build GRCh38
rs764056598 Chr.1: 196910428-196910428 on Build GRCh38
rs752548939 Chr.1: 196910465-196910465 on Build GRCh38
rs762884559 Chr.1: 196910314-196910314 on Build GRCh38
rs959411831 Chr.1: 196910238-196910238 on Build GRCh38
rs956889654 Chr.1: 196910276-196910276 on Build GRCh38
rs267598272 Chr.1: 196910427-196910427 on Build GRCh38
rs1056541773 Chr.1: 196910499-196910499 on Build GRCh38
rs528613959 Chr.1: 196910261-196910261 on Build GRCh38
rs766701460 Chr.1: 196910287-196910287 on Build GRCh38
rs759829132 Chr.1: 196910393-196910393 on Build GRCh38
rs192069928 Chr.1: 196910139-196910139 on Build GRCh38
rs751699894 Chr.1: 196910318-196910318 on Build GRCh38
rs1040867320 Chr.1: 196910095-196910095 on Build GRCh38
rs910990459 Chr.1: 196910051-196910051 on Build GRCh38
rs777172829 Chr.1: 196910471-196910471 on Build GRCh38
rs202044205 Chr.1: 196910279-196910279 on Build GRCh38
rs929814541 Chr.1: 196910340-196910340 on Build GRCh38
rs562079661 Chr.1: 196910272-196910272 on Build GRCh38
rs570664388 Chr.1: 196910406-196910406 on Build GRCh38
rs373614100 Chr.1: 196910166-196910171 on Build GRCh38
rs749214375 Chr.1: 196910255-196910255 on Build GRCh38

View Full Product Details