Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241848592-241849188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO1
Entrez Gene ID
9156
Gene Name
exonuclease 1
Gene Aliases
HEX1, hExoI
UniGene
Hs.498248
Gene Chromosome Location
Chr.1: 241847967-241889939 on Build GRCh38
Transcripts
NM_001319224.1 NM_003686.4 NM_006027.4 NM_130398.3 XM_006711840.2 XM_011544321.1 XM_011544322.1 XM_011544323.2 XM_011544324.2 XM_017002793.1

SNP Detail

SNP ID Chromosome Location
rs115925092 Chr.1: 241848822-241848822 on Build GRCh38
rs1635517 Chr.1: 241848731-241848731 on Build GRCh38
rs561884388 Chr.1: 241848620-241848620 on Build GRCh38
rs978226357 Chr.1: 241848623-241848623 on Build GRCh38
rs990902171 Chr.1: 241849141-241849141 on Build GRCh38
rs1776177 Chr.1: 241848847-241848847 on Build GRCh38
rs957527292 Chr.1: 241848818-241848818 on Build GRCh38
rs4149852 Chr.1: 241849083-241849083 on Build GRCh38
rs993626495 Chr.1: 241848727-241848727 on Build GRCh38
rs966354159 Chr.1: 241848683-241848683 on Build GRCh38
rs866466242 Chr.1: 241848739-241848739 on Build GRCh38
rs997838602 Chr.1: 241848777-241848777 on Build GRCh38
rs1045771074 Chr.1: 241848717-241848717 on Build GRCh38
rs1020650270 Chr.1: 241848673-241848673 on Build GRCh38
rs568690881 Chr.1: 241848636-241848636 on Build GRCh38
rs186184994 Chr.1: 241848740-241848740 on Build GRCh38
rs4149851 Chr.1: 241848791-241848791 on Build GRCh38
rs916125027 Chr.1: 241848670-241848670 on Build GRCh38
rs1032505672 Chr.1: 241848705-241848705 on Build GRCh38
rs146486523 Chr.1: 241849025-241849025 on Build GRCh38
rs113146389 Chr.1: 241848787-241848787 on Build GRCh38
rs4149850 Chr.1: 241848650-241848650 on Build GRCh38
rs547626726 Chr.1: 241848721-241848721 on Build GRCh38
rs554009836 Chr.1: 241849014-241849014 on Build GRCh38
rs372980962 Chr.1: 241849027-241849027 on Build GRCh38
rs1055380623 Chr.1: 241848828-241848828 on Build GRCh38
rs571797194 Chr.1: 241848886-241848886 on Build GRCh38
rs919908119 Chr.1: 241849109-241849110 on Build GRCh38
rs887831003 Chr.1: 241849064-241849064 on Build GRCh38
rs539207153 Chr.1: 241848967-241848967 on Build GRCh38
rs955528611 Chr.1: 241849049-241849049 on Build GRCh38
rs961609449 Chr.1: 241849101-241849101 on Build GRCh38
rs921406106 Chr.1: 241848748-241848748 on Build GRCh38
rs111962605 Chr.1: 241848833-241848833 on Build GRCh38
rs1017484067 Chr.1: 241848761-241848761 on Build GRCh38
rs1054926193 Chr.1: 241848643-241848643 on Build GRCh38
rs938073952 Chr.1: 241848823-241848823 on Build GRCh38
rs1038491803 Chr.1: 241848816-241848816 on Build GRCh38
rs917790970 Chr.1: 241849151-241849151 on Build GRCh38
rs775175195 Chr.1: 241848684-241848684 on Build GRCh38
rs528585350 Chr.1: 241848841-241848841 on Build GRCh38
rs966763403 Chr.1: 241848788-241848788 on Build GRCh38
rs763895762 Chr.1: 241848754-241848754 on Build GRCh38
rs765182600 Chr.1: 241848861-241848861 on Build GRCh38
rs916080605 Chr.1: 241848831-241848831 on Build GRCh38
rs750411041 Chr.1: 241848901-241848901 on Build GRCh38
rs1031973697 Chr.1: 241848986-241848986 on Build GRCh38
rs947532308 Chr.1: 241849059-241849059 on Build GRCh38
rs761454450 Chr.1: 241848646-241848646 on Build GRCh38
rs997468531 Chr.1: 241849116-241849116 on Build GRCh38
rs755055883 Chr.1: 241848931-241848931 on Build GRCh38
rs1006256443 Chr.1: 241849070-241849070 on Build GRCh38
rs887632949 Chr.1: 241848743-241848743 on Build GRCh38
rs567556449 Chr.1: 241848771-241848771 on Build GRCh38
rs111888950 Chr.1: 241848723-241848723 on Build GRCh38
rs531712621 Chr.1: 241849139-241849139 on Build GRCh38
rs4149853 Chr.1: 241849106-241849106 on Build GRCh38
rs896295919 Chr.1: 241848830-241848830 on Build GRCh38
rs924802240 Chr.1: 241848626-241848626 on Build GRCh38
rs925000197 Chr.1: 241848809-241848809 on Build GRCh38
rs1030634318 Chr.1: 241849147-241849147 on Build GRCh38
rs995918932 Chr.1: 241849016-241849016 on Build GRCh38

View Full Product Details