Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80078434-80078969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM254
Entrez Gene ID
80195
Gene Name
transmembrane protein 254
Gene Aliases
C10orf57, bA369J21.6
UniGene
Hs.169982
Gene Chromosome Location
Chr.10: 80078646-80092552 on Build GRCh38
Transcripts
NM_001270370.1 NM_001270371.1 NM_001270372.1 NM_001270373.1 NM_025125.3 XM_011540192.2 XM_017016679.1 NR_027428.1 NR_027429.1 NR_027430.1 NR_027431.2 NR_027432.1

Gene Symbol
TMEM254-AS1
Entrez Gene ID
219347
Gene Name
TMEM254 antisense RNA 1
Gene Aliases
-
UniGene
Hs.524453
Gene Chromosome Location
Chr.10: 80046233-80079193 on Build GRCh38
Transcripts
NM_001270370.1 NM_001270371.1 NM_001270372.1 NM_001270373.1 NM_025125.3 XM_011540192.2 XM_017016679.1 NR_027428.1 NR_027429.1 NR_027430.1 NR_027431.2 NR_027432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7260461 c.73T>A p.F25I Chr.10: 80078772-80078772 on Build GRCh38
7260460 c.73T>A p.F25I Chr.10: 80078772-80078772 on Build GRCh38
466009 c.7A>C p.T3P Chr.10: 80078706-80078706 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374821446 Chr.10: 80078520-80078520 on Build GRCh38
rs527532645 Chr.10: 80078546-80078546 on Build GRCh38
rs947550388 Chr.10: 80078865-80078865 on Build GRCh38
rs756208333 Chr.10: 80078703-80078703 on Build GRCh38
rs951054242 Chr.10: 80078819-80078819 on Build GRCh38
rs904741412 Chr.10: 80078924-80078924 on Build GRCh38
rs368889575 Chr.10: 80078552-80078552 on Build GRCh38
rs961869776 Chr.10: 80078612-80078612 on Build GRCh38
rs866716196 Chr.10: 80078781-80078781 on Build GRCh38
rs992631283 Chr.10: 80078555-80078555 on Build GRCh38
rs958682273 Chr.10: 80078554-80078555 on Build GRCh38
rs753325020 Chr.10: 80078759-80078759 on Build GRCh38
rs755759010 Chr.10: 80078750-80078750 on Build GRCh38
rs928316250 Chr.10: 80078846-80078846 on Build GRCh38
rs112292409 Chr.10: 80078831-80078831 on Build GRCh38
rs753046640 Chr.10: 80078827-80078827 on Build GRCh38
rs3824727 Chr.10: 80078900-80078900 on Build GRCh38
rs769924602 Chr.10: 80078700-80078700 on Build GRCh38
rs200905634 Chr.10: 80078469-80078469 on Build GRCh38
rs373526590 Chr.10: 80078574-80078574 on Build GRCh38
rs766884477 Chr.10: 80078717-80078717 on Build GRCh38
rs1027515903 Chr.10: 80078820-80078820 on Build GRCh38
rs971893615 Chr.10: 80078616-80078616 on Build GRCh38
rs917060695 Chr.10: 80078604-80078604 on Build GRCh38
rs140099300 Chr.10: 80078857-80078857 on Build GRCh38
rs757228293 Chr.10: 80078775-80078775 on Build GRCh38
rs942774898 Chr.10: 80078666-80078666 on Build GRCh38
rs768790185 Chr.10: 80078686-80078686 on Build GRCh38
rs397778570 Chr.10: 80078471-80078472 on Build GRCh38
rs751902920 Chr.10: 80078729-80078729 on Build GRCh38
rs968963074 Chr.10: 80078508-80078508 on Build GRCh38
rs78187241 Chr.10: 80078906-80078906 on Build GRCh38
rs771366026 Chr.10: 80078806-80078806 on Build GRCh38
rs545197259 Chr.10: 80078466-80078466 on Build GRCh38
rs991277489 Chr.10: 80078852-80078852 on Build GRCh38
rs556801637 Chr.10: 80078918-80078918 on Build GRCh38
rs763609843 Chr.10: 80078713-80078713 on Build GRCh38
rs368705890 Chr.10: 80078677-80078677 on Build GRCh38
rs745647460 Chr.10: 80078683-80078683 on Build GRCh38
rs151085716 Chr.10: 80078761-80078761 on Build GRCh38
rs1010830146 Chr.10: 80078484-80078484 on Build GRCh38
rs1055937792 Chr.10: 80078949-80078949 on Build GRCh38
rs760369724 Chr.10: 80078828-80078828 on Build GRCh38
rs772128720 Chr.10: 80078510-80078510 on Build GRCh38
rs759827152 Chr.10: 80078821-80078821 on Build GRCh38
rs1041944701 Chr.10: 80078868-80078868 on Build GRCh38
rs946164395 Chr.10: 80078915-80078915 on Build GRCh38
rs1018059922 Chr.10: 80078784-80078784 on Build GRCh38
rs889153441 Chr.10: 80078899-80078899 on Build GRCh38
rs942679473 Chr.10: 80078883-80078883 on Build GRCh38
rs570736083 Chr.10: 80078633-80078633 on Build GRCh38
rs904771218 Chr.10: 80078725-80078725 on Build GRCh38
rs568205287 Chr.10: 80078851-80078851 on Build GRCh38
rs997938445 Chr.10: 80078722-80078722 on Build GRCh38
rs541845834 Chr.10: 80078937-80078937 on Build GRCh38
rs761996334 Chr.10: 80078694-80078694 on Build GRCh38
rs763092337 Chr.10: 80078800-80078800 on Build GRCh38
rs372087539 Chr.10: 80078798-80078798 on Build GRCh38
rs66971032 Chr.10: 80078470-80078471 on Build GRCh38
rs181443659 Chr.10: 80078925-80078925 on Build GRCh38
rs773418315 Chr.10: 80078710-80078710 on Build GRCh38
rs775879676 Chr.10: 80078787-80078787 on Build GRCh38
rs933616964 Chr.10: 80078878-80078878 on Build GRCh38
rs759899782 Chr.10: 80078735-80078735 on Build GRCh38
rs375260687 Chr.10: 80078823-80078823 on Build GRCh38
rs1055958213 Chr.10: 80078628-80078628 on Build GRCh38
rs1000428633 Chr.10: 80078739-80078739 on Build GRCh38
rs373681540 Chr.10: 80078756-80078756 on Build GRCh38
rs1033975607 Chr.10: 80078747-80078747 on Build GRCh38
rs188953443 Chr.10: 80078812-80078812 on Build GRCh38
rs778868171 Chr.10: 80078776-80078776 on Build GRCh38
rs372311572 Chr.10: 80078742-80078742 on Build GRCh38
rs755566772 Chr.10: 80078663-80078663 on Build GRCh38
rs553900170 Chr.10: 80078874-80078874 on Build GRCh38
rs113611798 Chr.10: 80078476-80078476 on Build GRCh38
rs547606727 Chr.10: 80078605-80078605 on Build GRCh38
rs200326310 Chr.10: 80078785-80078785 on Build GRCh38
rs1051707491 Chr.10: 80078882-80078882 on Build GRCh38
rs869046357 Chr.10: 80078472-80078473 on Build GRCh38
rs775174068 Chr.10: 80078678-80078678 on Build GRCh38
rs763707216 Chr.10: 80078835-80078835 on Build GRCh38
rs1037756264 Chr.10: 80078901-80078901 on Build GRCh38
rs1020456766 Chr.10: 80078485-80078485 on Build GRCh38
rs777672833 Chr.10: 80078812-80078812 on Build GRCh38
rs774866772 Chr.10: 80078810-80078810 on Build GRCh38
rs748443554 Chr.10: 80078794-80078794 on Build GRCh38
rs971860746 Chr.10: 80078813-80078813 on Build GRCh38
rs889184646 Chr.10: 80078748-80078748 on Build GRCh38
rs3842235 Chr.10: 80078469-80078470 on Build GRCh38
rs1048411010 Chr.10: 80078640-80078640 on Build GRCh38
rs928384909 Chr.10: 80078535-80078535 on Build GRCh38
rs752332350 Chr.10: 80078743-80078743 on Build GRCh38
rs781275424 Chr.10: 80078790-80078790 on Build GRCh38
rs930557845 Chr.10: 80078617-80078617 on Build GRCh38
rs375330093 Chr.10: 80078693-80078693 on Build GRCh38
rs199727769 Chr.10: 80078460-80078460 on Build GRCh38
rs749289811 Chr.10: 80078669-80078669 on Build GRCh38

View Full Product Details