Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114435509-114436101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM1
Entrez Gene ID
3983
Gene Name
actin binding LIM protein 1
Gene Aliases
ABLIM, LIMAB1, LIMATIN, abLIM-1
UniGene
Hs.438236
Gene Chromosome Location
Chr.10: 114431110-114821643 on Build GRCh38
Transcripts
NM_001003407.1 NM_001003408.1 NM_001322882.1 NM_001322883.1 NM_001322884.1 NM_001322885.1 NM_001322886.1 NM_001322887.1 NM_001322888.1 NM_001322889.1 NM_001322890.1 NM_001322891.1 NM_001322892.1 NM_001322893.1 NM_001322894.1 NM_001322895.1 NM_001322896.1 NM_001322897.1 NM_001322898.1 NM_001322899.1 NM_001322900.1 NM_002313.5 NM_006720.3 XM_005269824.2 XM_005269826.1 XM_005269827.4 XM_006717837.1 XM_006717846.3 XM_006717847.1 XM_011539801.1 XM_011539802.1 XM_011539803.1 XM_011539805.1 XM_017016242.1 XM_017016243.1 XM_017016244.1 XM_017016245.1 XM_017016246.1 XM_017016247.1 XM_017016248.1 XM_017016249.1 XM_017016250.1 XM_017016251.1 XM_017016252.1 XM_017016253.1 XM_017016254.1 XM_017016255.1 XM_017016256.1 XM_017016258.1 XM_017016259.1

SNP Detail

SNP ID Chromosome Location
rs987122540 Chr.10: 114435947-114435947 on Build GRCh38
rs934578255 Chr.10: 114436064-114436064 on Build GRCh38
rs547499059 Chr.10: 114435858-114435858 on Build GRCh38
rs41293060 Chr.10: 114435835-114435835 on Build GRCh38
rs1045572400 Chr.10: 114436045-114436045 on Build GRCh38
rs1036368870 Chr.10: 114436067-114436067 on Build GRCh38
rs112726690 Chr.10: 114436022-114436022 on Build GRCh38
rs149419975 Chr.10: 114436082-114436082 on Build GRCh38
rs962355045 Chr.10: 114435588-114435589 on Build GRCh38
rs919498948 Chr.10: 114435980-114435980 on Build GRCh38
rs765122538 Chr.10: 114435910-114435910 on Build GRCh38
rs575491521 Chr.10: 114435834-114435834 on Build GRCh38
rs1039404886 Chr.10: 114435981-114435981 on Build GRCh38
rs907127231 Chr.10: 114435888-114435888 on Build GRCh38
rs747100903 Chr.10: 114436074-114436074 on Build GRCh38
rs115007772 Chr.10: 114435641-114435641 on Build GRCh38
rs185144991 Chr.10: 114435895-114435895 on Build GRCh38
rs760840887 Chr.10: 114435830-114435830 on Build GRCh38
rs1017934183 Chr.10: 114435569-114435569 on Build GRCh38
rs1046765765 Chr.10: 114436023-114436023 on Build GRCh38
rs774489726 Chr.10: 114435749-114435749 on Build GRCh38
rs1057139 Chr.10: 114436017-114436017 on Build GRCh38
rs1006553967 Chr.10: 114435810-114435810 on Build GRCh38
rs914544479 Chr.10: 114435691-114435691 on Build GRCh38
rs991574669 Chr.10: 114435689-114435689 on Build GRCh38
rs1049370447 Chr.10: 114435537-114435537 on Build GRCh38
rs947384259 Chr.10: 114435693-114435696 on Build GRCh38
rs879736626 Chr.10: 114435973-114435973 on Build GRCh38
rs79672630 Chr.10: 114435922-114435922 on Build GRCh38
rs1013153672 Chr.10: 114435632-114435632 on Build GRCh38
rs1045970111 Chr.10: 114435860-114435860 on Build GRCh38
rs377052963 Chr.10: 114435940-114435940 on Build GRCh38
rs534733206 Chr.10: 114435588-114435588 on Build GRCh38
rs150283420 Chr.10: 114435771-114435771 on Build GRCh38
rs548338608 Chr.10: 114435921-114435921 on Build GRCh38
rs531236888 Chr.10: 114435974-114435974 on Build GRCh38
rs1031308039 Chr.10: 114435688-114435688 on Build GRCh38
rs944948915 Chr.10: 114435774-114435774 on Build GRCh38
rs376537977 Chr.10: 114435949-114435949 on Build GRCh38
rs1003451682 Chr.10: 114435849-114435849 on Build GRCh38
rs527541249 Chr.10: 114435848-114435848 on Build GRCh38
rs554862103 Chr.10: 114435742-114435742 on Build GRCh38
rs7919465 Chr.10: 114436051-114436051 on Build GRCh38
rs935144195 Chr.10: 114435743-114435743 on Build GRCh38
rs879567739 Chr.10: 114435640-114435640 on Build GRCh38
rs74160408 Chr.10: 114435705-114435705 on Build GRCh38
rs10978 Chr.10: 114435840-114435840 on Build GRCh38
rs563940685 Chr.10: 114435839-114435839 on Build GRCh38
rs986686194 Chr.10: 114435755-114435755 on Build GRCh38
rs1011644878 Chr.10: 114435920-114435920 on Build GRCh38
rs112346723 Chr.10: 114436021-114436021 on Build GRCh38
rs923843665 Chr.10: 114435935-114435935 on Build GRCh38
rs181359735 Chr.10: 114435890-114435890 on Build GRCh38
rs1025839251 Chr.10: 114435836-114435836 on Build GRCh38
rs928703018 Chr.10: 114435686-114435686 on Build GRCh38
rs998556374 Chr.10: 114435670-114435670 on Build GRCh38
rs569995789 Chr.10: 114435923-114435923 on Build GRCh38
rs948780968 Chr.10: 114436027-114436027 on Build GRCh38
rs972044606 Chr.10: 114435805-114435805 on Build GRCh38
rs772169221 Chr.10: 114435731-114435731 on Build GRCh38
rs541489220 Chr.10: 114435717-114435717 on Build GRCh38
rs972263868 Chr.10: 114435978-114435978 on Build GRCh38
rs971303184 Chr.10: 114435642-114435642 on Build GRCh38
rs989893091 Chr.10: 114435739-114435739 on Build GRCh38

View Full Product Details