Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95956425-95956877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMDHD1
Entrez Gene ID
144193
Gene Name
amidohydrolase domain containing 1
Gene Aliases
HMFT1272
UniGene
Hs.424907
Gene Chromosome Location
Chr.12: 95943293-95968592 on Build GRCh38
Transcripts
NM_152435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2046459 c.452C>T p.T151M Chr.12: 95956827-95956827 on Build GRCh38
7236503 c.389C>T p.T130I Chr.12: 95956764-95956764 on Build GRCh38
5446401 c.464_465insTGT p.C155_K156insV Chr.12: 95956839-95956840 on Build GRCh38
7236648 c.426C>G p.L142L Chr.12: 95956801-95956801 on Build GRCh38
1364988 c.373C>T p.R125C Chr.12: 95956748-95956748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755703276 Chr.12: 95956747-95956747 on Build GRCh38
rs746297553 Chr.12: 95956757-95956757 on Build GRCh38
rs138716150 Chr.12: 95956666-95956666 on Build GRCh38
rs779552415 Chr.12: 95956755-95956755 on Build GRCh38
rs377372971 Chr.12: 95956571-95956571 on Build GRCh38
rs530496824 Chr.12: 95956742-95956742 on Build GRCh38
rs145626384 Chr.12: 95956715-95956715 on Build GRCh38
rs764118926 Chr.12: 95956704-95956704 on Build GRCh38
rs551304292 Chr.12: 95956779-95956779 on Build GRCh38
rs751426964 Chr.12: 95956727-95956727 on Build GRCh38
rs779518192 Chr.12: 95956815-95956815 on Build GRCh38
rs1043621391 Chr.12: 95956516-95956516 on Build GRCh38
rs766353218 Chr.12: 95956781-95956781 on Build GRCh38
rs772293939 Chr.12: 95956760-95956760 on Build GRCh38
rs748266716 Chr.12: 95956851-95956851 on Build GRCh38
rs758813223 Chr.12: 95956832-95956832 on Build GRCh38
rs759403725 Chr.12: 95956787-95956787 on Build GRCh38
rs201374830 Chr.12: 95956812-95956812 on Build GRCh38
rs966074213 Chr.12: 95956465-95956465 on Build GRCh38
rs552095079 Chr.12: 95956445-95956445 on Build GRCh38
rs4762651 Chr.12: 95956476-95956476 on Build GRCh38
rs369034222 Chr.12: 95956780-95956780 on Build GRCh38
rs766298210 Chr.12: 95956723-95956723 on Build GRCh38
rs372986173 Chr.12: 95956654-95956654 on Build GRCh38
rs192045864 Chr.12: 95956638-95956638 on Build GRCh38
rs925143393 Chr.12: 95956795-95956795 on Build GRCh38
rs1018694556 Chr.12: 95956503-95956528 on Build GRCh38
rs746321378 Chr.12: 95956827-95956827 on Build GRCh38
rs913683696 Chr.12: 95956485-95956485 on Build GRCh38
rs752248228 Chr.12: 95956741-95956741 on Build GRCh38
rs897953184 Chr.12: 95956609-95956609 on Build GRCh38
rs1045688478 Chr.12: 95956595-95956595 on Build GRCh38
rs1007826530 Chr.12: 95956667-95956667 on Build GRCh38
rs540496571 Chr.12: 95956643-95956643 on Build GRCh38
rs749701037 Chr.12: 95956842-95956842 on Build GRCh38
rs534815977 Chr.12: 95956849-95956849 on Build GRCh38
rs187366868 Chr.12: 95956626-95956626 on Build GRCh38
rs767164703 Chr.12: 95956790-95956790 on Build GRCh38
rs763445534 Chr.12: 95956749-95956749 on Build GRCh38
rs375853196 Chr.12: 95956703-95956703 on Build GRCh38
rs201085031 Chr.12: 95956749-95956749 on Build GRCh38
rs1042965599 Chr.12: 95956510-95956510 on Build GRCh38
rs527424065 Chr.12: 95956818-95956818 on Build GRCh38
rs768922385 Chr.12: 95956844-95956844 on Build GRCh38
rs762821619 Chr.12: 95956720-95956720 on Build GRCh38
rs754706271 Chr.12: 95956733-95956733 on Build GRCh38
rs775813472 Chr.12: 95956773-95956773 on Build GRCh38
rs757909459 Chr.12: 95956754-95956754 on Build GRCh38
rs774124190 Chr.12: 95956783-95956783 on Build GRCh38
rs772400510 Chr.12: 95956690-95956690 on Build GRCh38
rs556499631 Chr.12: 95956464-95956464 on Build GRCh38
rs769824213 Chr.12: 95956854-95956854 on Build GRCh38
rs937951124 Chr.12: 95956544-95956544 on Build GRCh38
rs200378404 Chr.12: 95956806-95956806 on Build GRCh38
rs1055848507 Chr.12: 95956475-95956475 on Build GRCh38
rs1055064885 Chr.12: 95956802-95956802 on Build GRCh38
rs773837719 Chr.12: 95956674-95956675 on Build GRCh38
rs759816920 Chr.12: 95956679-95956679 on Build GRCh38
rs993534377 Chr.12: 95956615-95956615 on Build GRCh38
rs780936530 Chr.12: 95956738-95956738 on Build GRCh38
rs986751544 Chr.12: 95956455-95956455 on Build GRCh38
rs982843448 Chr.12: 95956730-95956730 on Build GRCh38
rs151046034 Chr.12: 95956793-95956793 on Build GRCh38
rs112657735 Chr.12: 95956837-95956837 on Build GRCh38
rs199693405 Chr.12: 95956808-95956808 on Build GRCh38
rs1055414367 Chr.12: 95956547-95956547 on Build GRCh38
rs755758291 Chr.12: 95956801-95956801 on Build GRCh38
rs894495763 Chr.12: 95956558-95956558 on Build GRCh38
rs945090387 Chr.12: 95956486-95956486 on Build GRCh38
rs778451768 Chr.12: 95956639-95956639 on Build GRCh38
rs143138689 Chr.12: 95956834-95956834 on Build GRCh38
rs928634093 Chr.12: 95956748-95956748 on Build GRCh38
rs756466605 Chr.12: 95956543-95956543 on Build GRCh38
rs966486330 Chr.12: 95956792-95956792 on Build GRCh38
rs771138674 Chr.12: 95956776-95956777 on Build GRCh38

View Full Product Details