Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69592980-69593576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT2
Entrez Gene ID
10576
Gene Name
chaperonin containing TCP1 subunit 2
Gene Aliases
99D8.1, CCT-beta, CCTB, HEL-S-100n, PRO1633, TCP-1-beta
UniGene
Hs.189772
Gene Chromosome Location
Chr.12: 69585428-69601577 on Build GRCh38
Transcripts
NM_001198842.1 NM_006431.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7080408 c.847C>A p.L283I Chr.12: 69593072-69593072 on Build GRCh38
943026 c.768A>G p.V256V Chr.12: 69592993-69592993 on Build GRCh38
4147425 c.783A>G p.T261T Chr.12: 69593008-69593008 on Build GRCh38
6907979 c.860T>C p.I287T Chr.12: 69593085-69593085 on Build GRCh38
1470631 c.780T>C p.S260S Chr.12: 69593005-69593005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879543757 Chr.12: 69593386-69593386 on Build GRCh38
rs979587319 Chr.12: 69593379-69593379 on Build GRCh38
rs879008739 Chr.12: 69593239-69593239 on Build GRCh38
rs896513885 Chr.12: 69593189-69593189 on Build GRCh38
rs968958164 Chr.12: 69593269-69593269 on Build GRCh38
rs1016755139 Chr.12: 69593266-69593266 on Build GRCh38
rs960238203 Chr.12: 69593225-69593225 on Build GRCh38
rs539249032 Chr.12: 69593337-69593337 on Build GRCh38
rs745676913 Chr.12: 69593013-69593013 on Build GRCh38
rs576051656 Chr.12: 69593493-69593493 on Build GRCh38
rs916130143 Chr.12: 69593265-69593266 on Build GRCh38
rs914166454 Chr.12: 69593350-69593350 on Build GRCh38
rs777539486 Chr.12: 69593326-69593326 on Build GRCh38
rs745840327 Chr.12: 69593127-69593127 on Build GRCh38
rs761734093 Chr.12: 69593044-69593044 on Build GRCh38
rs868722288 Chr.12: 69593009-69593009 on Build GRCh38
rs766860904 Chr.12: 69593490-69593490 on Build GRCh38
rs1013009944 Chr.12: 69593480-69593480 on Build GRCh38
rs753399620 Chr.12: 69593116-69593116 on Build GRCh38
rs1037688788 Chr.12: 69593401-69593401 on Build GRCh38
rs369840085 Chr.12: 69593034-69593034 on Build GRCh38
rs932432382 Chr.12: 69593347-69593347 on Build GRCh38
rs771949290 Chr.12: 69593024-69593024 on Build GRCh38
rs934267533 Chr.12: 69593007-69593007 on Build GRCh38
rs148760765 Chr.12: 69593534-69593534 on Build GRCh38
rs553636864 Chr.12: 69593281-69593281 on Build GRCh38
rs551629629 Chr.12: 69593066-69593066 on Build GRCh38
rs975124686 Chr.12: 69593283-69593283 on Build GRCh38
rs745354900 Chr.12: 69593295-69593295 on Build GRCh38
rs780051604 Chr.12: 69593149-69593149 on Build GRCh38
rs763861518 Chr.12: 69593129-69593129 on Build GRCh38
rs532721380 Chr.12: 69593309-69593312 on Build GRCh38
rs747282578 Chr.12: 69593032-69593032 on Build GRCh38
rs773661302 Chr.12: 69593059-69593059 on Build GRCh38
rs756899750 Chr.12: 69593510-69593510 on Build GRCh38
rs77944709 Chr.12: 69593399-69593399 on Build GRCh38
rs1035699110 Chr.12: 69593421-69593421 on Build GRCh38
rs924296823 Chr.12: 69593304-69593304 on Build GRCh38
rs1043434 Chr.12: 69593008-69593008 on Build GRCh38
rs764887949 Chr.12: 69593513-69593513 on Build GRCh38
rs763651446 Chr.12: 69593110-69593110 on Build GRCh38
rs751903258 Chr.12: 69593076-69593076 on Build GRCh38
rs776661184 Chr.12: 69593469-69593469 on Build GRCh38
rs746793912 Chr.12: 69593461-69593461 on Build GRCh38
rs945601737 Chr.12: 69593366-69593366 on Build GRCh38
rs748282427 Chr.12: 69593478-69593478 on Build GRCh38
rs774766161 Chr.12: 69593492-69593492 on Build GRCh38
rs941293730 Chr.12: 69593394-69593395 on Build GRCh38
rs758474417 Chr.12: 69593552-69593552 on Build GRCh38
rs897849111 Chr.12: 69593407-69593407 on Build GRCh38
rs557573914 Chr.12: 69593391-69593391 on Build GRCh38
rs955255071 Chr.12: 69593329-69593329 on Build GRCh38
rs1055658846 Chr.12: 69593529-69593529 on Build GRCh38
rs535389481 Chr.12: 69593224-69593224 on Build GRCh38
rs753483720 Chr.12: 69593128-69593129 on Build GRCh38
rs549325342 Chr.12: 69593212-69593212 on Build GRCh38
rs778935918 Chr.12: 69593125-69593125 on Build GRCh38
rs755856520 Chr.12: 69593078-69593078 on Build GRCh38
rs758386451 Chr.12: 69593144-69593144 on Build GRCh38
rs749254276 Chr.12: 69593041-69593041 on Build GRCh38
rs201295125 Chr.12: 69593516-69593516 on Build GRCh38
rs753388666 Chr.12: 69593507-69593507 on Build GRCh38
rs530820234 Chr.12: 69593181-69593181 on Build GRCh38
rs746862418 Chr.12: 69593421-69593423 on Build GRCh38
rs778655085 Chr.12: 69593010-69593010 on Build GRCh38
rs565580558 Chr.12: 69593297-69593297 on Build GRCh38
rs1004062362 Chr.12: 69593248-69593248 on Build GRCh38
rs773249670 Chr.12: 69593485-69593485 on Build GRCh38
rs903959494 Chr.12: 69593077-69593077 on Build GRCh38
rs766774842 Chr.12: 69593069-69593069 on Build GRCh38
rs991621607 Chr.12: 69593247-69593247 on Build GRCh38
rs552533563 Chr.12: 69593367-69593367 on Build GRCh38
rs141561396 Chr.12: 69593220-69593220 on Build GRCh38
rs765789089 Chr.12: 69593055-69593055 on Build GRCh38
rs763503631 Chr.12: 69593486-69593486 on Build GRCh38
rs763520566 Chr.12: 69593500-69593500 on Build GRCh38
rs994011581 Chr.12: 69593214-69593214 on Build GRCh38
rs976941146 Chr.12: 69593309-69593309 on Build GRCh38
rs570084885 Chr.12: 69593168-69593168 on Build GRCh38
rs938071508 Chr.12: 69593084-69593084 on Build GRCh38
rs986464622 Chr.12: 69593340-69593340 on Build GRCh38
rs775393702 Chr.12: 69593026-69593026 on Build GRCh38
rs1028063555 Chr.12: 69593285-69593285 on Build GRCh38
rs768928958 Chr.12: 69593465-69593465 on Build GRCh38
rs895912664 Chr.12: 69593467-69593467 on Build GRCh38
rs756669993 Chr.12: 69593123-69593123 on Build GRCh38
rs201962311 Chr.12: 69593005-69593005 on Build GRCh38
rs770719405 Chr.12: 69593482-69593482 on Build GRCh38

View Full Product Details