Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110041783-110042382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf76
Entrez Gene ID
400073
Gene Name
chromosome 12 open reading frame 76
Gene Aliases
-
UniGene
Hs.44817
Gene Chromosome Location
Chr.12: 110041177-110067695 on Build GRCh38
Transcripts
NM_207435.2 XM_017019297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285987 c.382G>T p.V128L Chr.12: 110042377-110042377 on Build GRCh38
118431 c.378G>A p.A126A Chr.12: 110042381-110042381 on Build GRCh38
934911 c.377C>T p.A126V Chr.12: 110042382-110042382 on Build GRCh38
5456257 c.406T>C p.*136R Chr.12: 110042353-110042353 on Build GRCh38
934910 c.392G>T p.R131I Chr.12: 110042367-110042367 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs982704842 Chr.12: 110042173-110042174 on Build GRCh38
rs767841323 Chr.12: 110041909-110041909 on Build GRCh38
rs888170138 Chr.12: 110042325-110042325 on Build GRCh38
rs747693711 Chr.12: 110042335-110042335 on Build GRCh38
rs766524138 Chr.12: 110042015-110042015 on Build GRCh38
rs756229703 Chr.12: 110042051-110042051 on Build GRCh38
rs551712600 Chr.12: 110042167-110042167 on Build GRCh38
rs878905115 Chr.12: 110041921-110041921 on Build GRCh38
rs750641242 Chr.12: 110042000-110042000 on Build GRCh38
rs754568927 Chr.12: 110042323-110042323 on Build GRCh38
rs1040968879 Chr.12: 110041837-110041837 on Build GRCh38
rs11550081 Chr.12: 110042313-110042313 on Build GRCh38
rs947510443 Chr.12: 110042097-110042097 on Build GRCh38
rs192178810 Chr.12: 110041877-110041877 on Build GRCh38
rs754943370 Chr.12: 110042109-110042109 on Build GRCh38
rs372750833 Chr.12: 110042301-110042301 on Build GRCh38
rs549658868 Chr.12: 110041920-110041921 on Build GRCh38
rs1043213432 Chr.12: 110041938-110041938 on Build GRCh38
rs899610622 Chr.12: 110041894-110041894 on Build GRCh38
rs181702859 Chr.12: 110042146-110042146 on Build GRCh38
rs533579661 Chr.12: 110042213-110042213 on Build GRCh38
rs764468007 Chr.12: 110041842-110041842 on Build GRCh38
rs375672765 Chr.12: 110041986-110041986 on Build GRCh38
rs752489942 Chr.12: 110042027-110042027 on Build GRCh38
rs973520889 Chr.12: 110042141-110042141 on Build GRCh38
rs969069008 Chr.12: 110041972-110041972 on Build GRCh38
rs202140938 Chr.12: 110041921-110041922 on Build GRCh38
rs999100585 Chr.12: 110041995-110041997 on Build GRCh38
rs780281581 Chr.12: 110042315-110042315 on Build GRCh38
rs758122633 Chr.12: 110042266-110042266 on Build GRCh38
rs774582060 Chr.12: 110042121-110042121 on Build GRCh38
rs1047460196 Chr.12: 110041825-110041825 on Build GRCh38
rs901004269 Chr.12: 110041851-110041851 on Build GRCh38
rs749174511 Chr.12: 110042354-110042354 on Build GRCh38
rs988985037 Chr.12: 110042056-110042056 on Build GRCh38
rs764928781 Chr.12: 110042190-110042190 on Build GRCh38
rs1023019053 Chr.12: 110042069-110042069 on Build GRCh38
rs960039770 Chr.12: 110042045-110042045 on Build GRCh38
rs762028084 Chr.12: 110041893-110041893 on Build GRCh38
rs920978743 Chr.12: 110042220-110042220 on Build GRCh38
rs904677727 Chr.12: 110042337-110042337 on Build GRCh38
rs758300256 Chr.12: 110042153-110042153 on Build GRCh38
rs1040937687 Chr.12: 110042202-110042202 on Build GRCh38
rs757871041 Chr.12: 110042305-110042305 on Build GRCh38
rs1045248266 Chr.12: 110042352-110042352 on Build GRCh38
rs780089425 Chr.12: 110042083-110042083 on Build GRCh38
rs374404314 Chr.12: 110042331-110042331 on Build GRCh38
rs751959269 Chr.12: 110041861-110041861 on Build GRCh38
rs943729762 Chr.12: 110041884-110041884 on Build GRCh38
rs746682172 Chr.12: 110042317-110042317 on Build GRCh38
rs566742489 Chr.12: 110042200-110042200 on Build GRCh38
rs942602472 Chr.12: 110041810-110041810 on Build GRCh38
rs753870389 Chr.12: 110042096-110042096 on Build GRCh38
rs1023319111 Chr.12: 110041946-110041946 on Build GRCh38
rs555325986 Chr.12: 110042338-110042338 on Build GRCh38
rs370206147 Chr.12: 110042350-110042350 on Build GRCh38
rs767315528 Chr.12: 110042202-110042202 on Build GRCh38
rs765886542 Chr.12: 110042306-110042306 on Build GRCh38
rs569073998 Chr.12: 110041921-110041921 on Build GRCh38
rs919756483 Chr.12: 110042166-110042166 on Build GRCh38

View Full Product Details