Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35825945-35826369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGO4
Entrez Gene ID
192670
Gene Name
argonaute 4, RISC catalytic component
Gene Aliases
EIF2C4
UniGene
Hs.744858
Gene Chromosome Location
Chr.1: 35807630-35857890 on Build GRCh38
Transcripts
NM_017629.3 XM_005270578.3 XM_005270579.3 XM_011540883.2 XM_011540884.2 XM_017000529.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638588 c.699C>G p.I233M Chr.1: 35825999-35825999 on Build GRCh38
908782 c.751G>T p.E251* Chr.1: 35826051-35826051 on Build GRCh38
213377 c.654G>T p.Q218H Chr.1: 35825954-35825954 on Build GRCh38
4965664 c.646C>T p.R216W Chr.1: 35825946-35825946 on Build GRCh38
70585 c.748A>G p.K250E Chr.1: 35826048-35826048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045253488 Chr.1: 35825965-35825965 on Build GRCh38
rs951842986 Chr.1: 35826343-35826343 on Build GRCh38
rs769878814 Chr.1: 35825987-35825987 on Build GRCh38
rs181044708 Chr.1: 35826245-35826245 on Build GRCh38
rs376648938 Chr.1: 35825970-35825970 on Build GRCh38
rs571809564 Chr.1: 35826290-35826290 on Build GRCh38
rs763638798 Chr.1: 35826110-35826110 on Build GRCh38
rs374712943 Chr.1: 35826100-35826100 on Build GRCh38
rs987438339 Chr.1: 35826307-35826307 on Build GRCh38
rs962272476 Chr.1: 35826284-35826284 on Build GRCh38
rs887377341 Chr.1: 35826139-35826139 on Build GRCh38
rs774932844 Chr.1: 35826040-35826040 on Build GRCh38
rs1021668348 Chr.1: 35826234-35826234 on Build GRCh38
rs768306920 Chr.1: 35826286-35826286 on Build GRCh38
rs1012952603 Chr.1: 35826182-35826182 on Build GRCh38
rs999278426 Chr.1: 35826256-35826256 on Build GRCh38
rs894681710 Chr.1: 35826174-35826174 on Build GRCh38
rs369911053 Chr.1: 35826079-35826079 on Build GRCh38
rs749366407 Chr.1: 35825997-35825997 on Build GRCh38
rs147992114 Chr.1: 35825972-35825972 on Build GRCh38
rs556510403 Chr.1: 35826279-35826279 on Build GRCh38
rs189308037 Chr.1: 35826086-35826086 on Build GRCh38
rs987609121 Chr.1: 35826236-35826236 on Build GRCh38
rs756229224 Chr.1: 35825978-35825978 on Build GRCh38
rs761168372 Chr.1: 35826111-35826111 on Build GRCh38
rs772771668 Chr.1: 35826029-35826029 on Build GRCh38
rs148256871 Chr.1: 35825996-35825996 on Build GRCh38
rs554957549 Chr.1: 35826034-35826034 on Build GRCh38
rs953451135 Chr.1: 35826197-35826197 on Build GRCh38
rs537934470 Chr.1: 35826184-35826184 on Build GRCh38
rs72659690 Chr.1: 35826149-35826149 on Build GRCh38
rs939147119 Chr.1: 35826056-35826056 on Build GRCh38
rs1037492733 Chr.1: 35826002-35826002 on Build GRCh38
rs781729877 Chr.1: 35826077-35826078 on Build GRCh38
rs747480741 Chr.1: 35826247-35826247 on Build GRCh38
rs760141856 Chr.1: 35826073-35826073 on Build GRCh38
rs920268272 Chr.1: 35826342-35826342 on Build GRCh38
rs748682202 Chr.1: 35826111-35826112 on Build GRCh38
rs1003152089 Chr.1: 35826144-35826144 on Build GRCh38

View Full Product Details