Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32013490-32014062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDRBS1
Entrez Gene ID
10657
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Gene Aliases
Sam68, p62, p68
UniGene
Hs.445893
Gene Chromosome Location
Chr.1: 32013694-32060859 on Build GRCh38
Transcripts
NM_001271878.1 NM_006559.2 NR_073498.1 NR_073499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4633794 c.29G>A p.R10H Chr.1: 32014024-32014024 on Build GRCh38
4881607 c.67C>A p.P23T Chr.1: 32014062-32014062 on Build GRCh38
4772345 c.65A>C p.D22A Chr.1: 32014060-32014060 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951207345 Chr.1: 32013880-32013880 on Build GRCh38
rs752713376 Chr.1: 32014037-32014037 on Build GRCh38
rs947210162 Chr.1: 32013825-32013825 on Build GRCh38
rs894277797 Chr.1: 32013660-32013660 on Build GRCh38
rs1022827668 Chr.1: 32013541-32013541 on Build GRCh38
rs1035775809 Chr.1: 32013886-32013886 on Build GRCh38
rs912579924 Chr.1: 32013991-32013991 on Build GRCh38
rs886696924 Chr.1: 32013641-32013641 on Build GRCh38
rs944005169 Chr.1: 32014026-32014026 on Build GRCh38
rs982518354 Chr.1: 32013885-32013885 on Build GRCh38
rs368464409 Chr.1: 32013771-32013771 on Build GRCh38
rs1012366103 Chr.1: 32013680-32013680 on Build GRCh38
rs139589535 Chr.1: 32013759-32013759 on Build GRCh38
rs540743924 Chr.1: 32013887-32013887 on Build GRCh38
rs1034983055 Chr.1: 32013517-32013517 on Build GRCh38
rs779236077 Chr.1: 32013947-32013947 on Build GRCh38
rs769652619 Chr.1: 32013988-32013988 on Build GRCh38
rs530783649 Chr.1: 32013984-32013984 on Build GRCh38
rs576447463 Chr.1: 32013760-32013760 on Build GRCh38
rs999926265 Chr.1: 32013705-32013705 on Build GRCh38
rs944480806 Chr.1: 32013608-32013608 on Build GRCh38
rs6667294 Chr.1: 32013951-32013951 on Build GRCh38
rs766284175 Chr.1: 32014028-32014028 on Build GRCh38
rs111483654 Chr.1: 32013523-32013523 on Build GRCh38
rs938567421 Chr.1: 32013655-32013655 on Build GRCh38
rs1045035277 Chr.1: 32013865-32013865 on Build GRCh38
rs982852439 Chr.1: 32013748-32013748 on Build GRCh38
rs115349425 Chr.1: 32013627-32013627 on Build GRCh38
rs891610883 Chr.1: 32013892-32013892 on Build GRCh38
rs769085204 Chr.1: 32013747-32013747 on Build GRCh38
rs772931806 Chr.1: 32014016-32014016 on Build GRCh38
rs955601806 Chr.1: 32013555-32013555 on Build GRCh38
rs1032812238 Chr.1: 32013717-32013717 on Build GRCh38
rs938604122 Chr.1: 32013780-32013780 on Build GRCh38
rs150564278 Chr.1: 32013678-32013678 on Build GRCh38
rs948151743 Chr.1: 32013662-32013662 on Build GRCh38
rs896521286 Chr.1: 32013841-32013841 on Build GRCh38
rs76080682 Chr.1: 32013930-32013930 on Build GRCh38
rs536031890 Chr.1: 32013524-32013524 on Build GRCh38
rs918176208 Chr.1: 32014019-32014019 on Build GRCh38
rs968561946 Chr.1: 32013923-32013923 on Build GRCh38
rs772493181 Chr.1: 32013961-32013961 on Build GRCh38
rs1052875512 Chr.1: 32013831-32013831 on Build GRCh38
rs377526856 Chr.1: 32013605-32013605 on Build GRCh38
rs911356486 Chr.1: 32013595-32013595 on Build GRCh38
rs768370132 Chr.1: 32013969-32013969 on Build GRCh38
rs746579121 Chr.1: 32013963-32013963 on Build GRCh38
rs1008448126 Chr.1: 32013719-32013719 on Build GRCh38
rs900028415 Chr.1: 32013861-32013861 on Build GRCh38
rs1031291818 Chr.1: 32013711-32013711 on Build GRCh38
rs1055555435 Chr.1: 32013659-32013659 on Build GRCh38
rs929611670 Chr.1: 32013630-32013630 on Build GRCh38
rs963798659 Chr.1: 32013596-32013596 on Build GRCh38
rs776431312 Chr.1: 32013982-32013982 on Build GRCh38
rs956163539 Chr.1: 32013920-32013920 on Build GRCh38
rs891377439 Chr.1: 32013714-32013714 on Build GRCh38
rs955944351 Chr.1: 32013574-32013574 on Build GRCh38
rs925860390 Chr.1: 32013571-32013571 on Build GRCh38
rs970281838 Chr.1: 32013548-32013548 on Build GRCh38
rs1012313889 Chr.1: 32013536-32013536 on Build GRCh38
rs760747849 Chr.1: 32014039-32014039 on Build GRCh38
rs982745439 Chr.1: 32013632-32013632 on Build GRCh38
rs999806343 Chr.1: 32013533-32013533 on Build GRCh38
rs574486117 Chr.1: 32013921-32013921 on Build GRCh38
rs928532700 Chr.1: 32013635-32013635 on Build GRCh38
rs542002614 Chr.1: 32013925-32013925 on Build GRCh38
rs757900703 Chr.1: 32013856-32013856 on Build GRCh38
rs951904057 Chr.1: 32013942-32013942 on Build GRCh38
rs987374388 Chr.1: 32013592-32013592 on Build GRCh38
rs745364198 Chr.1: 32013516-32013516 on Build GRCh38
rs746667276 Chr.1: 32013739-32013739 on Build GRCh38
rs868111502 Chr.1: 32013845-32013845 on Build GRCh38
rs764353405 Chr.1: 32013947-32013957 on Build GRCh38
rs757671366 Chr.1: 32013945-32013945 on Build GRCh38
rs762917162 Chr.1: 32014024-32014024 on Build GRCh38
rs903934148 Chr.1: 32013829-32013829 on Build GRCh38
rs903944179 Chr.1: 32013866-32013866 on Build GRCh38
rs371122373 Chr.1: 32013735-32013735 on Build GRCh38
rs935392778 Chr.1: 32013830-32013830 on Build GRCh38
rs964186327 Chr.1: 32013728-32013728 on Build GRCh38
rs1018469229 Chr.1: 32013724-32013724 on Build GRCh38
rs995663670 Chr.1: 32013871-32013871 on Build GRCh38
rs12746359 Chr.1: 32014036-32014036 on Build GRCh38
rs765746078 Chr.1: 32013874-32013874 on Build GRCh38
rs771543382 Chr.1: 32013576-32013576 on Build GRCh38
rs764259849 Chr.1: 32014041-32014041 on Build GRCh38
rs1056718089 Chr.1: 32013836-32013836 on Build GRCh38
rs974499563 Chr.1: 32013782-32013782 on Build GRCh38
rs991394559 Chr.1: 32013903-32013903 on Build GRCh38
rs922154628 Chr.1: 32013618-32013618 on Build GRCh38
rs367965800 Chr.1: 32013889-32013889 on Build GRCh38
rs1023717060 Chr.1: 32013697-32013697 on Build GRCh38
rs764747249 Chr.1: 32013666-32013666 on Build GRCh38
rs921677973 Chr.1: 32013784-32013785 on Build GRCh38
rs965725793 Chr.1: 32013905-32013905 on Build GRCh38
rs978734901 Chr.1: 32013566-32013569 on Build GRCh38
rs574555583 Chr.1: 32013800-32013800 on Build GRCh38
rs1043823754 Chr.1: 32013827-32013827 on Build GRCh38
rs967998460 Chr.1: 32013530-32013530 on Build GRCh38
rs540375364 Chr.1: 32013822-32013822 on Build GRCh38
rs1031657708 Chr.1: 32013534-32013534 on Build GRCh38
rs891100704 Chr.1: 32013848-32013848 on Build GRCh38
rs924437024 Chr.1: 32013941-32013941 on Build GRCh38
rs1038742832 Chr.1: 32013612-32013612 on Build GRCh38
rs1018518241 Chr.1: 32013859-32013859 on Build GRCh38
rs951111199 Chr.1: 32013740-32013740 on Build GRCh38

View Full Product Details