Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167547961-167548523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREG1
Entrez Gene ID
8804
Gene Name
cellular repressor of E1A stimulated genes 1
Gene Aliases
CREG
UniGene
Hs.5710
Gene Chromosome Location
Chr.1: 167541013-167553819 on Build GRCh38
Transcripts
NM_003851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5793472 c.355-6C>G p.? Chr.1: 167548127-167548127 on Build GRCh38
5378326 c.434C>T p.P145L Chr.1: 167548042-167548042 on Build GRCh38
3477805 c.433C>A p.P145T Chr.1: 167548043-167548043 on Build GRCh38
4978238 c.386T>A p.L129* Chr.1: 167548090-167548090 on Build GRCh38
5910157 c.361C>T p.P121S Chr.1: 167548115-167548115 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs143114354 Chr.1: 167548120-167548120 on Build GRCh38
rs866369758 Chr.1: 167548045-167548045 on Build GRCh38
rs371833839 Chr.1: 167548148-167548148 on Build GRCh38
rs74119804 Chr.1: 167548315-167548315 on Build GRCh38
rs150841373 Chr.1: 167548197-167548197 on Build GRCh38
rs868252102 Chr.1: 167548179-167548179 on Build GRCh38
rs984319973 Chr.1: 167548491-167548491 on Build GRCh38
rs778294403 Chr.1: 167548091-167548091 on Build GRCh38
rs765953608 Chr.1: 167548071-167548071 on Build GRCh38
rs987880466 Chr.1: 167548230-167548230 on Build GRCh38
rs984003250 Chr.1: 167548198-167548198 on Build GRCh38
rs997849421 Chr.1: 167548468-167548468 on Build GRCh38
rs537683141 Chr.1: 167548326-167548326 on Build GRCh38
rs939953738 Chr.1: 167548238-167548238 on Build GRCh38
rs569456371 Chr.1: 167548440-167548440 on Build GRCh38
rs1054632835 Chr.1: 167548411-167548411 on Build GRCh38
rs898537767 Chr.1: 167548388-167548388 on Build GRCh38
rs560351191 Chr.1: 167548159-167548159 on Build GRCh38
rs552955866 Chr.1: 167548263-167548263 on Build GRCh38
rs150526402 Chr.1: 167548084-167548084 on Build GRCh38
rs900089300 Chr.1: 167548269-167548269 on Build GRCh38
rs1023422089 Chr.1: 167548435-167548435 on Build GRCh38
rs892019807 Chr.1: 167548414-167548414 on Build GRCh38
rs190742658 Chr.1: 167548425-167548425 on Build GRCh38
rs1028023932 Chr.1: 167548473-167548473 on Build GRCh38
rs142047332 Chr.1: 167548131-167548131 on Build GRCh38
rs139296899 Chr.1: 167548249-167548249 on Build GRCh38
rs950058660 Chr.1: 167548306-167548306 on Build GRCh38
rs756673611 Chr.1: 167548088-167548088 on Build GRCh38
rs771173297 Chr.1: 167548411-167548412 on Build GRCh38
rs200702293 Chr.1: 167548129-167548129 on Build GRCh38
rs558450486 Chr.1: 167548229-167548229 on Build GRCh38
rs768500518 Chr.1: 167548012-167548012 on Build GRCh38
rs1000738647 Chr.1: 167548403-167548403 on Build GRCh38
rs764761435 Chr.1: 167548036-167548036 on Build GRCh38
rs11446137 Chr.1: 167548480-167548481 on Build GRCh38
rs775447345 Chr.1: 167547996-167547996 on Build GRCh38
rs932929050 Chr.1: 167548314-167548314 on Build GRCh38
rs74119803 Chr.1: 167548125-167548125 on Build GRCh38
rs905985740 Chr.1: 167548318-167548318 on Build GRCh38
rs772117438 Chr.1: 167547993-167547993 on Build GRCh38
rs142420831 Chr.1: 167548026-167548026 on Build GRCh38
rs895862727 Chr.1: 167547989-167547989 on Build GRCh38
rs111711324 Chr.1: 167547991-167547992 on Build GRCh38
rs552499196 Chr.1: 167548342-167548342 on Build GRCh38
rs748867487 Chr.1: 167548492-167548492 on Build GRCh38
rs373714949 Chr.1: 167548134-167548134 on Build GRCh38
rs997026400 Chr.1: 167547986-167547986 on Build GRCh38
rs753208958 Chr.1: 167548239-167548239 on Build GRCh38
rs541979271 Chr.1: 167548308-167548308 on Build GRCh38
rs574728620 Chr.1: 167548297-167548297 on Build GRCh38
rs953826382 Chr.1: 167548476-167548476 on Build GRCh38
rs375517047 Chr.1: 167548156-167548156 on Build GRCh38
rs745896297 Chr.1: 167547991-167547991 on Build GRCh38
rs1019185990 Chr.1: 167548449-167548449 on Build GRCh38
rs773681782 Chr.1: 167548381-167548381 on Build GRCh38
rs369944597 Chr.1: 167548174-167548174 on Build GRCh38
rs762596750 Chr.1: 167548065-167548065 on Build GRCh38
rs776233790 Chr.1: 167548019-167548019 on Build GRCh38
rs781368173 Chr.1: 167548111-167548111 on Build GRCh38
rs921039263 Chr.1: 167548194-167548194 on Build GRCh38
rs964998139 Chr.1: 167548453-167548453 on Build GRCh38
rs11542635 Chr.1: 167548048-167548048 on Build GRCh38
rs754082886 Chr.1: 167548096-167548096 on Build GRCh38
rs1033672610 Chr.1: 167548123-167548123 on Build GRCh38
rs764467815 Chr.1: 167548168-167548168 on Build GRCh38
rs374855258 Chr.1: 167548040-167548040 on Build GRCh38
rs1014365545 Chr.1: 167548001-167548001 on Build GRCh38
rs749966825 Chr.1: 167548063-167548063 on Build GRCh38
rs114355836 Chr.1: 167548245-167548245 on Build GRCh38
rs1004369692 Chr.1: 167548360-167548360 on Build GRCh38
rs760604408 Chr.1: 167548002-167548002 on Build GRCh38
rs970082867 Chr.1: 167548448-167548448 on Build GRCh38
rs757530382 Chr.1: 167548099-167548099 on Build GRCh38
rs771778878 Chr.1: 167548113-167548113 on Build GRCh38
rs913653519 Chr.1: 167548498-167548498 on Build GRCh38
rs772777074 Chr.1: 167548136-167548136 on Build GRCh38

View Full Product Details