Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10034386-10034884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCD2
Entrez Gene ID
2177
Gene Name
Fanconi anemia complementation group D2
Gene Aliases
FA-D2, FA4, FACD, FAD, FAD2, FANCD
UniGene
Hs.208388
Gene Chromosome Location
Chr.3: 10026387-10101937 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880913 c.247C>T p.L83L Chr.3: 10034510-10034510 on Build GRCh38
4653582 c.352G>A p.E118K Chr.3: 10034773-10034773 on Build GRCh38
6377427 c.377G>T p.S126I Chr.3: 10034798-10034798 on Build GRCh38
4930570 c.249G>C p.L83L Chr.3: 10034512-10034512 on Build GRCh38
256924 c.283G>T p.E95* Chr.3: 10034704-10034704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45593431 Chr.3: 10034815-10034815 on Build GRCh38
rs368448399 Chr.3: 10034465-10034465 on Build GRCh38
rs375506300 Chr.3: 10034514-10034514 on Build GRCh38
rs758809407 Chr.3: 10034544-10034544 on Build GRCh38
rs925093701 Chr.3: 10034523-10034523 on Build GRCh38
rs774299094 Chr.3: 10034731-10034731 on Build GRCh38
rs953809345 Chr.3: 10034446-10034446 on Build GRCh38
rs757581413 Chr.3: 10034824-10034824 on Build GRCh38
rs779406688 Chr.3: 10034645-10034645 on Build GRCh38
rs762278011 Chr.3: 10034689-10034691 on Build GRCh38
rs764507146 Chr.3: 10034797-10034797 on Build GRCh38
rs778092384 Chr.3: 10034542-10034542 on Build GRCh38
rs779309780 Chr.3: 10034831-10034831 on Build GRCh38
rs564528633 Chr.3: 10034611-10034611 on Build GRCh38
rs775474773 Chr.3: 10034543-10034544 on Build GRCh38
rs755028947 Chr.3: 10034515-10034515 on Build GRCh38
rs769771419 Chr.3: 10034665-10034665 on Build GRCh38
rs374031026 Chr.3: 10034445-10034445 on Build GRCh38
rs889869430 Chr.3: 10034557-10034557 on Build GRCh38
rs776858997 Chr.3: 10034668-10034668 on Build GRCh38
rs773138169 Chr.3: 10034669-10034669 on Build GRCh38
rs771573600 Chr.3: 10034467-10034467 on Build GRCh38
rs763311345 Chr.3: 10034855-10034856 on Build GRCh38
rs190531212 Chr.3: 10034664-10034664 on Build GRCh38
rs893386787 Chr.3: 10034629-10034629 on Build GRCh38
rs778286669 Chr.3: 10034432-10034432 on Build GRCh38
rs756313149 Chr.3: 10034786-10034786 on Build GRCh38
rs768821254 Chr.3: 10034667-10034668 on Build GRCh38
rs770835633 Chr.3: 10034490-10034490 on Build GRCh38
rs566769898 Chr.3: 10034777-10034777 on Build GRCh38
rs752880741 Chr.3: 10034534-10034534 on Build GRCh38
rs867061954 Chr.3: 10034537-10034537 on Build GRCh38
rs547064803 Chr.3: 10034852-10034852 on Build GRCh38
rs749176909 Chr.3: 10034452-10034453 on Build GRCh38
rs746511202 Chr.3: 10034658-10034658 on Build GRCh38
rs55932659 Chr.3: 10034641-10034641 on Build GRCh38
rs373904360 Chr.3: 10034427-10034427 on Build GRCh38
rs942669213 Chr.3: 10034586-10034586 on Build GRCh38
rs749849880 Chr.3: 10034550-10034550 on Build GRCh38
rs1005885079 Chr.3: 10034771-10034771 on Build GRCh38
rs376680433 Chr.3: 10034448-10034448 on Build GRCh38
rs936603288 Chr.3: 10034533-10034533 on Build GRCh38
rs202196826 Chr.3: 10034717-10034717 on Build GRCh38
rs1019208945 Chr.3: 10034855-10034855 on Build GRCh38
rs756513527 Chr.3: 10034536-10034536 on Build GRCh38
rs746739014 Chr.3: 10034460-10034460 on Build GRCh38
rs1037892058 Chr.3: 10034715-10034715 on Build GRCh38
rs371915501 Chr.3: 10034509-10034509 on Build GRCh38
rs866806142 Chr.3: 10034774-10034774 on Build GRCh38
rs1054419166 Chr.3: 10034625-10034625 on Build GRCh38
rs755150961 Chr.3: 10034420-10034420 on Build GRCh38
rs200335298 Chr.3: 10034776-10034776 on Build GRCh38
rs749767303 Chr.3: 10034441-10034441 on Build GRCh38
rs867535949 Chr.3: 10034719-10034719 on Build GRCh38
rs757697678 Chr.3: 10034555-10034555 on Build GRCh38
rs200170760 Chr.3: 10034449-10034449 on Build GRCh38
rs768164124 Chr.3: 10034659-10034659 on Build GRCh38
rs958452816 Chr.3: 10034769-10034769 on Build GRCh38
rs762742734 Chr.3: 10034684-10034684 on Build GRCh38
rs1004943548 Chr.3: 10034704-10034704 on Build GRCh38
rs868566205 Chr.3: 10034734-10034734 on Build GRCh38
rs143936557 Chr.3: 10034732-10034732 on Build GRCh38
rs954659193 Chr.3: 10034860-10034860 on Build GRCh38
rs765082165 Chr.3: 10034493-10034493 on Build GRCh38
rs773320606 Chr.3: 10034494-10034494 on Build GRCh38
rs866255711 Chr.3: 10034753-10034753 on Build GRCh38
rs371395855 Chr.3: 10034579-10034579 on Build GRCh38
rs916618083 Chr.3: 10034578-10034578 on Build GRCh38
rs761722300 Chr.3: 10034479-10034479 on Build GRCh38
rs149618265 Chr.3: 10034421-10034421 on Build GRCh38
rs748530086 Chr.3: 10034425-10034425 on Build GRCh38
rs369743385 Chr.3: 10034833-10034833 on Build GRCh38
rs762788048 Chr.3: 10034499-10034499 on Build GRCh38
rs766241610 Chr.3: 10034707-10034707 on Build GRCh38
rs767659342 Chr.3: 10034519-10034519 on Build GRCh38
rs907335908 Chr.3: 10034680-10034680 on Build GRCh38

View Full Product Details