Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 65253504-65253968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR2
Entrez Gene ID
10097
Gene Name
ARP2 actin related protein 2 homolog
Gene Aliases
ARP2
UniGene
Hs.643727
Gene Chromosome Location
Chr.2: 65227695-65271253 on Build GRCh38
Transcripts
NM_001005386.2 NM_005722.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403219 c.464G>A p.G155D Chr.2: 65253728-65253728 on Build GRCh38
7135383 c.592C>T p.L198F Chr.2: 65253856-65253856 on Build GRCh38
6403220 c.449G>A p.G150D Chr.2: 65253728-65253728 on Build GRCh38
3659924 c.565A>G p.I189V Chr.2: 65253829-65253829 on Build GRCh38
5480042 c.585+10A>C p.? Chr.2: 65253874-65253874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs268871 Chr.2: 65253874-65253874 on Build GRCh38
rs750224932 Chr.2: 65253688-65253688 on Build GRCh38
rs143267480 Chr.2: 65253609-65253609 on Build GRCh38
rs915155538 Chr.2: 65253622-65253622 on Build GRCh38
rs145726035 Chr.2: 65253829-65253829 on Build GRCh38
rs765250381 Chr.2: 65253756-65253756 on Build GRCh38
rs3217129 Chr.2: 65253880-65253881 on Build GRCh38
rs746337748 Chr.2: 65253887-65253887 on Build GRCh38
rs201196839 Chr.2: 65253876-65253877 on Build GRCh38
rs770326689 Chr.2: 65253894-65253894 on Build GRCh38
rs937820331 Chr.2: 65253576-65253576 on Build GRCh38
rs758920285 Chr.2: 65253547-65253548 on Build GRCh38
rs200556229 Chr.2: 65253738-65253738 on Build GRCh38
rs544840778 Chr.2: 65253886-65253886 on Build GRCh38
rs989341096 Chr.2: 65253575-65253575 on Build GRCh38
rs1022529652 Chr.2: 65253654-65253654 on Build GRCh38
rs775596446 Chr.2: 65253766-65253766 on Build GRCh38
rs1017570398 Chr.2: 65253540-65253540 on Build GRCh38
rs778601258 Chr.2: 65253708-65253708 on Build GRCh38
rs866989160 Chr.2: 65253833-65253833 on Build GRCh38
rs182673501 Chr.2: 65253895-65253895 on Build GRCh38
rs113106233 Chr.2: 65253548-65253552 on Build GRCh38
rs17030083 Chr.2: 65253783-65253783 on Build GRCh38
rs964614409 Chr.2: 65253561-65253561 on Build GRCh38
rs755810885 Chr.2: 65253806-65253806 on Build GRCh38
rs1042056369 Chr.2: 65253629-65253629 on Build GRCh38
rs747388020 Chr.2: 65253722-65253722 on Build GRCh38
rs746675963 Chr.2: 65253882-65253882 on Build GRCh38
rs760120471 Chr.2: 65253747-65253747 on Build GRCh38
rs771339519 Chr.2: 65253737-65253737 on Build GRCh38
rs927990253 Chr.2: 65253753-65253753 on Build GRCh38
rs756076650 Chr.2: 65253689-65253689 on Build GRCh38
rs747758062 Chr.2: 65253709-65253709 on Build GRCh38
rs1047892522 Chr.2: 65253939-65253939 on Build GRCh38
rs187796877 Chr.2: 65253937-65253937 on Build GRCh38
rs754381415 Chr.2: 65253707-65253707 on Build GRCh38
rs756439882 Chr.2: 65253940-65253940 on Build GRCh38
rs1008819060 Chr.2: 65253528-65253528 on Build GRCh38
rs201046285 Chr.2: 65253765-65253765 on Build GRCh38
rs200929514 Chr.2: 65253549-65253549 on Build GRCh38
rs945314693 Chr.2: 65253624-65253624 on Build GRCh38
rs990774220 Chr.2: 65253873-65253873 on Build GRCh38
rs989760159 Chr.2: 65253591-65253591 on Build GRCh38
rs754991965 Chr.2: 65253852-65253852 on Build GRCh38
rs760135923 Chr.2: 65253634-65253634 on Build GRCh38
rs373133141 Chr.2: 65253915-65253915 on Build GRCh38
rs764368291 Chr.2: 65253804-65253804 on Build GRCh38
rs533246838 Chr.2: 65253620-65253620 on Build GRCh38
rs753682530 Chr.2: 65253844-65253844 on Build GRCh38
rs1038523154 Chr.2: 65253901-65253901 on Build GRCh38
rs966837914 Chr.2: 65253677-65253677 on Build GRCh38
rs1015814090 Chr.2: 65253910-65253910 on Build GRCh38
rs58684918 Chr.2: 65253549-65253553 on Build GRCh38
rs778531443 Chr.2: 65253868-65253868 on Build GRCh38
rs750220184 Chr.2: 65253805-65253805 on Build GRCh38
rs749265847 Chr.2: 65253700-65253700 on Build GRCh38
rs766203615 Chr.2: 65253842-65253842 on Build GRCh38
rs879562674 Chr.2: 65253859-65253859 on Build GRCh38
rs771887992 Chr.2: 65253711-65253711 on Build GRCh38
rs17851188 Chr.2: 65253793-65253793 on Build GRCh38

View Full Product Details