Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54426957-54427534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH1
Entrez Gene ID
10175
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNIH, CNIH-1, CNIL, TGAM77
UniGene
Hs.294603
Gene Chromosome Location
Chr.14: 54426929-54441430 on Build GRCh38
Transcripts
NM_005776.2

SNP Detail

SNP ID Chromosome Location
rs887117272 Chr.14: 54427311-54427311 on Build GRCh38
rs151200006 Chr.14: 54427108-54427109 on Build GRCh38
rs896268740 Chr.14: 54427450-54427450 on Build GRCh38
rs960685949 Chr.14: 54427173-54427173 on Build GRCh38
rs1004189783 Chr.14: 54427332-54427332 on Build GRCh38
rs746790873 Chr.14: 54427438-54427438 on Build GRCh38
rs138616880 Chr.14: 54427125-54427125 on Build GRCh38
rs191170776 Chr.14: 54427487-54427487 on Build GRCh38
rs1004304421 Chr.14: 54427023-54427023 on Build GRCh38
rs1002687013 Chr.14: 54427419-54427419 on Build GRCh38
rs934301915 Chr.14: 54427289-54427289 on Build GRCh38
rs943102326 Chr.14: 54427259-54427259 on Build GRCh38
rs779887686 Chr.14: 54427412-54427412 on Build GRCh38
rs904309257 Chr.14: 54427326-54427326 on Build GRCh38
rs1026231578 Chr.14: 54427453-54427453 on Build GRCh38
rs1013510371 Chr.14: 54427458-54427458 on Build GRCh38
rs756177374 Chr.14: 54427205-54427205 on Build GRCh38
rs562162583 Chr.14: 54427401-54427401 on Build GRCh38
rs556690216 Chr.14: 54427147-54427147 on Build GRCh38
rs1016999039 Chr.14: 54427053-54427053 on Build GRCh38
rs972825176 Chr.14: 54427003-54427003 on Build GRCh38
rs970893250 Chr.14: 54427429-54427430 on Build GRCh38
rs540474402 Chr.14: 54427394-54427394 on Build GRCh38
rs767832583 Chr.14: 54427049-54427049 on Build GRCh38
rs369586916 Chr.14: 54427127-54427127 on Build GRCh38
rs887105932 Chr.14: 54427012-54427012 on Build GRCh38
rs74707690 Chr.14: 54427339-54427339 on Build GRCh38
rs372903257 Chr.14: 54427354-54427354 on Build GRCh38
rs1034046423 Chr.14: 54427336-54427336 on Build GRCh38
rs571714133 Chr.14: 54427274-54427274 on Build GRCh38
rs749273065 Chr.14: 54427296-54427296 on Build GRCh38
rs984599831 Chr.14: 54427473-54427473 on Build GRCh38
rs925008556 Chr.14: 54427254-54427254 on Build GRCh38
rs546277794 Chr.14: 54427024-54427024 on Build GRCh38
rs1021262363 Chr.14: 54427465-54427465 on Build GRCh38
rs1000343156 Chr.14: 54427329-54427329 on Build GRCh38
rs574055625 Chr.14: 54427407-54427407 on Build GRCh38
rs184763336 Chr.14: 54427505-54427505 on Build GRCh38
rs755308848 Chr.14: 54427042-54427042 on Build GRCh38
rs551455734 Chr.14: 54427183-54427183 on Build GRCh38
rs1051746498 Chr.14: 54427291-54427291 on Build GRCh38
rs1051134827 Chr.14: 54426999-54426999 on Build GRCh38
rs942682404 Chr.14: 54427493-54427496 on Build GRCh38
rs913570144 Chr.14: 54427216-54427216 on Build GRCh38
rs966366517 Chr.14: 54427497-54427497 on Build GRCh38
rs539372402 Chr.14: 54427304-54427304 on Build GRCh38
rs952861522 Chr.14: 54427468-54427468 on Build GRCh38
rs979163981 Chr.14: 54427231-54427231 on Build GRCh38
rs1012520645 Chr.14: 54427347-54427347 on Build GRCh38
rs752865236 Chr.14: 54427186-54427186 on Build GRCh38
rs898606038 Chr.14: 54427373-54427373 on Build GRCh38
rs745760205 Chr.14: 54427284-54427284 on Build GRCh38
rs182483411 Chr.14: 54427079-54427079 on Build GRCh38
rs186764033 Chr.14: 54427386-54427386 on Build GRCh38
rs778736819 Chr.14: 54427229-54427229 on Build GRCh38
rs561360079 Chr.14: 54426989-54426989 on Build GRCh38
rs1002014853 Chr.14: 54427103-54427103 on Build GRCh38

View Full Product Details