Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1089050-1089647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF8
Entrez Gene ID
390664
Gene Name
C1q and tumor necrosis factor related protein 8
Gene Aliases
CTRP8, UNQ5829
UniGene
Hs.527853
Gene Chromosome Location
Chr.16: 1088226-1096306 on Build GRCh38
Transcripts
NM_207419.3 XM_011522483.2

SNP Detail

SNP ID Chromosome Location
rs767667782 Chr.16: 1089591-1089591 on Build GRCh38
rs541206861 Chr.16: 1089424-1089424 on Build GRCh38
rs550076541 Chr.16: 1089087-1089087 on Build GRCh38
rs568278375 Chr.16: 1089583-1089583 on Build GRCh38
rs563602425 Chr.16: 1089479-1089479 on Build GRCh38
rs190971273 Chr.16: 1089202-1089202 on Build GRCh38
rs944979797 Chr.16: 1089512-1089512 on Build GRCh38
rs757084278 Chr.16: 1089550-1089550 on Build GRCh38
rs924568589 Chr.16: 1089205-1089205 on Build GRCh38
rs201198103 Chr.16: 1089088-1089093 on Build GRCh38
rs1024691822 Chr.16: 1089589-1089589 on Build GRCh38
rs934646359 Chr.16: 1089245-1089245 on Build GRCh38
rs1044491759 Chr.16: 1089490-1089490 on Build GRCh38
rs532398662 Chr.16: 1089488-1089488 on Build GRCh38
rs147500286 Chr.16: 1089432-1089432 on Build GRCh38
rs952079197 Chr.16: 1089084-1089084 on Build GRCh38
rs543312865 Chr.16: 1089457-1089457 on Build GRCh38
rs957431696 Chr.16: 1089628-1089628 on Build GRCh38
rs920670943 Chr.16: 1089135-1089135 on Build GRCh38
rs1019265498 Chr.16: 1089388-1089388 on Build GRCh38
rs1001387325 Chr.16: 1089619-1089619 on Build GRCh38
rs930822418 Chr.16: 1089493-1089493 on Build GRCh38
rs913021715 Chr.16: 1089505-1089505 on Build GRCh38
rs971201919 Chr.16: 1089304-1089304 on Build GRCh38
rs945768267 Chr.16: 1089347-1089347 on Build GRCh38
rs1028070285 Chr.16: 1089548-1089548 on Build GRCh38
rs917324956 Chr.16: 1089273-1089273 on Build GRCh38
rs917197573 Chr.16: 1089297-1089297 on Build GRCh38
rs1006193586 Chr.16: 1089226-1089226 on Build GRCh38
rs887844653 Chr.16: 1089411-1089411 on Build GRCh38
rs904305281 Chr.16: 1089558-1089558 on Build GRCh38
rs996961832 Chr.16: 1089539-1089539 on Build GRCh38
rs73494066 Chr.16: 1089116-1089116 on Build GRCh38
rs996623511 Chr.16: 1089421-1089421 on Build GRCh38
rs539032531 Chr.16: 1089094-1089094 on Build GRCh38
rs112994424 Chr.16: 1089363-1089363 on Build GRCh38
rs892016028 Chr.16: 1089168-1089168 on Build GRCh38
rs924514753 Chr.16: 1089444-1089444 on Build GRCh38
rs1057188108 Chr.16: 1089270-1089270 on Build GRCh38
rs968585055 Chr.16: 1089425-1089425 on Build GRCh38
rs569934951 Chr.16: 1089088-1089088 on Build GRCh38
rs1040679306 Chr.16: 1089180-1089180 on Build GRCh38
rs1003824099 Chr.16: 1089387-1089387 on Build GRCh38
rs927025704 Chr.16: 1089328-1089328 on Build GRCh38
rs565819406 Chr.16: 1089566-1089566 on Build GRCh38
rs572208321 Chr.16: 1089412-1089412 on Build GRCh38
rs541902318 Chr.16: 1089609-1089609 on Build GRCh38
rs1013579173 Chr.16: 1089430-1089430 on Build GRCh38
rs1034298325 Chr.16: 1089621-1089621 on Build GRCh38
rs886676160 Chr.16: 1089510-1089510 on Build GRCh38
rs532587594 Chr.16: 1089493-1089494 on Build GRCh38
rs1047914652 Chr.16: 1089506-1089506 on Build GRCh38
rs973912261 Chr.16: 1089121-1089121 on Build GRCh38
rs780954207 Chr.16: 1089579-1089579 on Build GRCh38
rs183188 Chr.16: 1089446-1089446 on Build GRCh38
rs972570780 Chr.16: 1089095-1089095 on Build GRCh38
rs917274459 Chr.16: 1089484-1089484 on Build GRCh38
rs972611289 Chr.16: 1089277-1089277 on Build GRCh38
rs367676588 Chr.16: 1089534-1089534 on Build GRCh38
rs183067593 Chr.16: 1089491-1089491 on Build GRCh38
rs441870 Chr.16: 1089274-1089274 on Build GRCh38
rs995461908 Chr.16: 1089334-1089334 on Build GRCh38
rs969822349 Chr.16: 1089200-1089200 on Build GRCh38
rs1054535045 Chr.16: 1089167-1089167 on Build GRCh38
rs528590498 Chr.16: 1089568-1089568 on Build GRCh38
rs558361708 Chr.16: 1089402-1089402 on Build GRCh38
rs902005837 Chr.16: 1089195-1089195 on Build GRCh38
rs1050597474 Chr.16: 1089398-1089398 on Build GRCh38
rs970084655 Chr.16: 1089594-1089594 on Build GRCh38
rs1047967508 Chr.16: 1089360-1089360 on Build GRCh38
rs566211336 Chr.16: 1089098-1089098 on Build GRCh38
rs992809056 Chr.16: 1089450-1089450 on Build GRCh38
rs980547022 Chr.16: 1089600-1089600 on Build GRCh38
rs1044933697 Chr.16: 1089325-1089325 on Build GRCh38
rs565101023 Chr.16: 1089361-1089362 on Build GRCh38
rs949914533 Chr.16: 1089112-1089112 on Build GRCh38
rs575903792 Chr.16: 1089324-1089324 on Build GRCh38
rs912870316 Chr.16: 1089344-1089344 on Build GRCh38
rs990291604 Chr.16: 1089503-1089503 on Build GRCh38
rs574768682 Chr.16: 1089165-1089165 on Build GRCh38
rs952236281 Chr.16: 1089217-1089217 on Build GRCh38
rs34188781 Chr.16: 1089275-1089276 on Build GRCh38
rs371649944 Chr.16: 1089406-1089406 on Build GRCh38
rs1054102387 Chr.16: 1089341-1089341 on Build GRCh38
rs1040660867 Chr.16: 1089522-1089522 on Build GRCh38
rs899343124 Chr.16: 1089332-1089332 on Build GRCh38
rs750970806 Chr.16: 1089281-1089281 on Build GRCh38
rs13333683 Chr.16: 1089582-1089582 on Build GRCh38
rs923489145 Chr.16: 1089535-1089535 on Build GRCh38
rs1010921735 Chr.16: 1089171-1089171 on Build GRCh38
rs554774971 Chr.16: 1089117-1089117 on Build GRCh38
rs71376322 Chr.16: 1089308-1089308 on Build GRCh38
rs557647969 Chr.16: 1089257-1089257 on Build GRCh38
rs930754503 Chr.16: 1089617-1089617 on Build GRCh38

View Full Product Details