Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68037920-68038372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
UniGene
Hs.534460
Gene Chromosome Location
Chr.16: 68022944-68079323 on Build GRCh38
Transcripts
NM_001271762.1 NM_001271763.1 NM_017803.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
344456 c.91G>T p.A31S Chr.16: 68038114-68038114 on Build GRCh38
471970 c.21T>A p.S7S Chr.16: 68038044-68038044 on Build GRCh38
3818318 c.11A>C p.N4T Chr.16: 68038034-68038034 on Build GRCh38
5851278 c.106G>A p.A36T Chr.16: 68038129-68038129 on Build GRCh38
6990566 c.73C>G p.L25V Chr.16: 68038096-68038096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551818375 Chr.16: 68038164-68038164 on Build GRCh38
rs183668312 Chr.16: 68037986-68037986 on Build GRCh38
rs563983494 Chr.16: 68038184-68038184 on Build GRCh38
rs778026093 Chr.16: 68038167-68038167 on Build GRCh38
rs767079041 Chr.16: 68038010-68038010 on Build GRCh38
rs569994678 Chr.16: 68038004-68038004 on Build GRCh38
rs773278927 Chr.16: 68037977-68037977 on Build GRCh38
rs775385955 Chr.16: 68038087-68038087 on Build GRCh38
rs755707118 Chr.16: 68038023-68038023 on Build GRCh38
rs75445468 Chr.16: 68037991-68037991 on Build GRCh38
rs967515491 Chr.16: 68037984-68037984 on Build GRCh38
rs1055988165 Chr.16: 68038346-68038346 on Build GRCh38
rs906626356 Chr.16: 68038319-68038319 on Build GRCh38
rs777253759 Chr.16: 68037994-68037994 on Build GRCh38
rs112586927 Chr.16: 68038291-68038292 on Build GRCh38
rs1042386675 Chr.16: 68038259-68038259 on Build GRCh38
rs1001616452 Chr.16: 68038297-68038297 on Build GRCh38
rs749421328 Chr.16: 68038295-68038295 on Build GRCh38
rs773589426 Chr.16: 68038014-68038015 on Build GRCh38
rs963403808 Chr.16: 68038107-68038107 on Build GRCh38
rs760687951 Chr.16: 68037979-68037979 on Build GRCh38
rs113202298 Chr.16: 68037997-68037997 on Build GRCh38
rs748469313 Chr.16: 68038078-68038078 on Build GRCh38
rs1009554907 Chr.16: 68037974-68037974 on Build GRCh38
rs958615905 Chr.16: 68038058-68038058 on Build GRCh38
rs774127544 Chr.16: 68038102-68038102 on Build GRCh38
rs780663427 Chr.16: 68038036-68038036 on Build GRCh38
rs943635708 Chr.16: 68038190-68038190 on Build GRCh38
rs762705423 Chr.16: 68037998-68037998 on Build GRCh38
rs919034285 Chr.16: 68038118-68038118 on Build GRCh38
rs374264321 Chr.16: 68038007-68038007 on Build GRCh38
rs770987078 Chr.16: 68038183-68038183 on Build GRCh38
rs779303777 Chr.16: 68038156-68038156 on Build GRCh38
rs758723834 Chr.16: 68038161-68038161 on Build GRCh38
rs12102597 Chr.16: 68038349-68038349 on Build GRCh38
rs192258237 Chr.16: 68037945-68037945 on Build GRCh38
rs755478505 Chr.16: 68038135-68038135 on Build GRCh38
rs377656104 Chr.16: 68038071-68038071 on Build GRCh38
rs187875252 Chr.16: 68038039-68038039 on Build GRCh38
rs768616402 Chr.16: 68037984-68037985 on Build GRCh38
rs763244911 Chr.16: 68038163-68038164 on Build GRCh38
rs200799318 Chr.16: 68038038-68038038 on Build GRCh38
rs138348659 Chr.16: 68038115-68038115 on Build GRCh38
rs929783039 Chr.16: 68038214-68038214 on Build GRCh38
rs11555012 Chr.16: 68038101-68038101 on Build GRCh38
rs868001273 Chr.16: 68038086-68038086 on Build GRCh38
rs562081425 Chr.16: 68038055-68038055 on Build GRCh38
rs374243258 Chr.16: 68038195-68038195 on Build GRCh38
rs749847774 Chr.16: 68038047-68038047 on Build GRCh38
rs915474763 Chr.16: 68038200-68038200 on Build GRCh38
rs375337192 Chr.16: 68038130-68038130 on Build GRCh38
rs749890860 Chr.16: 68038015-68038015 on Build GRCh38
rs753060459 Chr.16: 68038158-68038158 on Build GRCh38
rs569037112 Chr.16: 68037993-68037994 on Build GRCh38
rs370838996 Chr.16: 68038111-68038111 on Build GRCh38
rs112318181 Chr.16: 68038291-68038291 on Build GRCh38
rs770183977 Chr.16: 68037994-68037997 on Build GRCh38
rs199527982 Chr.16: 68038002-68038002 on Build GRCh38
rs150058704 Chr.16: 68038258-68038258 on Build GRCh38
rs992762460 Chr.16: 68037942-68037942 on Build GRCh38
rs576386610 Chr.16: 68038003-68038003 on Build GRCh38
rs547481966 Chr.16: 68038353-68038353 on Build GRCh38
rs370308611 Chr.16: 68038171-68038171 on Build GRCh38
rs893768571 Chr.16: 68037946-68037946 on Build GRCh38
rs367898219 Chr.16: 68038160-68038160 on Build GRCh38
rs568014494 Chr.16: 68038332-68038332 on Build GRCh38
rs532141248 Chr.16: 68038084-68038084 on Build GRCh38
rs1002744333 Chr.16: 68038025-68038025 on Build GRCh38
rs551925747 Chr.16: 68038085-68038085 on Build GRCh38
rs192210401 Chr.16: 68038131-68038131 on Build GRCh38
rs772281950 Chr.16: 68038081-68038081 on Build GRCh38

View Full Product Details