Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84872737-84873183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISPLD2
Entrez Gene ID
83716
Gene Name
cysteine rich secretory protein LCCL domain containing 2
Gene Aliases
CRISP11, LCRISP2
UniGene
Hs.513779
Gene Chromosome Location
Chr.16: 84819981-84909510 on Build GRCh38
Transcripts
NM_031476.3 XM_005256190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851649 c.983C>T p.S328L Chr.16: 84872993-84872993 on Build GRCh38
974479 c.1015G>A p.G339R Chr.16: 84873025-84873025 on Build GRCh38
3356870 c.1059C>T p.N353N Chr.16: 84873069-84873069 on Build GRCh38
6145259 c.996C>T p.C332C Chr.16: 84873006-84873006 on Build GRCh38
3513043 c.1080G>A p.V360V Chr.16: 84873090-84873090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775661750 Chr.16: 84873084-84873084 on Build GRCh38
rs113744408 Chr.16: 84872844-84872844 on Build GRCh38
rs753528852 Chr.16: 84873109-84873109 on Build GRCh38
rs543357038 Chr.16: 84872854-84872854 on Build GRCh38
rs1022496616 Chr.16: 84872839-84872839 on Build GRCh38
rs772999342 Chr.16: 84873150-84873150 on Build GRCh38
rs181184036 Chr.16: 84873108-84873108 on Build GRCh38
rs916485878 Chr.16: 84873093-84873093 on Build GRCh38
rs190350027 Chr.16: 84873154-84873154 on Build GRCh38
rs758348611 Chr.16: 84872948-84872948 on Build GRCh38
rs929242127 Chr.16: 84872912-84872912 on Build GRCh38
rs201353419 Chr.16: 84873088-84873088 on Build GRCh38
rs1028915603 Chr.16: 84872761-84872761 on Build GRCh38
rs781515338 Chr.16: 84873135-84873135 on Build GRCh38
rs777641847 Chr.16: 84872951-84872951 on Build GRCh38
rs762791289 Chr.16: 84873087-84873087 on Build GRCh38
rs895101485 Chr.16: 84872899-84872899 on Build GRCh38
rs536109763 Chr.16: 84873062-84873062 on Build GRCh38
rs747546737 Chr.16: 84873070-84873070 on Build GRCh38
rs749513904 Chr.16: 84873140-84873140 on Build GRCh38
rs939250902 Chr.16: 84872887-84872887 on Build GRCh38
rs953378475 Chr.16: 84872762-84872762 on Build GRCh38
rs143175101 Chr.16: 84873090-84873090 on Build GRCh38
rs779433800 Chr.16: 84872994-84872994 on Build GRCh38
rs35153102 Chr.16: 84873122-84873122 on Build GRCh38
rs200084653 Chr.16: 84872987-84872987 on Build GRCh38
rs746002704 Chr.16: 84873137-84873137 on Build GRCh38
rs976190252 Chr.16: 84872900-84872900 on Build GRCh38
rs200575316 Chr.16: 84873025-84873025 on Build GRCh38
rs990133952 Chr.16: 84872771-84872771 on Build GRCh38
rs1005058458 Chr.16: 84872935-84872935 on Build GRCh38
rs111874417 Chr.16: 84872960-84872960 on Build GRCh38
rs778484442 Chr.16: 84873069-84873069 on Build GRCh38
rs773494107 Chr.16: 84872848-84872848 on Build GRCh38
rs138955762 Chr.16: 84873063-84873063 on Build GRCh38
rs113335623 Chr.16: 84872975-84872975 on Build GRCh38
rs184915941 Chr.16: 84873010-84873010 on Build GRCh38
rs748473995 Chr.16: 84872995-84872995 on Build GRCh38
rs772736629 Chr.16: 84872997-84872997 on Build GRCh38
rs774215475 Chr.16: 84873147-84873147 on Build GRCh38
rs529520154 Chr.16: 84872798-84872798 on Build GRCh38
rs751481884 Chr.16: 84873030-84873030 on Build GRCh38
rs984645008 Chr.16: 84873116-84873116 on Build GRCh38
rs756766940 Chr.16: 84872963-84872963 on Build GRCh38
rs1048785727 Chr.16: 84872914-84872914 on Build GRCh38
rs747279745 Chr.16: 84873127-84873127 on Build GRCh38
rs968172876 Chr.16: 84873048-84873048 on Build GRCh38
rs977930822 Chr.16: 84872835-84872835 on Build GRCh38
rs752427016 Chr.16: 84872946-84872946 on Build GRCh38
rs752515073 Chr.16: 84873018-84873018 on Build GRCh38
rs887494591 Chr.16: 84872920-84872920 on Build GRCh38
rs751103537 Chr.16: 84872959-84872959 on Build GRCh38
rs776789199 Chr.16: 84873076-84873076 on Build GRCh38
rs946136808 Chr.16: 84872824-84872824 on Build GRCh38
rs760826398 Chr.16: 84872886-84872886 on Build GRCh38
rs1056390018 Chr.16: 84872890-84872890 on Build GRCh38
rs765389169 Chr.16: 84873016-84873016 on Build GRCh38
rs965704194 Chr.16: 84872898-84872898 on Build GRCh38
rs957211448 Chr.16: 84872823-84872823 on Build GRCh38
rs759853821 Chr.16: 84873138-84873138 on Build GRCh38
rs780762209 Chr.16: 84872986-84872986 on Build GRCh38
rs548735529 Chr.16: 84873041-84873041 on Build GRCh38
rs771277704 Chr.16: 84873071-84873071 on Build GRCh38
rs948517031 Chr.16: 84873143-84873143 on Build GRCh38
rs770036422 Chr.16: 84873080-84873080 on Build GRCh38
rs758901132 Chr.16: 84873114-84873114 on Build GRCh38
rs147512774 Chr.16: 84873101-84873101 on Build GRCh38
rs1048861317 Chr.16: 84872956-84872956 on Build GRCh38
rs182019495 Chr.16: 84873157-84873157 on Build GRCh38
rs759809994 Chr.16: 84873013-84873013 on Build GRCh38
rs757808459 Chr.16: 84873130-84873130 on Build GRCh38
rs990643416 Chr.16: 84872978-84872978 on Build GRCh38
rs762699158 Chr.16: 84873023-84873023 on Build GRCh38
rs560739101 Chr.16: 84872777-84872777 on Build GRCh38
rs200215115 Chr.16: 84873105-84873105 on Build GRCh38
rs138350685 Chr.16: 84873106-84873106 on Build GRCh38
rs186989358 Chr.16: 84873148-84873148 on Build GRCh38
rs150278341 Chr.16: 84873042-84873042 on Build GRCh38
rs530342677 Chr.16: 84872863-84872863 on Build GRCh38
rs1003960667 Chr.16: 84872767-84872767 on Build GRCh38
rs773822971 Chr.16: 84873001-84873001 on Build GRCh38
rs376122777 Chr.16: 84873009-84873009 on Build GRCh38
rs551430867 Chr.16: 84872938-84872938 on Build GRCh38
rs902016233 Chr.16: 84872974-84872974 on Build GRCh38
rs947682417 Chr.16: 84872911-84872911 on Build GRCh38
rs34582606 Chr.16: 84873039-84873039 on Build GRCh38
rs765984600 Chr.16: 84873155-84873155 on Build GRCh38
rs759889773 Chr.16: 84872819-84872819 on Build GRCh38
rs375824731 Chr.16: 84873007-84873007 on Build GRCh38
rs373672055 Chr.16: 84872993-84872993 on Build GRCh38
rs775753587 Chr.16: 84873139-84873139 on Build GRCh38
rs562439341 Chr.16: 84872894-84872894 on Build GRCh38
rs776785949 Chr.16: 84873012-84873012 on Build GRCh38
rs558176005 Chr.16: 84873126-84873126 on Build GRCh38
rs531310661 Chr.16: 84872902-84872902 on Build GRCh38
rs373572693 Chr.16: 84873146-84873146 on Build GRCh38

View Full Product Details